This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01042034_m1
See other ATG4C GE Assays ›
Gene Symbol:
ATG4C
Gene Name:
autophagy related 4C cysteine peptidase
Gene Aliases:
APG4-C, APG4C, AUTL1, AUTL3
Chromosome Location:
Chr.1: 62784106 - 62865270 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01042034_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATG4C
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_032852.3 NP_116241.2 2 - 3 315 178
NM_178221.2 NP_835739.1 2 - 3 267 178
XM_005271288.2 XP_005271345.1 2 - 3 258 178
XM_011542313.2 XP_011540615.1 2 - 3 289 178
XM_017002600.1 XP_016858089.1 2 - 3 289 178
XM_017002601.1 XP_016858090.1 2 - 3 267 178
GenBank mRNA AJ312234.1 - 1 - 2 120 178
AJ320169.1 - 1 - 2 124 178
AK027773.1 - 2 - 3 286 178
BC008395.1 - 2 - 3 212 178
BC018678.1 - 2 - 3 273 178
BC033024.1 - 2 - 3 274 4838543 178
BI518482.1 - 2 - 3 300 178
DB183325.1 - 2 - 3 267 178

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100008564.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01000710.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ATG4C

Panther Classification:

Molecular Function -

cysteine protease

Gene Ontology Categories:

Function(s) Process(es)

autophagosome assembly
mitophagy
C-terminal protein lipidation
proteolysis
protein targeting to membrane
autophagy
protein processing
nucleophagy
protein delipidation
cysteine-type endopeptidase activity
peptidase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR01011351.hg.1 TC01000710.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01011351.hg.1 ENST00000317868.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01011351.hg.1 ENST00000371120.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01011351.hg.1 ENST00000540025.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01011351.hg.1 NM_032852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01011351.hg.1 NM_178221.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000317868.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000317868.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000317868.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000317868.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000317868.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000317868.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000317868.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000317868.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000317868.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000317868.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000317868.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000317868.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000317868.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000317868.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000317868.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000317868.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000317868.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000317868.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000317868.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000371118.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000371118.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000371118.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000371118.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000371118.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000371118.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000371118.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000371120.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000371120.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000371120.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000371120.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000371120.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000371120.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000371120.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000371120.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000371120.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000371120.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000371120.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000371120.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000371120.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000371120.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000371120.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000371120.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000371120.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000371120.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000414558.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000414558.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000414558.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000414558.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000414558.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000443289.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000443289.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000443289.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000443289.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000443289.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 ENST00000443289.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 NM_032852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 NM_032852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 NM_032852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 NM_032852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 NM_032852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 NM_032852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 NM_032852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 NM_032852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 NM_032852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 NM_032852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 NM_032852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 NM_032852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 NM_032852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 NM_032852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 NM_032852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 NM_032852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 NM_032852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 NM_032852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 NM_032852.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 NM_178221.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 NM_178221.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 NM_178221.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 NM_178221.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 NM_178221.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 NM_178221.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 NM_178221.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 NM_178221.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 NM_178221.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 NM_178221.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 NM_178221.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 NM_178221.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 NM_178221.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 NM_178221.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 NM_178221.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 NM_178221.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 NM_178221.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008564.hg.1 NM_178221.2 Refseq
Affymetrix Human Clariom D Assay PSR0100160783.hg.1 ENST00000317868.8 Ensembl
Affymetrix Human Clariom D Assay PSR0100160783.hg.1 ENST00000371118.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100160783.hg.1 ENST00000371120.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100160783.hg.1 ENST00000443289.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100160783.hg.1 NM_032852.3 Refseq
Affymetrix Human Clariom D Assay PSR0100160783.hg.1 NM_178221.2 Refseq
Affymetrix Human Clariom D Assay PSR0100160783.hg.1 ENST00000317868.8 Ensembl
Affymetrix Human Clariom D Assay PSR0100160783.hg.1 ENST00000371118.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100160783.hg.1 ENST00000371120.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100160783.hg.1 ENST00000443289.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100160783.hg.1 NM_032852.3 Refseq
Affymetrix Human Clariom D Assay PSR0100160783.hg.1 NM_178221.2 Refseq
Affymetrix Human Clariom D Assay PSR0100160783.hg.1 TC0100008564.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000317868.8 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000317868.8 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000317868.8 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000317868.8 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000317868.8 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000317868.8 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000317868.8 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000317868.8 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000317868.8 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000317868.8 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000317868.8 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000317868.8 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000317868.8 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000317868.8 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000317868.8 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000317868.8 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000317868.8 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000317868.8 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000317868.8 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000371118.1 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000371118.1 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000371118.1 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000371118.1 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000371118.1 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000371118.1 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000371118.1 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000371120.7 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000371120.7 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000371120.7 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000371120.7 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000371120.7 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000371120.7 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000371120.7 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000371120.7 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000371120.7 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000371120.7 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000371120.7 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000371120.7 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000371120.7 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000371120.7 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000371120.7 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000371120.7 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000371120.7 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000371120.7 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000414558.2 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000414558.2 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000414558.2 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000414558.2 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000414558.2 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000443289.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000443289.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000443289.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000443289.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000443289.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 ENST00000443289.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008564.hg.1 NM_032852.3 Refseq
Affymetrix Human Clariom D Assay TC0100008564.hg.1 NM_032852.3 Refseq
Affymetrix Human Clariom D Assay TC0100008564.hg.1 NM_032852.3 Refseq
Affymetrix Human Clariom D Assay TC0100008564.hg.1 NM_032852.3 Refseq
Affymetrix Human Clariom D Assay TC0100008564.hg.1 NM_032852.3 Refseq
Affymetrix Human Clariom D Assay TC0100008564.hg.1 NM_032852.3 Refseq
Affymetrix Human Clariom D Assay TC0100008564.hg.1 NM_032852.3 Refseq
Affymetrix Human Clariom D Assay TC0100008564.hg.1 NM_032852.3 Refseq
Affymetrix Human Clariom D Assay TC0100008564.hg.1 NM_032852.3 Refseq
Affymetrix Human Clariom D Assay TC0100008564.hg.1 NM_032852.3 Refseq
Affymetrix Human Clariom D Assay TC0100008564.hg.1 NM_032852.3 Refseq
Affymetrix Human Clariom D Assay TC0100008564.hg.1 NM_032852.3 Refseq
Affymetrix Human Clariom D Assay TC0100008564.hg.1 NM_032852.3 Refseq
Affymetrix Human Clariom D Assay TC0100008564.hg.1 NM_032852.3 Refseq
Affymetrix Human Clariom D Assay TC0100008564.hg.1 NM_032852.3 Refseq
Affymetrix Human Clariom D Assay TC0100008564.hg.1 NM_032852.3 Refseq
Affymetrix Human Clariom D Assay TC0100008564.hg.1 NM_032852.3 Refseq
Affymetrix Human Clariom D Assay TC0100008564.hg.1 NM_032852.3 Refseq
Affymetrix Human Clariom D Assay TC0100008564.hg.1 NM_032852.3 Refseq
Affymetrix Human Clariom D Assay TC0100008564.hg.1 NM_178221.2 Refseq
Affymetrix Human Clariom D Assay TC0100008564.hg.1 NM_178221.2 Refseq
Affymetrix Human Clariom D Assay TC0100008564.hg.1 NM_178221.2 Refseq
Affymetrix Human Clariom D Assay TC0100008564.hg.1 NM_178221.2 Refseq
Affymetrix Human Clariom D Assay TC0100008564.hg.1 NM_178221.2 Refseq
Affymetrix Human Clariom D Assay TC0100008564.hg.1 NM_178221.2 Refseq
Affymetrix Human Clariom D Assay TC0100008564.hg.1 NM_178221.2 Refseq
Affymetrix Human Clariom D Assay TC0100008564.hg.1 NM_178221.2 Refseq
Affymetrix Human Clariom D Assay TC0100008564.hg.1 NM_178221.2 Refseq
Affymetrix Human Clariom D Assay TC0100008564.hg.1 NM_178221.2 Refseq
Affymetrix Human Clariom D Assay TC0100008564.hg.1 NM_178221.2 Refseq
Affymetrix Human Clariom D Assay TC0100008564.hg.1 NM_178221.2 Refseq
Affymetrix Human Clariom D Assay TC0100008564.hg.1 NM_178221.2 Refseq
Affymetrix Human Clariom D Assay TC0100008564.hg.1 NM_178221.2 Refseq
Affymetrix Human Clariom D Assay TC0100008564.hg.1 NM_178221.2 Refseq
Affymetrix Human Clariom D Assay TC0100008564.hg.1 NM_178221.2 Refseq
Affymetrix Human Clariom D Assay TC0100008564.hg.1 NM_178221.2 Refseq
Affymetrix Human Clariom D Assay TC0100008564.hg.1 NM_178221.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0