This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01042230_g1
See other RRP8 GE Assays ›
Gene Symbol:
RRP8
Gene Name:
ribosomal RNA processing 8, methyltransferase, homolog (yeast)
Gene Aliases:
KIAA0409, NML
Chromosome Location:
Chr.11: 6514830 - 6603678 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01042230_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RRP8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_015324.3 NP_056139.1 5 - 6 1303 96
XM_011519955.2 XP_011518257.1 5 - 6 1331 96
XM_017017423.1 XP_016872912.1 5 - 6 1380 96
GenBank mRNA AB007869.1 - 5 - 6 1179 96
BC001071.2 - 5 - 6 1234 3505288 96
D87060.1 - 5 - 6 1155 96

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1100013138.hg.1
This assay targets the same location as HTA transcript cluster(s) TC11003503.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene RRP8

Gene Ontology Categories:

Function(s) Process(es)

chromatin silencing at rDNA
transcription, DNA-templated
rRNA processing
covalent chromatin modification
methylation
cellular response to glucose starvation
regulation of transcription by glucose
positive regulation of cell cycle arrest
intrinsic apoptotic signaling pathway by p53 class mediator
protein binding
S-adenosylmethionine-dependent methyltransferase activity
methylated histone binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 ENST00000254605.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 ENST00000254605.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 ENST00000254605.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 ENST00000254605.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 ENST00000254605.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 ENST00000254605.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 ENST00000254605.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 ENST00000254605.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 ENST00000254605.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 ENST00000254605.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 ENST00000254605.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 ENST00000254605.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 ENST00000526352.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 ENST00000526352.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 ENST00000526352.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 ENST00000526352.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 ENST00000526352.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 ENST00000526352.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 ENST00000526352.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 ENST00000526352.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 ENST00000526352.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 ENST00000530762.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 ENST00000530762.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 ENST00000530762.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 ENST00000530762.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 ENST00000530762.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 ENST00000530762.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 ENST00000533907.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 ENST00000533907.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 ENST00000533907.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 ENST00000533907.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 ENST00000533907.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 ENST00000533907.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 ENST00000533907.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 ENST00000533907.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 ENST00000533907.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 ENST00000533907.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 ENST00000533907.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 ENST00000533907.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 ENST00000533907.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 ENST00000534343.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 ENST00000534343.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 ENST00000534343.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 ENST00000534343.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 ENST00000534343.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 ENST00000534343.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 NM_015324.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 NM_015324.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 NM_015324.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 NM_015324.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 NM_015324.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 NM_015324.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 NM_015324.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 NM_015324.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 NM_015324.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 NM_015324.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 NM_015324.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013138.hg.1 NM_015324.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11017095.hg.1 ENST00000254605.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11017095.hg.1 ENST00000533907.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11017095.hg.1 ENST00000534343.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11017095.hg.1 NM_015324.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11017095.hg.1 TC11003503.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR11017096.hg.1 ENST00000254605.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11017096.hg.1 ENST00000533907.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11017096.hg.1 ENST00000534343.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11017096.hg.1 NM_015324.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11017096.hg.1 TC11003503.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC1100013138.hg.1 ENST00000254605.10 Ensembl
Affymetrix Human Clariom D Assay TC1100013138.hg.1 ENST00000254605.10 Ensembl
Affymetrix Human Clariom D Assay TC1100013138.hg.1 ENST00000254605.10 Ensembl
Affymetrix Human Clariom D Assay TC1100013138.hg.1 ENST00000254605.10 Ensembl
Affymetrix Human Clariom D Assay TC1100013138.hg.1 ENST00000254605.10 Ensembl
Affymetrix Human Clariom D Assay TC1100013138.hg.1 ENST00000254605.10 Ensembl
Affymetrix Human Clariom D Assay TC1100013138.hg.1 ENST00000254605.10 Ensembl
Affymetrix Human Clariom D Assay TC1100013138.hg.1 ENST00000254605.10 Ensembl
Affymetrix Human Clariom D Assay TC1100013138.hg.1 ENST00000254605.10 Ensembl
Affymetrix Human Clariom D Assay TC1100013138.hg.1 ENST00000254605.10 Ensembl
Affymetrix Human Clariom D Assay TC1100013138.hg.1 ENST00000254605.10 Ensembl
Affymetrix Human Clariom D Assay TC1100013138.hg.1 ENST00000254605.10 Ensembl
Affymetrix Human Clariom D Assay TC1100013138.hg.1 ENST00000526352.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013138.hg.1 ENST00000526352.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013138.hg.1 ENST00000526352.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013138.hg.1 ENST00000526352.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013138.hg.1 ENST00000526352.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013138.hg.1 ENST00000526352.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013138.hg.1 ENST00000526352.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013138.hg.1 ENST00000526352.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013138.hg.1 ENST00000526352.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013138.hg.1 ENST00000530762.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013138.hg.1 ENST00000530762.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013138.hg.1 ENST00000530762.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013138.hg.1 ENST00000530762.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013138.hg.1 ENST00000530762.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013138.hg.1 ENST00000530762.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013138.hg.1 ENST00000533907.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013138.hg.1 ENST00000533907.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013138.hg.1 ENST00000533907.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013138.hg.1 ENST00000533907.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013138.hg.1 ENST00000533907.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013138.hg.1 ENST00000533907.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013138.hg.1 ENST00000533907.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013138.hg.1 ENST00000533907.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013138.hg.1 ENST00000533907.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013138.hg.1 ENST00000533907.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013138.hg.1 ENST00000533907.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013138.hg.1 ENST00000533907.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013138.hg.1 ENST00000533907.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013138.hg.1 ENST00000534343.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013138.hg.1 ENST00000534343.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013138.hg.1 ENST00000534343.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013138.hg.1 ENST00000534343.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013138.hg.1 ENST00000534343.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013138.hg.1 ENST00000534343.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013138.hg.1 NM_015324.3 Refseq
Affymetrix Human Clariom D Assay TC1100013138.hg.1 NM_015324.3 Refseq
Affymetrix Human Clariom D Assay TC1100013138.hg.1 NM_015324.3 Refseq
Affymetrix Human Clariom D Assay TC1100013138.hg.1 NM_015324.3 Refseq
Affymetrix Human Clariom D Assay TC1100013138.hg.1 NM_015324.3 Refseq
Affymetrix Human Clariom D Assay TC1100013138.hg.1 NM_015324.3 Refseq
Affymetrix Human Clariom D Assay TC1100013138.hg.1 NM_015324.3 Refseq
Affymetrix Human Clariom D Assay TC1100013138.hg.1 NM_015324.3 Refseq
Affymetrix Human Clariom D Assay TC1100013138.hg.1 NM_015324.3 Refseq
Affymetrix Human Clariom D Assay TC1100013138.hg.1 NM_015324.3 Refseq
Affymetrix Human Clariom D Assay TC1100013138.hg.1 NM_015324.3 Refseq
Affymetrix Human Clariom D Assay TC1100013138.hg.1 NM_015324.3 Refseq
Affymetrix Human Clariom D Assay PSR1100203445.hg.1 ENST00000254605.10 Ensembl
Affymetrix Human Clariom D Assay PSR1100203445.hg.1 ENST00000526352.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100203445.hg.1 ENST00000533907.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100203445.hg.1 ENST00000534343.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100203445.hg.1 NM_015324.3 Refseq
Affymetrix Human Clariom D Assay PSR1100203445.hg.1 ENST00000254605.10 Ensembl
Affymetrix Human Clariom D Assay PSR1100203445.hg.1 ENST00000526352.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100203445.hg.1 ENST00000533907.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100203445.hg.1 ENST00000534343.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100203445.hg.1 NM_015324.3 Refseq
Affymetrix Human Clariom D Assay PSR1100203445.hg.1 TC1100013138.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1100203446.hg.1 ENST00000254605.10 Ensembl
Affymetrix Human Clariom D Assay PSR1100203446.hg.1 ENST00000526352.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100203446.hg.1 ENST00000533907.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100203446.hg.1 ENST00000534343.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100203446.hg.1 NM_015324.3 Refseq
Affymetrix Human Clariom D Assay PSR1100203446.hg.1 ENST00000254605.10 Ensembl
Affymetrix Human Clariom D Assay PSR1100203446.hg.1 ENST00000526352.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100203446.hg.1 ENST00000533907.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100203446.hg.1 ENST00000534343.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100203446.hg.1 NM_015324.3 Refseq
Affymetrix Human Clariom D Assay PSR1100203446.hg.1 TC1100013138.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline