This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01042838_m1
See other TRAPPC6B GE Assays ›
Gene Symbol:
TRAPPC6B
Gene Name:
trafficking protein particle complex 6B
Gene Aliases:
TPC6
Chromosome Location:
Chr.14: 39147811 - 39170430 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01042838_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TRAPPC6B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001079537.1 NP_001073005.1 3 - 4 596 137
XM_011536409.2 XP_011534711.1 4 - 5 616 137
XM_017020964.1 XP_016876453.1 3 - 4 569 137
GenBank mRNA AK310730.1 - 3 - 4 406 137
AK315900.1 - 3 - 4 524 137
AL833179.1 - 3 - 4 480 137
BX161510.1 - 3 - 4 488 137
BX248013.1 - 3 - 4 552 137

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1400009006.hg
This assay targets the same location as HTA transcript cluster(s) TC14001064.hg

Additional Information:

This assay is in the 5'UTR region of the AK315900.1,BX248013.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

COPII vesicle coating
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR14012123.hg.1 ENST00000330149.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14012123.hg.1 ENST00000557764.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14012123.hg.1 NM_001079537.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR14012123.hg.1 TC14001064.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000330149.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000330149.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000330149.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000330149.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000330149.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000330149.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000330149.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000330149.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000330149.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000330149.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000330149.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000347691.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000347691.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000347691.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000347691.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000347691.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000347691.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000347691.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000347691.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000347691.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000347691.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000347691.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000469361.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000469361.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000469361.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000469361.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000469361.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000469361.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000469361.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000469361.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000469361.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000469361.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000469361.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000469361.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000469361.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000469361.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000469361.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000469361.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000554018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000554018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000554018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000554018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000554018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000555269.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000555269.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000555269.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000555269.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000555269.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000555269.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000555269.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000555269.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000555269.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000555269.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000556765.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000556765.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000556765.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000556765.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000556765.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000556765.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000556765.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000556765.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000557764.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000557764.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000557764.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000557764.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000557764.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000557764.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000557764.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 ENST00000557764.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 NM_001079537.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 NM_001079537.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 NM_001079537.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 NM_001079537.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 NM_001079537.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 NM_001079537.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 NM_001079537.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 NM_001079537.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 NM_001079537.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 NM_001079537.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 NM_001079537.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 NM_001079537.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 NM_001079537.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 NM_177452.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 NM_177452.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 NM_177452.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 NM_177452.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 NM_177452.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 NM_177452.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 NM_177452.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 NM_177452.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 NM_177452.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 NM_177452.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 NM_177452.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009006.hg.1 NM_177452.3 Refseq
Affymetrix Human Clariom D Assay PSR1400166300.hg.1 ENST00000330149.9 Ensembl
Affymetrix Human Clariom D Assay PSR1400166300.hg.1 ENST00000469361.5 Ensembl
Affymetrix Human Clariom D Assay PSR1400166300.hg.1 ENST00000555269.5 Ensembl
Affymetrix Human Clariom D Assay PSR1400166300.hg.1 ENST00000557764.5 Ensembl
Affymetrix Human Clariom D Assay PSR1400166300.hg.1 NM_001079537.1 Refseq
Affymetrix Human Clariom D Assay PSR1400166300.hg.1 ENST00000330149.9 Ensembl
Affymetrix Human Clariom D Assay PSR1400166300.hg.1 ENST00000469361.5 Ensembl
Affymetrix Human Clariom D Assay PSR1400166300.hg.1 ENST00000555269.5 Ensembl
Affymetrix Human Clariom D Assay PSR1400166300.hg.1 ENST00000557764.5 Ensembl
Affymetrix Human Clariom D Assay PSR1400166300.hg.1 NM_001079537.1 Refseq
Affymetrix Human Clariom D Assay PSR1400166300.hg.1 TC1400009006.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000330149.9 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000330149.9 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000330149.9 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000330149.9 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000330149.9 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000330149.9 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000330149.9 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000330149.9 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000330149.9 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000330149.9 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000330149.9 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000347691.9 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000347691.9 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000347691.9 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000347691.9 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000347691.9 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000347691.9 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000347691.9 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000347691.9 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000347691.9 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000347691.9 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000347691.9 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000469361.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000469361.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000469361.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000469361.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000469361.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000469361.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000469361.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000469361.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000469361.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000469361.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000469361.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000469361.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000469361.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000469361.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000469361.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000469361.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000554018.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000554018.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000554018.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000554018.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000554018.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000555269.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000555269.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000555269.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000555269.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000555269.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000555269.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000555269.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000555269.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000555269.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000555269.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000556765.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000556765.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000556765.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000556765.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000556765.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000556765.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000556765.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000556765.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000557764.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000557764.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000557764.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000557764.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000557764.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000557764.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000557764.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 ENST00000557764.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009006.hg.1 NM_001079537.1 Refseq
Affymetrix Human Clariom D Assay TC1400009006.hg.1 NM_001079537.1 Refseq
Affymetrix Human Clariom D Assay TC1400009006.hg.1 NM_001079537.1 Refseq
Affymetrix Human Clariom D Assay TC1400009006.hg.1 NM_001079537.1 Refseq
Affymetrix Human Clariom D Assay TC1400009006.hg.1 NM_001079537.1 Refseq
Affymetrix Human Clariom D Assay TC1400009006.hg.1 NM_001079537.1 Refseq
Affymetrix Human Clariom D Assay TC1400009006.hg.1 NM_001079537.1 Refseq
Affymetrix Human Clariom D Assay TC1400009006.hg.1 NM_001079537.1 Refseq
Affymetrix Human Clariom D Assay TC1400009006.hg.1 NM_001079537.1 Refseq
Affymetrix Human Clariom D Assay TC1400009006.hg.1 NM_001079537.1 Refseq
Affymetrix Human Clariom D Assay TC1400009006.hg.1 NM_001079537.1 Refseq
Affymetrix Human Clariom D Assay TC1400009006.hg.1 NM_001079537.1 Refseq
Affymetrix Human Clariom D Assay TC1400009006.hg.1 NM_001079537.1 Refseq
Affymetrix Human Clariom D Assay TC1400009006.hg.1 NM_177452.3 Refseq
Affymetrix Human Clariom D Assay TC1400009006.hg.1 NM_177452.3 Refseq
Affymetrix Human Clariom D Assay TC1400009006.hg.1 NM_177452.3 Refseq
Affymetrix Human Clariom D Assay TC1400009006.hg.1 NM_177452.3 Refseq
Affymetrix Human Clariom D Assay TC1400009006.hg.1 NM_177452.3 Refseq
Affymetrix Human Clariom D Assay TC1400009006.hg.1 NM_177452.3 Refseq
Affymetrix Human Clariom D Assay TC1400009006.hg.1 NM_177452.3 Refseq
Affymetrix Human Clariom D Assay TC1400009006.hg.1 NM_177452.3 Refseq
Affymetrix Human Clariom D Assay TC1400009006.hg.1 NM_177452.3 Refseq
Affymetrix Human Clariom D Assay TC1400009006.hg.1 NM_177452.3 Refseq
Affymetrix Human Clariom D Assay TC1400009006.hg.1 NM_177452.3 Refseq
Affymetrix Human Clariom D Assay TC1400009006.hg.1 NM_177452.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-f5rhb:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0