This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01043697_m1
See other KAT6B GE Assays ›
Gene Symbol:
KAT6B
Gene Name:
lysine acetyltransferase 6B
Gene Aliases:
GTPTS, MORF, MOZ2, MYST4, ZC2HC6B, qkf, querkopf
Chromosome Location:
Chr.10: 74824927 - 75032623 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01043697_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: KAT6B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001256468.1 NP_001243397.1 7 - 8 1765 140
NM_012330.3 NP_036462.2 7 - 8 1765 140
XM_005269664.2 XP_005269721.1 7 - 8 1826 140
XM_017016000.1 XP_016871489.1 7 - 8 1674 140
XM_017016001.1 XP_016871490.1 7 - 8 1538 140
XM_017016002.1 XP_016871491.1 7 - 8 1777 140
XM_017016003.1 XP_016871492.1 6 - 7 1448 140
XM_017016004.1 XP_016871493.1 7 - 8 1674 140
XM_017016005.1 XP_016871494.1 7 - 8 1674 140
GenBank mRNA AF119230.1 - 6 - 7 1379 140
AF119231.1 - 6 - 7 1379 140
AF217500.1 - 7 - 8 1552 140
BC150618.1 - 7 - 8 1525 9056988 140

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene KAT6B

Panther Classification:

Molecular Function -

histone modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

nucleosome assembly
transcription, DNA-templated
regulation of transcription, DNA-templated
histone acetylation
histone H3 acetylation
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
DNA binding
histone acetyltransferase activity
protein binding
transcription factor binding
zinc ion binding
acetyltransferase activity
protein complex binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0