| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07008336.hg.1 |
ENST00000289495.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07008336.hg.1 |
ENST00000340694.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07008336.hg.1 |
ENST00000424654.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07008336.hg.1 |
ENST00000433360.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07008336.hg.1 |
ENST00000433881.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07008336.hg.1 |
ENST00000456331.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07008336.hg.1 |
NM_001166160.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07008336.hg.1 |
NM_001166161.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07008336.hg.1 |
NM_001166162.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07008336.hg.1 |
NM_001166163.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07008336.hg.1 |
NM_017650.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07008336.hg.1 |
TC07000565.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0700158548.hg.1 |
ENST00000289495.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700158548.hg.1 |
ENST00000340694.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700158548.hg.1 |
ENST00000424654.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700158548.hg.1 |
ENST00000433360.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700158548.hg.1 |
ENST00000433881.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700158548.hg.1 |
ENST00000456331.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700158548.hg.1 |
NM_001166160.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700158548.hg.1 |
NM_001166161.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700158548.hg.1 |
NM_001166162.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700158548.hg.1 |
NM_001166163.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700158548.hg.1 |
NM_017650.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700158548.hg.1 |
ENST00000289495.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700158548.hg.1 |
ENST00000340694.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700158548.hg.1 |
ENST00000424654.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700158548.hg.1 |
ENST00000433360.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700158548.hg.1 |
ENST00000433881.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700158548.hg.1 |
ENST00000456331.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700158548.hg.1 |
NM_001166160.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700158548.hg.1 |
NM_001166161.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700158548.hg.1 |
NM_001166162.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700158548.hg.1 |
NM_001166163.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700158548.hg.1 |
NM_017650.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700158548.hg.1 |
TC0700008375.hg.1 |
Clariom D TC |