This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01044568_g1
See other ERAL1 GE Assays ›
Gene Symbol:
ERAL1
Gene Name:
Era like 12S mitochondrial rRNA chaperone 1
Gene Aliases:
CEGA, ERA, ERA-W, ERAL1A, H-ERA, HERA-A, HERA-B
Chromosome Location:
Chr.17: 28854957 - 28861067 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01044568_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ERAL1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001317985.1 NP_001304914.1 7 - 8 1034 73
NM_005702.3 NP_005693.1 7 - 8 1037 73
GenBank mRNA AB049388.1 - 7 - 8 985 73
AF082657.1 - 7 - 8 978 73
AK023342.1 - 7 - 8 978 73
AY007435.1 - 7 - 8 980 73
BC019094.2 - 7 - 8 969 4893068 73
BX537542.1 - 6 - 7 1906 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1700007399.hg.1
This assay targets the same location as HTA transcript cluster(s) TC17000332.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ERAL1

Panther Classification:

Molecular Function -

RNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

ribosomal small subunit assembly
GTPase activity
protein binding
GTP binding
rRNA binding
ribosomal small subunit binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR17004115.hg.1 ENST00000254928.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR17004115.hg.1 ENST00000461894.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR17004115.hg.1 NM_005702.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR17004115.hg.1 TC17000332.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR17004114.hg.1 ENST00000254928.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR17004114.hg.1 ENST00000461894.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR17004114.hg.1 NM_005702.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR17004114.hg.1 TC17000332.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000412138.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000412138.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000412138.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000412138.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000412138.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000412138.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000412138.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000412138.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000412138.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000461894.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000461894.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000461894.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000461894.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000461894.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000461894.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000461894.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000461894.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000461894.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000461894.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000461894.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000461894.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000471992.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000471992.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000471992.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000471992.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000471992.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000471992.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000471992.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000471992.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000471992.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000471992.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000471992.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000577942.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000577942.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000577942.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000577942.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000577942.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000577942.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000577942.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000578001.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000578001.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000578001.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000578001.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000580917.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000580917.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000580917.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000580917.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000580917.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000580917.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000580917.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000583487.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000583487.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000583487.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000583487.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR17004113.hg.1 ENST00000254928.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR17004113.hg.1 NM_005702.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR17004113.hg.1 TC17000332.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1700152743.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Clariom D Assay PSR1700152743.hg.1 ENST00000471992.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700152743.hg.1 ENST00000577942.5 Ensembl
Affymetrix Human Clariom D Assay PSR1700152743.hg.1 NM_005702.3 Refseq
Affymetrix Human Clariom D Assay PSR1700152743.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Clariom D Assay PSR1700152743.hg.1 ENST00000471992.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700152743.hg.1 ENST00000577942.5 Ensembl
Affymetrix Human Clariom D Assay PSR1700152743.hg.1 NM_005702.3 Refseq
Affymetrix Human Clariom D Assay PSR1700152743.hg.1 TC1700007399.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1700152745.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Clariom D Assay PSR1700152745.hg.1 ENST00000461894.5 Ensembl
Affymetrix Human Clariom D Assay PSR1700152745.hg.1 ENST00000471992.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700152745.hg.1 ENST00000577942.5 Ensembl
Affymetrix Human Clariom D Assay PSR1700152745.hg.1 ENST00000580917.2 Ensembl
Affymetrix Human Clariom D Assay PSR1700152745.hg.1 ENST00000583487.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700152745.hg.1 NM_005702.3 Refseq
Affymetrix Human Clariom D Assay PSR1700152745.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Clariom D Assay PSR1700152745.hg.1 ENST00000461894.5 Ensembl
Affymetrix Human Clariom D Assay PSR1700152745.hg.1 ENST00000471992.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700152745.hg.1 ENST00000577942.5 Ensembl
Affymetrix Human Clariom D Assay PSR1700152745.hg.1 ENST00000580917.2 Ensembl
Affymetrix Human Clariom D Assay PSR1700152745.hg.1 ENST00000583487.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700152745.hg.1 NM_005702.3 Refseq
Affymetrix Human Clariom D Assay PSR1700152745.hg.1 TC1700007399.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000412138.6 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000412138.6 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000412138.6 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000412138.6 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000412138.6 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000412138.6 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000412138.6 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000412138.6 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000412138.6 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000461894.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000461894.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000461894.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000461894.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000461894.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000461894.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000461894.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000461894.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000461894.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000461894.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000461894.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000461894.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000471992.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000471992.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000471992.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000471992.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000471992.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000471992.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000471992.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000471992.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000471992.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000471992.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000471992.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000577942.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000577942.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000577942.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000577942.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000577942.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000577942.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000577942.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000578001.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000578001.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000578001.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000578001.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000580917.2 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000580917.2 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000580917.2 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000580917.2 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000580917.2 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000580917.2 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000580917.2 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000583487.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000583487.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000583487.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000583487.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Clariom D Assay TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Clariom D Assay TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Clariom D Assay TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Clariom D Assay TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Clariom D Assay TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Clariom D Assay TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Clariom D Assay TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Clariom D Assay TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Clariom D Assay TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Clariom D Assay TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Clariom D Assay TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Clariom D Assay TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Clariom D Assay TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Clariom D Assay PSR1700152746.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Clariom D Assay PSR1700152746.hg.1 ENST00000461894.5 Ensembl
Affymetrix Human Clariom D Assay PSR1700152746.hg.1 ENST00000471992.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700152746.hg.1 ENST00000583487.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700152746.hg.1 NM_005702.3 Refseq
Affymetrix Human Clariom D Assay PSR1700152746.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Clariom D Assay PSR1700152746.hg.1 ENST00000461894.5 Ensembl
Affymetrix Human Clariom D Assay PSR1700152746.hg.1 ENST00000471992.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700152746.hg.1 ENST00000583487.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700152746.hg.1 NM_005702.3 Refseq
Affymetrix Human Clariom D Assay PSR1700152746.hg.1 TC1700007399.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0