This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01044786_m1
See other ELP4 GE Assays ›
Gene Symbol:
ELP4
Gene Name:
elongator acetyltransferase complex subunit 4
Gene Aliases:
AN, C11orf19, PAX6NEB, PAXNEB, dJ68P15A.1, hELP4
Chromosome Location:
Chr.11: 31509729 - 31784525 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01044786_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ELP4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001288725.1 NP_001275654.1 7 - 8 989 96
NM_001288726.1 NP_001275655.1 7 - 8 986 96
NM_019040.4 NP_061913.3 7 - 8 986 96
GenBank mRNA AK000505.1 - 7 - 8 965 96
AK304885.1 - 7 - 8 948 96
AL136677.1 - 7 - 8 943 96
BC012514.1 - 7 - 8 937 4509903 96

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1100007185.hg.1
This assay targets the same location as HTA transcript cluster(s) TC11000303.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ELP4

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription from RNA polymerase II promoter
transcription elongation from RNA polymerase II promoter
histone H3 acetylation
histone H4 acetylation
regulation of protein kinase activity
RNA polymerase II core binding
histone acetyltransferase activity
protein binding
phosphorylase kinase regulator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR11003865.hg.1 ENST00000350638.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11003865.hg.1 ENST00000379163.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11003865.hg.1 ENST00000395934.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11003865.hg.1 NM_019040.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11003865.hg.1 TC11000303.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 ENST00000350638.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 ENST00000350638.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 ENST00000350638.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 ENST00000350638.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 ENST00000350638.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 ENST00000350638.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 ENST00000350638.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 ENST00000350638.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 ENST00000350638.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 ENST00000350638.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 ENST00000350638.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 ENST00000350638.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 ENST00000350638.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 ENST00000379163.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 ENST00000379163.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 ENST00000379163.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 ENST00000379163.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 ENST00000379163.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 ENST00000379163.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 ENST00000379163.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 ENST00000379163.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 ENST00000379163.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 ENST00000379163.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 ENST00000379163.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 ENST00000379163.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 ENST00000379163.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 ENST00000395934.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 ENST00000395934.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 ENST00000395934.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 ENST00000395934.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 ENST00000395934.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 ENST00000395934.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 ENST00000395934.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 ENST00000395934.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 ENST00000395934.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 ENST00000395934.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 ENST00000395934.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 ENST00000395934.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 ENST00000395934.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 ENST00000395934.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 ENST00000474374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 ENST00000474374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 ENST00000474374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 ENST00000474374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 ENST00000474374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 ENST00000474374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 ENST00000474374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 ENST00000474374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 ENST00000474374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 NM_001288725.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 NM_001288725.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 NM_001288725.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 NM_001288725.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 NM_001288725.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 NM_001288725.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 NM_001288725.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 NM_001288725.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 NM_001288725.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 NM_001288725.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 NM_001288725.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 NM_001288725.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 NM_001288725.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 NM_001288725.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 NM_001288725.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 NM_001288726.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 NM_001288726.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 NM_001288726.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 NM_001288726.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 NM_001288726.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 NM_001288726.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 NM_001288726.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 NM_001288726.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 NM_001288726.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 NM_001288726.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 NM_001288726.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 NM_001288726.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 NM_001288726.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 NM_001288726.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 NM_001288726.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 NM_001288726.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 NM_019040.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 NM_019040.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 NM_019040.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 NM_019040.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 NM_019040.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 NM_019040.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 NM_019040.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 NM_019040.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 NM_019040.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 NM_019040.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 NM_019040.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 NM_019040.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 NM_019040.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007185.hg.1 NM_019040.4 Refseq
Affymetrix Human Clariom D Assay PSR1100150542.hg.1 ENST00000350638.9 Ensembl
Affymetrix Human Clariom D Assay PSR1100150542.hg.1 ENST00000379163.9 Ensembl
Affymetrix Human Clariom D Assay PSR1100150542.hg.1 ENST00000395934.2 Ensembl
Affymetrix Human Clariom D Assay PSR1100150542.hg.1 ENST00000474374.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100150542.hg.1 NM_001288725.1 Refseq
Affymetrix Human Clariom D Assay PSR1100150542.hg.1 NM_001288726.1 Refseq
Affymetrix Human Clariom D Assay PSR1100150542.hg.1 NM_019040.4 Refseq
Affymetrix Human Clariom D Assay PSR1100150542.hg.1 ENST00000350638.9 Ensembl
Affymetrix Human Clariom D Assay PSR1100150542.hg.1 ENST00000379163.9 Ensembl
Affymetrix Human Clariom D Assay PSR1100150542.hg.1 ENST00000395934.2 Ensembl
Affymetrix Human Clariom D Assay PSR1100150542.hg.1 ENST00000474374.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100150542.hg.1 NM_001288725.1 Refseq
Affymetrix Human Clariom D Assay PSR1100150542.hg.1 NM_001288726.1 Refseq
Affymetrix Human Clariom D Assay PSR1100150542.hg.1 NM_019040.4 Refseq
Affymetrix Human Clariom D Assay PSR1100150542.hg.1 TC1100007185.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1100007185.hg.1 ENST00000350638.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007185.hg.1 ENST00000350638.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007185.hg.1 ENST00000350638.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007185.hg.1 ENST00000350638.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007185.hg.1 ENST00000350638.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007185.hg.1 ENST00000350638.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007185.hg.1 ENST00000350638.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007185.hg.1 ENST00000350638.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007185.hg.1 ENST00000350638.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007185.hg.1 ENST00000350638.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007185.hg.1 ENST00000350638.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007185.hg.1 ENST00000350638.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007185.hg.1 ENST00000350638.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007185.hg.1 ENST00000379163.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007185.hg.1 ENST00000379163.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007185.hg.1 ENST00000379163.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007185.hg.1 ENST00000379163.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007185.hg.1 ENST00000379163.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007185.hg.1 ENST00000379163.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007185.hg.1 ENST00000379163.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007185.hg.1 ENST00000379163.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007185.hg.1 ENST00000379163.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007185.hg.1 ENST00000379163.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007185.hg.1 ENST00000379163.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007185.hg.1 ENST00000379163.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007185.hg.1 ENST00000379163.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007185.hg.1 ENST00000395934.2 Ensembl
Affymetrix Human Clariom D Assay TC1100007185.hg.1 ENST00000395934.2 Ensembl
Affymetrix Human Clariom D Assay TC1100007185.hg.1 ENST00000395934.2 Ensembl
Affymetrix Human Clariom D Assay TC1100007185.hg.1 ENST00000395934.2 Ensembl
Affymetrix Human Clariom D Assay TC1100007185.hg.1 ENST00000395934.2 Ensembl
Affymetrix Human Clariom D Assay TC1100007185.hg.1 ENST00000395934.2 Ensembl
Affymetrix Human Clariom D Assay TC1100007185.hg.1 ENST00000395934.2 Ensembl
Affymetrix Human Clariom D Assay TC1100007185.hg.1 ENST00000395934.2 Ensembl
Affymetrix Human Clariom D Assay TC1100007185.hg.1 ENST00000395934.2 Ensembl
Affymetrix Human Clariom D Assay TC1100007185.hg.1 ENST00000395934.2 Ensembl
Affymetrix Human Clariom D Assay TC1100007185.hg.1 ENST00000395934.2 Ensembl
Affymetrix Human Clariom D Assay TC1100007185.hg.1 ENST00000395934.2 Ensembl
Affymetrix Human Clariom D Assay TC1100007185.hg.1 ENST00000395934.2 Ensembl
Affymetrix Human Clariom D Assay TC1100007185.hg.1 ENST00000395934.2 Ensembl
Affymetrix Human Clariom D Assay TC1100007185.hg.1 ENST00000474374.5 Ensembl
Affymetrix Human Clariom D Assay TC1100007185.hg.1 ENST00000474374.5 Ensembl
Affymetrix Human Clariom D Assay TC1100007185.hg.1 ENST00000474374.5 Ensembl
Affymetrix Human Clariom D Assay TC1100007185.hg.1 ENST00000474374.5 Ensembl
Affymetrix Human Clariom D Assay TC1100007185.hg.1 ENST00000474374.5 Ensembl
Affymetrix Human Clariom D Assay TC1100007185.hg.1 ENST00000474374.5 Ensembl
Affymetrix Human Clariom D Assay TC1100007185.hg.1 ENST00000474374.5 Ensembl
Affymetrix Human Clariom D Assay TC1100007185.hg.1 ENST00000474374.5 Ensembl
Affymetrix Human Clariom D Assay TC1100007185.hg.1 ENST00000474374.5 Ensembl
Affymetrix Human Clariom D Assay TC1100007185.hg.1 NM_001288725.1 Refseq
Affymetrix Human Clariom D Assay TC1100007185.hg.1 NM_001288725.1 Refseq
Affymetrix Human Clariom D Assay TC1100007185.hg.1 NM_001288725.1 Refseq
Affymetrix Human Clariom D Assay TC1100007185.hg.1 NM_001288725.1 Refseq
Affymetrix Human Clariom D Assay TC1100007185.hg.1 NM_001288725.1 Refseq
Affymetrix Human Clariom D Assay TC1100007185.hg.1 NM_001288725.1 Refseq
Affymetrix Human Clariom D Assay TC1100007185.hg.1 NM_001288725.1 Refseq
Affymetrix Human Clariom D Assay TC1100007185.hg.1 NM_001288725.1 Refseq
Affymetrix Human Clariom D Assay TC1100007185.hg.1 NM_001288725.1 Refseq
Affymetrix Human Clariom D Assay TC1100007185.hg.1 NM_001288725.1 Refseq
Affymetrix Human Clariom D Assay TC1100007185.hg.1 NM_001288725.1 Refseq
Affymetrix Human Clariom D Assay TC1100007185.hg.1 NM_001288725.1 Refseq
Affymetrix Human Clariom D Assay TC1100007185.hg.1 NM_001288725.1 Refseq
Affymetrix Human Clariom D Assay TC1100007185.hg.1 NM_001288725.1 Refseq
Affymetrix Human Clariom D Assay TC1100007185.hg.1 NM_001288725.1 Refseq
Affymetrix Human Clariom D Assay TC1100007185.hg.1 NM_001288726.1 Refseq
Affymetrix Human Clariom D Assay TC1100007185.hg.1 NM_001288726.1 Refseq
Affymetrix Human Clariom D Assay TC1100007185.hg.1 NM_001288726.1 Refseq
Affymetrix Human Clariom D Assay TC1100007185.hg.1 NM_001288726.1 Refseq
Affymetrix Human Clariom D Assay TC1100007185.hg.1 NM_001288726.1 Refseq
Affymetrix Human Clariom D Assay TC1100007185.hg.1 NM_001288726.1 Refseq
Affymetrix Human Clariom D Assay TC1100007185.hg.1 NM_001288726.1 Refseq
Affymetrix Human Clariom D Assay TC1100007185.hg.1 NM_001288726.1 Refseq
Affymetrix Human Clariom D Assay TC1100007185.hg.1 NM_001288726.1 Refseq
Affymetrix Human Clariom D Assay TC1100007185.hg.1 NM_001288726.1 Refseq
Affymetrix Human Clariom D Assay TC1100007185.hg.1 NM_001288726.1 Refseq
Affymetrix Human Clariom D Assay TC1100007185.hg.1 NM_001288726.1 Refseq
Affymetrix Human Clariom D Assay TC1100007185.hg.1 NM_001288726.1 Refseq
Affymetrix Human Clariom D Assay TC1100007185.hg.1 NM_001288726.1 Refseq
Affymetrix Human Clariom D Assay TC1100007185.hg.1 NM_001288726.1 Refseq
Affymetrix Human Clariom D Assay TC1100007185.hg.1 NM_001288726.1 Refseq
Affymetrix Human Clariom D Assay TC1100007185.hg.1 NM_019040.4 Refseq
Affymetrix Human Clariom D Assay TC1100007185.hg.1 NM_019040.4 Refseq
Affymetrix Human Clariom D Assay TC1100007185.hg.1 NM_019040.4 Refseq
Affymetrix Human Clariom D Assay TC1100007185.hg.1 NM_019040.4 Refseq
Affymetrix Human Clariom D Assay TC1100007185.hg.1 NM_019040.4 Refseq
Affymetrix Human Clariom D Assay TC1100007185.hg.1 NM_019040.4 Refseq
Affymetrix Human Clariom D Assay TC1100007185.hg.1 NM_019040.4 Refseq
Affymetrix Human Clariom D Assay TC1100007185.hg.1 NM_019040.4 Refseq
Affymetrix Human Clariom D Assay TC1100007185.hg.1 NM_019040.4 Refseq
Affymetrix Human Clariom D Assay TC1100007185.hg.1 NM_019040.4 Refseq
Affymetrix Human Clariom D Assay TC1100007185.hg.1 NM_019040.4 Refseq
Affymetrix Human Clariom D Assay TC1100007185.hg.1 NM_019040.4 Refseq
Affymetrix Human Clariom D Assay TC1100007185.hg.1 NM_019040.4 Refseq
Affymetrix Human Clariom D Assay TC1100007185.hg.1 NM_019040.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0