This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01045501_m1
See other PRSS22 GE Assays ›
Gene Symbol:
PRSS22
Gene Name:
protease, serine 22
Gene Aliases:
BSSP-4, SP001LA, hBSSP-4
Chromosome Location:
Chr.16: 2852727 - 2858630 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01045501_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PRSS22
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_022119.3 NP_071402.1 1 - 2 151 119
XM_005255473.3 XP_005255530.1 2 - 3 250 119
GenBank mRNA AB010779.1 - 1 - 2 88 119
AF321182.1 - 1 - 2 102 119
AY358396.1 - 1 - 2 137 119
BC009726.1 - 1 - 2 123 3899480 119
BQ893064.1 - 159 119

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1600009147.hg.1
This assay targets the same location as HTA transcript cluster(s) TC16000795.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PRSS22

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
serine-type endopeptidase activity
serine-type peptidase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR16011641.hg.1 ENST00000161006.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16011641.hg.1 NM_022119.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16011641.hg.1 TC16000795.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR16011642.hg.1 ENST00000161006.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16011642.hg.1 NM_022119.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16011642.hg.1 TC16000795.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR16011640.hg.1 ENST00000161006.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16011640.hg.1 NM_022119.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16011640.hg.1 TC16000795.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000161006.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000161006.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000161006.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000161006.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000161006.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000161006.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000161006.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000161006.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000161006.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000161006.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000161006.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000161006.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000161006.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000161006.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000570950.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000570950.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000570950.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000570950.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000570950.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000570950.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000570950.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000570950.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000571228.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000571228.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000571228.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000571228.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000571228.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000571228.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000571228.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000571228.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000571228.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000572061.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000572061.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000572061.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000572061.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000572061.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000572061.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000572061.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000572061.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000574768.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000574768.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000574768.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000574768.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000574768.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000574768.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000574768.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000575164.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000575164.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000575164.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000575164.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000575164.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000575164.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000575164.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000575164.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000575164.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000575164.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000575164.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000576381.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000576381.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000576381.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000576381.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000576381.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000576381.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000576381.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000576381.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000576381.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000576381.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000576381.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000577177.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000577177.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000577177.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000577177.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 ENST00000577177.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 NM_022119.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 NM_022119.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 NM_022119.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 NM_022119.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 NM_022119.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 NM_022119.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 NM_022119.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 NM_022119.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 NM_022119.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 NM_022119.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 NM_022119.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 NM_022119.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 NM_022119.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009147.hg.1 NM_022119.3 Refseq
Affymetrix Human Clariom D Assay PSR1600169678.hg.1 ENST00000161006.7 Ensembl
Affymetrix Human Clariom D Assay PSR1600169678.hg.1 ENST00000571228.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600169678.hg.1 ENST00000574768.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600169678.hg.1 NM_022119.3 Refseq
Affymetrix Human Clariom D Assay PSR1600169678.hg.1 ENST00000161006.7 Ensembl
Affymetrix Human Clariom D Assay PSR1600169678.hg.1 ENST00000571228.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600169678.hg.1 ENST00000574768.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600169678.hg.1 NM_022119.3 Refseq
Affymetrix Human Clariom D Assay PSR1600169678.hg.1 TC1600009147.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1600169672.hg.1 ENST00000161006.7 Ensembl
Affymetrix Human Clariom D Assay PSR1600169672.hg.1 ENST00000570950.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600169672.hg.1 ENST00000571228.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600169672.hg.1 ENST00000574768.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600169672.hg.1 ENST00000576381.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600169672.hg.1 ENST00000577177.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600169672.hg.1 NM_022119.3 Refseq
Affymetrix Human Clariom D Assay PSR1600169672.hg.1 ENST00000161006.7 Ensembl
Affymetrix Human Clariom D Assay PSR1600169672.hg.1 ENST00000570950.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600169672.hg.1 ENST00000571228.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600169672.hg.1 ENST00000574768.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600169672.hg.1 ENST00000576381.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600169672.hg.1 ENST00000577177.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600169672.hg.1 NM_022119.3 Refseq
Affymetrix Human Clariom D Assay PSR1600169672.hg.1 TC1600009147.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000161006.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000161006.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000161006.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000161006.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000161006.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000161006.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000161006.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000161006.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000161006.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000161006.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000161006.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000161006.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000161006.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000161006.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000570950.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000570950.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000570950.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000570950.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000570950.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000570950.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000570950.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000570950.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000571228.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000571228.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000571228.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000571228.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000571228.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000571228.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000571228.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000571228.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000571228.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000572061.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000572061.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000572061.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000572061.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000572061.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000572061.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000572061.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000572061.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000574768.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000574768.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000574768.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000574768.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000574768.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000574768.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000574768.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000575164.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000575164.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000575164.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000575164.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000575164.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000575164.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000575164.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000575164.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000575164.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000575164.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000575164.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000576381.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000576381.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000576381.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000576381.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000576381.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000576381.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000576381.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000576381.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000576381.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000576381.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000576381.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000577177.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000577177.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000577177.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000577177.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 ENST00000577177.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009147.hg.1 NM_022119.3 Refseq
Affymetrix Human Clariom D Assay TC1600009147.hg.1 NM_022119.3 Refseq
Affymetrix Human Clariom D Assay TC1600009147.hg.1 NM_022119.3 Refseq
Affymetrix Human Clariom D Assay TC1600009147.hg.1 NM_022119.3 Refseq
Affymetrix Human Clariom D Assay TC1600009147.hg.1 NM_022119.3 Refseq
Affymetrix Human Clariom D Assay TC1600009147.hg.1 NM_022119.3 Refseq
Affymetrix Human Clariom D Assay TC1600009147.hg.1 NM_022119.3 Refseq
Affymetrix Human Clariom D Assay TC1600009147.hg.1 NM_022119.3 Refseq
Affymetrix Human Clariom D Assay TC1600009147.hg.1 NM_022119.3 Refseq
Affymetrix Human Clariom D Assay TC1600009147.hg.1 NM_022119.3 Refseq
Affymetrix Human Clariom D Assay TC1600009147.hg.1 NM_022119.3 Refseq
Affymetrix Human Clariom D Assay TC1600009147.hg.1 NM_022119.3 Refseq
Affymetrix Human Clariom D Assay TC1600009147.hg.1 NM_022119.3 Refseq
Affymetrix Human Clariom D Assay TC1600009147.hg.1 NM_022119.3 Refseq
Affymetrix Human Clariom D Assay PSR1600169677.hg.1 ENST00000161006.7 Ensembl
Affymetrix Human Clariom D Assay PSR1600169677.hg.1 ENST00000571228.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600169677.hg.1 ENST00000574768.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600169677.hg.1 NM_022119.3 Refseq
Affymetrix Human Clariom D Assay PSR1600169677.hg.1 ENST00000161006.7 Ensembl
Affymetrix Human Clariom D Assay PSR1600169677.hg.1 ENST00000571228.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600169677.hg.1 ENST00000574768.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600169677.hg.1 NM_022119.3 Refseq
Affymetrix Human Clariom D Assay PSR1600169677.hg.1 TC1600009147.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0