This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01045830_g1
See other SMG8 GE Assays ›
Gene Symbol:
SMG8
Gene Name:
SMG8, nonsense mediated mRNA decay factor
Gene Aliases:
C17orf71
Chromosome Location:
Chr.17: 59210010 - 59215250 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01045830_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SMG8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_018149.6 NP_060619.4 2 - 3 1945 129
GenBank mRNA AF349467.1 - 2 - 3 1945 129
AK001449.1 - 2 - 3 1094 129
AK026858.1 - 2 - 3 1918 129
AL834490.1 - 2 - 3 1924 129
BC020957.1 - 2 - 3 1113 3851898 129

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1700012286.hg.1
This assay targets the same location as HTA transcript cluster(s) TC17000719.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SMG8

Gene Ontology Categories:

Function(s) Process(es)

nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
regulation of protein kinase activity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR17009387.hg.1 ENST00000300917.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR17009387.hg.1 ENST00000543872.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR17009387.hg.1 NM_018149.6 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR17009387.hg.1 TC17000719.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000300917.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000300917.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000300917.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000300917.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000300917.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000300917.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000300917.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000300917.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000300917.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000300917.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000300917.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000300917.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000300917.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000300917.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000543872.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000543872.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000543872.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000543872.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000543872.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000543872.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000543872.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000543872.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000543872.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000543872.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000543872.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000543872.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000543872.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000543872.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000543872.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000543872.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000578922.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000578922.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000578922.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000578922.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000578922.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000578922.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000578922.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000578922.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000578922.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000580498.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000580498.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000580498.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000580498.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000580498.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000580498.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000580798.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000580798.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000580798.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000580798.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000582469.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000582469.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000582469.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000582469.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000582469.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000582469.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 ENST00000582469.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 NM_018149.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 NM_018149.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 NM_018149.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 NM_018149.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 NM_018149.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 NM_018149.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 NM_018149.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 NM_018149.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 NM_018149.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 NM_018149.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 NM_018149.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 NM_018149.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 NM_018149.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012286.hg.1 NM_018149.6 Refseq
Affymetrix Human Clariom D Assay PSR1700203316.hg.1 ENST00000300917.9 Ensembl
Affymetrix Human Clariom D Assay PSR1700203316.hg.1 ENST00000543872.6 Ensembl
Affymetrix Human Clariom D Assay PSR1700203316.hg.1 NM_018149.6 Refseq
Affymetrix Human Clariom D Assay PSR1700203316.hg.1 TC1700012286.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1700203316.hg.1 ENST00000300917.9 Ensembl
Affymetrix Human Clariom D Assay PSR1700203316.hg.1 ENST00000543872.6 Ensembl
Affymetrix Human Clariom D Assay PSR1700203316.hg.1 NM_018149.6 Refseq
Affymetrix Human Clariom D Assay PSR1700203315.hg.1 ENST00000300917.9 Ensembl
Affymetrix Human Clariom D Assay PSR1700203315.hg.1 ENST00000543872.6 Ensembl
Affymetrix Human Clariom D Assay PSR1700203315.hg.1 ENST00000580498.5 Ensembl
Affymetrix Human Clariom D Assay PSR1700203315.hg.1 NM_018149.6 Refseq
Affymetrix Human Clariom D Assay PSR1700203315.hg.1 TC1700012286.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1700203315.hg.1 ENST00000300917.9 Ensembl
Affymetrix Human Clariom D Assay PSR1700203315.hg.1 ENST00000543872.6 Ensembl
Affymetrix Human Clariom D Assay PSR1700203315.hg.1 ENST00000580498.5 Ensembl
Affymetrix Human Clariom D Assay PSR1700203315.hg.1 NM_018149.6 Refseq
Affymetrix Human Clariom D Assay PSR1700203313.hg.1 ENST00000300917.9 Ensembl
Affymetrix Human Clariom D Assay PSR1700203313.hg.1 ENST00000543872.6 Ensembl
Affymetrix Human Clariom D Assay PSR1700203313.hg.1 ENST00000580498.5 Ensembl
Affymetrix Human Clariom D Assay PSR1700203313.hg.1 ENST00000580798.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700203313.hg.1 ENST00000582469.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700203313.hg.1 NM_018149.6 Refseq
Affymetrix Human Clariom D Assay PSR1700203313.hg.1 TC1700012286.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1700203313.hg.1 ENST00000300917.9 Ensembl
Affymetrix Human Clariom D Assay PSR1700203313.hg.1 ENST00000543872.6 Ensembl
Affymetrix Human Clariom D Assay PSR1700203313.hg.1 ENST00000580498.5 Ensembl
Affymetrix Human Clariom D Assay PSR1700203313.hg.1 ENST00000580798.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700203313.hg.1 ENST00000582469.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700203313.hg.1 NM_018149.6 Refseq
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000300917.9 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000300917.9 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000300917.9 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000300917.9 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000300917.9 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000300917.9 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000300917.9 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000300917.9 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000300917.9 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000300917.9 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000300917.9 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000300917.9 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000300917.9 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000300917.9 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000543872.6 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000543872.6 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000543872.6 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000543872.6 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000543872.6 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000543872.6 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000543872.6 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000543872.6 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000543872.6 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000543872.6 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000543872.6 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000543872.6 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000543872.6 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000543872.6 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000543872.6 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000543872.6 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000578922.1 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000578922.1 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000578922.1 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000578922.1 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000578922.1 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000578922.1 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000578922.1 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000578922.1 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000578922.1 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000580498.5 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000580498.5 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000580498.5 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000580498.5 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000580498.5 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000580498.5 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000580798.1 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000580798.1 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000580798.1 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000580798.1 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000582469.1 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000582469.1 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000582469.1 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000582469.1 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000582469.1 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000582469.1 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 ENST00000582469.1 Ensembl
Affymetrix Human Clariom D Assay TC1700012286.hg.1 NM_018149.6 Refseq
Affymetrix Human Clariom D Assay TC1700012286.hg.1 NM_018149.6 Refseq
Affymetrix Human Clariom D Assay TC1700012286.hg.1 NM_018149.6 Refseq
Affymetrix Human Clariom D Assay TC1700012286.hg.1 NM_018149.6 Refseq
Affymetrix Human Clariom D Assay TC1700012286.hg.1 NM_018149.6 Refseq
Affymetrix Human Clariom D Assay TC1700012286.hg.1 NM_018149.6 Refseq
Affymetrix Human Clariom D Assay TC1700012286.hg.1 NM_018149.6 Refseq
Affymetrix Human Clariom D Assay TC1700012286.hg.1 NM_018149.6 Refseq
Affymetrix Human Clariom D Assay TC1700012286.hg.1 NM_018149.6 Refseq
Affymetrix Human Clariom D Assay TC1700012286.hg.1 NM_018149.6 Refseq
Affymetrix Human Clariom D Assay TC1700012286.hg.1 NM_018149.6 Refseq
Affymetrix Human Clariom D Assay TC1700012286.hg.1 NM_018149.6 Refseq
Affymetrix Human Clariom D Assay TC1700012286.hg.1 NM_018149.6 Refseq
Affymetrix Human Clariom D Assay TC1700012286.hg.1 NM_018149.6 Refseq
Affymetrix Human Clariom D Assay PSR1700203314.hg.1 ENST00000300917.9 Ensembl
Affymetrix Human Clariom D Assay PSR1700203314.hg.1 ENST00000543872.6 Ensembl
Affymetrix Human Clariom D Assay PSR1700203314.hg.1 ENST00000580498.5 Ensembl
Affymetrix Human Clariom D Assay PSR1700203314.hg.1 ENST00000582469.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700203314.hg.1 NM_018149.6 Refseq
Affymetrix Human Clariom D Assay PSR1700203314.hg.1 TC1700012286.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1700203314.hg.1 ENST00000300917.9 Ensembl
Affymetrix Human Clariom D Assay PSR1700203314.hg.1 ENST00000543872.6 Ensembl
Affymetrix Human Clariom D Assay PSR1700203314.hg.1 ENST00000580498.5 Ensembl
Affymetrix Human Clariom D Assay PSR1700203314.hg.1 ENST00000582469.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700203314.hg.1 NM_018149.6 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline