This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01045889_m1
See other MSI1 GE Assays ›
Gene Symbol:
MSI1
Gene Name:
musashi RNA binding protein 1
Gene Aliases:
-
Chromosome Location:
Chr.12: 120339655 - 120369180 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01045889_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MSI1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_002442.3 NP_002433.1 4 - 5 354 106
XM_006719403.3 XP_006719466.1 4 - 5 348 106
XM_011538361.2 XP_011536663.1 4 - 5 348 106
XM_011538362.2 XP_011536664.1 4 - 5 348 106
XM_011538363.2 XP_011536665.1 4 - 5 348 106
GenBank mRNA AB012851.1 - 4 - 5 329 106
AB072590.1 - 1 - 2 76 106
AB072591.1 - 1 - 2 76 106
AB072592.1 - 1 - 2 76 106
AB073212.1 - 1 - 2 69 106

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1200012159.hg.1
This assay targets the same location as HTA transcript cluster(s) TC12002043.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MSI1

Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

nervous system development
nucleotide binding
RNA binding
poly(U) RNA binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR12026846.hg.1 ENST00000257552.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12026846.hg.1 NM_002442.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12026846.hg.1 TC12002043.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000546622.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000546622.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000546622.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000546622.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000546622.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000546985.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000546985.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000546985.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000546985.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000546985.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000546985.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000546985.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000546985.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000546985.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Clariom D Assay PSR1200194834.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Clariom D Assay PSR1200194834.hg.1 ENST00000546985.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200194834.hg.1 NM_002442.3 Refseq
Affymetrix Human Clariom D Assay PSR1200194834.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Clariom D Assay PSR1200194834.hg.1 ENST00000546985.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200194834.hg.1 NM_002442.3 Refseq
Affymetrix Human Clariom D Assay PSR1200194834.hg.1 TC1200012159.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000546622.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000546622.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000546622.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000546622.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000546622.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000546985.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000546985.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000546985.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000546985.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000546985.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000546985.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000546985.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000546985.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000546985.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Clariom D Assay TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Clariom D Assay TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Clariom D Assay TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Clariom D Assay TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Clariom D Assay TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Clariom D Assay TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Clariom D Assay TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Clariom D Assay TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Clariom D Assay TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Clariom D Assay TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Clariom D Assay TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Clariom D Assay TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Clariom D Assay TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Clariom D Assay TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Clariom D Assay TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Clariom D Assay TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Clariom D Assay TC1200012159.hg.1 NM_002442.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0