This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01045892_m1
See other MSI1 GE Assays ›
Gene Symbol:
MSI1
Gene Name:
musashi RNA binding protein 1
Gene Aliases:
-
Chromosome Location:
Chr.12: 120339655 - 120369180 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01045892_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MSI1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_002442.3 NP_002433.1 7 - 8 538 77
XM_006719403.3 XP_006719466.1 7 - 8 532 77
XM_006719404.3 XP_006719467.1 2 - 3 317 77
XM_011538361.2 XP_011536663.1 7 - 8 532 77
XM_011538362.2 XP_011536664.1 7 - 8 532 77
XM_011538363.2 XP_011536665.1 7 - 8 532 77
XM_011538364.2 XP_011536666.1 3 - 4 1082 77
XM_011538365.2 XP_011536667.1 4 - 5 201 77
XM_011538366.2 XP_011536668.1 4 - 5 1209 77
XM_011538368.2 XP_011536670.1 2 - 3 157 77
XM_011538370.2 XP_011536672.1 2 - 3 83 77
XM_011538371.2 XP_011536673.1 2 - 3 109 77
GenBank mRNA AB012851.1 - 7 - 8 513 77
AB072590.1 - 4 - 5 260 77
AB072591.1 - 4 - 5 260 77
AB072592.1 - 4 - 5 260 77
AB073212.1 - 4 - 5 253 77
BX393884.2 - 7 - 8 538 77

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1200012159.hg.1
This assay targets the same location as HTA transcript cluster(s) TC12002043.hg.1

Additional Information:

This assay is in the 5'UTR region of the XM_006719404.3,XM_011538368.2,XM_011538370.2,XM_011538371.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MSI1

Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

nervous system development
nucleotide binding
RNA binding
poly(U) RNA binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000546622.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000546622.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000546622.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000546622.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000546622.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000546985.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000546985.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000546985.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000546985.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000546985.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000546985.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000546985.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000546985.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 ENST00000546985.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12026841.hg.1 ENST00000257552.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12026841.hg.1 ENST00000546622.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12026841.hg.1 NM_002442.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12026841.hg.1 TC12002043.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR12026842.hg.1 ENST00000257552.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12026842.hg.1 ENST00000546622.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12026842.hg.1 NM_002442.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12026842.hg.1 TC12002043.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000546622.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000546622.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000546622.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000546622.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000546622.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000546985.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000546985.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000546985.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000546985.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000546985.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000546985.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000546985.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000546985.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 ENST00000546985.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Clariom D Assay TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Clariom D Assay TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Clariom D Assay TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Clariom D Assay TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Clariom D Assay TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Clariom D Assay TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Clariom D Assay TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Clariom D Assay TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Clariom D Assay TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Clariom D Assay TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Clariom D Assay TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Clariom D Assay TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Clariom D Assay TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Clariom D Assay TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Clariom D Assay TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Clariom D Assay TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Clariom D Assay TC1200012159.hg.1 NM_002442.3 Refseq
Affymetrix Human Clariom D Assay PSR1200194825.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Clariom D Assay PSR1200194825.hg.1 ENST00000546622.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200194825.hg.1 ENST00000546985.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200194825.hg.1 NM_002442.3 Refseq
Affymetrix Human Clariom D Assay PSR1200194825.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Clariom D Assay PSR1200194825.hg.1 ENST00000546622.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200194825.hg.1 ENST00000546985.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200194825.hg.1 NM_002442.3 Refseq
Affymetrix Human Clariom D Assay PSR1200194825.hg.1 TC1200012159.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1200194829.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Clariom D Assay PSR1200194829.hg.1 ENST00000546622.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200194829.hg.1 ENST00000546985.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200194829.hg.1 NM_002442.3 Refseq
Affymetrix Human Clariom D Assay PSR1200194829.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Clariom D Assay PSR1200194829.hg.1 ENST00000546622.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200194829.hg.1 ENST00000546985.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200194829.hg.1 NM_002442.3 Refseq
Affymetrix Human Clariom D Assay PSR1200194829.hg.1 TC1200012159.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1200194827.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Clariom D Assay PSR1200194827.hg.1 ENST00000546622.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200194827.hg.1 ENST00000546985.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200194827.hg.1 NM_002442.3 Refseq
Affymetrix Human Clariom D Assay PSR1200194827.hg.1 ENST00000257552.6 Ensembl
Affymetrix Human Clariom D Assay PSR1200194827.hg.1 ENST00000546622.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200194827.hg.1 ENST00000546985.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200194827.hg.1 NM_002442.3 Refseq
Affymetrix Human Clariom D Assay PSR1200194827.hg.1 TC1200012159.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0