This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01045909_g1
See other DGAT2 GE Assays ›
Gene Symbol:
DGAT2
Gene Name:
diacylglycerol O-acyltransferase 2
Gene Aliases:
ARAT, GS1999FULL, HMFN1045
Chromosome Location:
Chr.11: 75768733 - 75801536 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01045909_g1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DGAT2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001253891.1 NP_001240820.1 2 - 3 487 76
NM_032564.4 NP_115953.2 3 - 4 616 76
XM_011545304.2 XP_011543606.1 3 - 4 320 76
GenBank mRNA AB048286.2 - 3 - 4 587 76
AB073384.1 - 2 - 3 164 76
AF384161.2 - 3 - 4 364 76
AL834287.1 - 3 - 4 495 76
AY358532.1 - 3 - 4 541 76
AY780647.1 - 2 - 3 219 76
BC015234.1 - 3 - 4 605 3903313 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1100008477.hg.1
This assay targets the same location as HTA transcript cluster(s) TC11000812.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene DGAT2

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

glycerol metabolic process
positive regulation of triglyceride biosynthetic process
triglyceride biosynthetic process
lipid storage
low-density lipoprotein particle clearance
long-chain fatty-acyl-CoA metabolic process
cellular triglyceride homeostasis
acylglycerol acyl-chain remodeling
retinol metabolic process
cholesterol homeostasis
positive regulation of gluconeogenesis
negative regulation of fatty acid oxidation
diacylglycerol metabolic process
regulation of lipoprotein metabolic process
fatty acid homeostasis
fat pad development
cellular response to oleic acid
regulation of cholesterol metabolic process
regulation of plasma lipoprotein particle levels
2-acylglycerol O-acyltransferase activity
diacylglycerol O-acyltransferase activity
protein homodimerization activity
retinol O-fatty-acyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000228027.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000228027.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000228027.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000228027.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000228027.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000228027.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000228027.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000228027.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000228027.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000228027.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000228027.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000228027.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000228027.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000376262.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000376262.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000376262.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000376262.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000376262.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000376262.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000376262.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000376262.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000376262.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000376262.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000376262.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000376262.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000603276.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000603276.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000603276.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000603276.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000603276.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000603276.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000603363.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000603363.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000603363.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000603363.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000603363.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000603363.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000603363.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000603363.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000603363.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000603865.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000603865.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000603865.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000603865.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000603865.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000603865.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000603865.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000603865.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000604733.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000604733.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000604733.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000604733.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000604733.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000604733.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000604733.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000604935.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000604935.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000604935.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000604935.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000604935.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000605099.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000605099.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000605099.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000605099.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000605099.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000605608.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000605608.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 ENST00000605608.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 NM_001253891.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 NM_001253891.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 NM_001253891.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 NM_001253891.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 NM_001253891.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 NM_001253891.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 NM_001253891.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 NM_001253891.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 NM_001253891.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 NM_001253891.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 NM_001253891.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 NM_001253891.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 NM_032564.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 NM_032564.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 NM_032564.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 NM_032564.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 NM_032564.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 NM_032564.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 NM_032564.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 NM_032564.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 NM_032564.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 NM_032564.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 NM_032564.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 NM_032564.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008477.hg.1 NM_032564.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11010373.hg.1 ENST00000228027.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11010373.hg.1 ENST00000376262.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11010373.hg.1 NM_001253891.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11010373.hg.1 NM_032564.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11010373.hg.1 TC11000812.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000228027.11 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000228027.11 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000228027.11 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000228027.11 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000228027.11 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000228027.11 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000228027.11 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000228027.11 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000228027.11 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000228027.11 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000228027.11 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000228027.11 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000228027.11 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000376262.7 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000376262.7 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000376262.7 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000376262.7 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000376262.7 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000376262.7 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000376262.7 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000376262.7 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000376262.7 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000376262.7 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000376262.7 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000376262.7 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000603276.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000603276.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000603276.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000603276.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000603276.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000603276.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000603363.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000603363.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000603363.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000603363.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000603363.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000603363.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000603363.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000603363.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000603363.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000603865.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000603865.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000603865.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000603865.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000603865.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000603865.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000603865.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000603865.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000604733.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000604733.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000604733.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000604733.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000604733.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000604733.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000604733.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000604935.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000604935.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000604935.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000604935.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000604935.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000605099.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000605099.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000605099.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000605099.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000605099.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000605608.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000605608.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 ENST00000605608.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008477.hg.1 NM_001253891.1 Refseq
Affymetrix Human Clariom D Assay TC1100008477.hg.1 NM_001253891.1 Refseq
Affymetrix Human Clariom D Assay TC1100008477.hg.1 NM_001253891.1 Refseq
Affymetrix Human Clariom D Assay TC1100008477.hg.1 NM_001253891.1 Refseq
Affymetrix Human Clariom D Assay TC1100008477.hg.1 NM_001253891.1 Refseq
Affymetrix Human Clariom D Assay TC1100008477.hg.1 NM_001253891.1 Refseq
Affymetrix Human Clariom D Assay TC1100008477.hg.1 NM_001253891.1 Refseq
Affymetrix Human Clariom D Assay TC1100008477.hg.1 NM_001253891.1 Refseq
Affymetrix Human Clariom D Assay TC1100008477.hg.1 NM_001253891.1 Refseq
Affymetrix Human Clariom D Assay TC1100008477.hg.1 NM_001253891.1 Refseq
Affymetrix Human Clariom D Assay TC1100008477.hg.1 NM_001253891.1 Refseq
Affymetrix Human Clariom D Assay TC1100008477.hg.1 NM_001253891.1 Refseq
Affymetrix Human Clariom D Assay TC1100008477.hg.1 NM_032564.4 Refseq
Affymetrix Human Clariom D Assay TC1100008477.hg.1 NM_032564.4 Refseq
Affymetrix Human Clariom D Assay TC1100008477.hg.1 NM_032564.4 Refseq
Affymetrix Human Clariom D Assay TC1100008477.hg.1 NM_032564.4 Refseq
Affymetrix Human Clariom D Assay TC1100008477.hg.1 NM_032564.4 Refseq
Affymetrix Human Clariom D Assay TC1100008477.hg.1 NM_032564.4 Refseq
Affymetrix Human Clariom D Assay TC1100008477.hg.1 NM_032564.4 Refseq
Affymetrix Human Clariom D Assay TC1100008477.hg.1 NM_032564.4 Refseq
Affymetrix Human Clariom D Assay TC1100008477.hg.1 NM_032564.4 Refseq
Affymetrix Human Clariom D Assay TC1100008477.hg.1 NM_032564.4 Refseq
Affymetrix Human Clariom D Assay TC1100008477.hg.1 NM_032564.4 Refseq
Affymetrix Human Clariom D Assay TC1100008477.hg.1 NM_032564.4 Refseq
Affymetrix Human Clariom D Assay TC1100008477.hg.1 NM_032564.4 Refseq
Affymetrix Human Clariom D Assay PSR1100162765.hg.1 ENST00000228027.11 Ensembl
Affymetrix Human Clariom D Assay PSR1100162765.hg.1 ENST00000376262.7 Ensembl
Affymetrix Human Clariom D Assay PSR1100162765.hg.1 ENST00000603276.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100162765.hg.1 ENST00000604733.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100162765.hg.1 ENST00000604935.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100162765.hg.1 ENST00000605099.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100162765.hg.1 NM_001253891.1 Refseq
Affymetrix Human Clariom D Assay PSR1100162765.hg.1 NM_032564.4 Refseq
Affymetrix Human Clariom D Assay PSR1100162765.hg.1 ENST00000228027.11 Ensembl
Affymetrix Human Clariom D Assay PSR1100162765.hg.1 ENST00000376262.7 Ensembl
Affymetrix Human Clariom D Assay PSR1100162765.hg.1 ENST00000603276.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100162765.hg.1 ENST00000604733.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100162765.hg.1 ENST00000604935.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100162765.hg.1 ENST00000605099.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100162765.hg.1 NM_001253891.1 Refseq
Affymetrix Human Clariom D Assay PSR1100162765.hg.1 NM_032564.4 Refseq
Affymetrix Human Clariom D Assay PSR1100162765.hg.1 TC1100008477.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0