This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01045937_m1
See other ASIC5 GE Assays ›
Gene Symbol:
ASIC5
Gene Name:
acid sensing ion channel subunit family member 5
Gene Aliases:
ACCN5, HINAC, INAC
Chromosome Location:
Chr.4: 155829699 - 155867303 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01045937_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ASIC5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_017419.2 NP_059115.1 8 - 9 1286 102
XM_017008291.1 XP_016863780.1 7 - 8 2190 102
GenBank mRNA AJ252011.1 - 8 - 9 1286 102

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0400012212.hg.1
This assay targets the same location as HTA transcript cluster(s) TC04001669.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ASIC5

Panther Classification:

Molecular Function -

ion channel

Gene Ontology Categories:

Function(s) Process(es)

proton transport
ion transmembrane transport
sodium ion transmembrane transport
hydrogen ion channel activity
acid-sensing ion channel activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0400012212.hg.1 ENST00000537611.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012212.hg.1 ENST00000537611.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012212.hg.1 ENST00000537611.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012212.hg.1 ENST00000537611.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012212.hg.1 ENST00000537611.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012212.hg.1 ENST00000537611.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012212.hg.1 ENST00000537611.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012212.hg.1 ENST00000537611.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012212.hg.1 ENST00000537611.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012212.hg.1 ENST00000537611.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012212.hg.1 ENST00000537611.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012212.hg.1 ENST00000537611.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012212.hg.1 NM_017419.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012212.hg.1 NM_017419.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012212.hg.1 NM_017419.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012212.hg.1 NM_017419.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012212.hg.1 NM_017419.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012212.hg.1 NM_017419.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012212.hg.1 NM_017419.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012212.hg.1 NM_017419.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012212.hg.1 NM_017419.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012212.hg.1 NM_017419.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012212.hg.1 NM_017419.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012212.hg.1 NM_017419.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04023256.hg.1 TC04001669.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR04023256.hg.1 ENST00000537611.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04023256.hg.1 NM_017419.2 Refseq
Affymetrix Human Clariom D Assay TC0400012212.hg.1 ENST00000537611.2 Ensembl
Affymetrix Human Clariom D Assay TC0400012212.hg.1 ENST00000537611.2 Ensembl
Affymetrix Human Clariom D Assay TC0400012212.hg.1 ENST00000537611.2 Ensembl
Affymetrix Human Clariom D Assay TC0400012212.hg.1 ENST00000537611.2 Ensembl
Affymetrix Human Clariom D Assay TC0400012212.hg.1 ENST00000537611.2 Ensembl
Affymetrix Human Clariom D Assay TC0400012212.hg.1 ENST00000537611.2 Ensembl
Affymetrix Human Clariom D Assay TC0400012212.hg.1 ENST00000537611.2 Ensembl
Affymetrix Human Clariom D Assay TC0400012212.hg.1 ENST00000537611.2 Ensembl
Affymetrix Human Clariom D Assay TC0400012212.hg.1 ENST00000537611.2 Ensembl
Affymetrix Human Clariom D Assay TC0400012212.hg.1 ENST00000537611.2 Ensembl
Affymetrix Human Clariom D Assay TC0400012212.hg.1 ENST00000537611.2 Ensembl
Affymetrix Human Clariom D Assay TC0400012212.hg.1 ENST00000537611.2 Ensembl
Affymetrix Human Clariom D Assay TC0400012212.hg.1 NM_017419.2 Refseq
Affymetrix Human Clariom D Assay TC0400012212.hg.1 NM_017419.2 Refseq
Affymetrix Human Clariom D Assay TC0400012212.hg.1 NM_017419.2 Refseq
Affymetrix Human Clariom D Assay TC0400012212.hg.1 NM_017419.2 Refseq
Affymetrix Human Clariom D Assay TC0400012212.hg.1 NM_017419.2 Refseq
Affymetrix Human Clariom D Assay TC0400012212.hg.1 NM_017419.2 Refseq
Affymetrix Human Clariom D Assay TC0400012212.hg.1 NM_017419.2 Refseq
Affymetrix Human Clariom D Assay TC0400012212.hg.1 NM_017419.2 Refseq
Affymetrix Human Clariom D Assay TC0400012212.hg.1 NM_017419.2 Refseq
Affymetrix Human Clariom D Assay TC0400012212.hg.1 NM_017419.2 Refseq
Affymetrix Human Clariom D Assay TC0400012212.hg.1 NM_017419.2 Refseq
Affymetrix Human Clariom D Assay TC0400012212.hg.1 NM_017419.2 Refseq
Affymetrix Human Clariom D Assay PSR0400183869.hg.1 ENST00000537611.2 Ensembl
Affymetrix Human Clariom D Assay PSR0400183869.hg.1 NM_017419.2 Refseq
Affymetrix Human Clariom D Assay PSR0400183869.hg.1 ENST00000537611.2 Ensembl
Affymetrix Human Clariom D Assay PSR0400183869.hg.1 NM_017419.2 Refseq
Affymetrix Human Clariom D Assay PSR0400183869.hg.1 TC0400012212.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0