This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01046137_m1
See other KAT6B GE Assays ›
Gene Symbol:
KAT6B
Gene Name:
lysine acetyltransferase 6B
Gene Aliases:
GTPTS, MORF, MOZ2, MYST4, ZC2HC6B, qkf, querkopf
Chromosome Location:
Chr.10: 74824927 - 75032623 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01046137_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: KAT6B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001256469.1 NP_001243398.1 8 - 9 1816 121
XM_017016006.1 XP_016871495.1 8 - 9 1725 121
XM_017016007.1 XP_016871496.1 8 - 9 1588 121
XM_017016008.1 XP_016871497.1 7 - 8 1499 121
XM_017016009.1 XP_016871498.1 8 - 9 1816 121
GenBank mRNA AB002381.2 - 6 - 7 1115 121
AF113514.1 - 7 - 8 1430 121
AK074816.1 - 8 - 9 1629 121
BC150270.1 - 6 - 7 1142 9007238 121

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000008081.hg.1

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

histone modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

nucleosome assembly
transcription, DNA-templated
regulation of transcription, DNA-templated
histone acetylation
histone H3 acetylation
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
DNA binding
histone acetyltransferase activity
protein binding
transcription factor binding
zinc ion binding
acetyltransferase activity
protein complex binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Clariom D Assay PSR1000154865.hg.1 ENST00000287239.8 Ensembl
Affymetrix Human Clariom D Assay PSR1000154865.hg.1 ENST00000372711.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000154865.hg.1 ENST00000372714.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000154865.hg.1 ENST00000372724.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000154865.hg.1 ENST00000372725.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000154865.hg.1 ENST00000490365.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000154865.hg.1 NM_001256468.1 Refseq
Affymetrix Human Clariom D Assay PSR1000154865.hg.1 NM_001256469.1 Refseq
Affymetrix Human Clariom D Assay PSR1000154865.hg.1 NM_012330.3 Refseq
Affymetrix Human Clariom D Assay PSR1000154865.hg.1 ENST00000287239.8 Ensembl
Affymetrix Human Clariom D Assay PSR1000154865.hg.1 ENST00000372711.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000154865.hg.1 ENST00000372714.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000154865.hg.1 ENST00000372724.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000154865.hg.1 ENST00000372725.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000154865.hg.1 ENST00000490365.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000154865.hg.1 NM_001256468.1 Refseq
Affymetrix Human Clariom D Assay PSR1000154865.hg.1 NM_001256469.1 Refseq
Affymetrix Human Clariom D Assay PSR1000154865.hg.1 NM_012330.3 Refseq
Affymetrix Human Clariom D Assay PSR1000154865.hg.1 TC1000008081.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vh84l:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0