This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01046161_m1
See other GPR107 GE Assays ›
Gene Symbol:
GPR107
Gene Name:
G protein-coupled receptor 107
Gene Aliases:
GCDRP, LUSTR1, bA138E2.2
Chromosome Location:
Chr.9: 130053706 - 130140169 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01046161_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPR107
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001287346.1 NP_001274275.1 13 - 14 1486 80
NM_020960.4 NP_066011.2 13 - 14 1490 80
XM_017014953.1 XP_016870442.1 13 - 14 1451 80
GenBank mRNA AF376725.1 - 13 - 14 1271 80
AK293092.1 - 13 - 14 1271 80
AL834359.1 - 12 - 13 1134 80
BC110518.2 - 13 - 14 1271 40033360 80

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC09000728.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GPR107

Panther Classification:

Molecular Function -

G-protein coupled receptor transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

clathrin-mediated endocytosis
clathrin heavy chain binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR09009156.hg.1 TC09000728.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR09009156.hg.1 ENST00000347136.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09009156.hg.1 ENST00000372406.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09009156.hg.1 ENST00000372410.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09009156.hg.1 ENST00000493417.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09009156.hg.1 NM_001136557.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09009156.hg.1 NM_001136558.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09009156.hg.1 NM_020960.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09009155.hg.1 TC09000728.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR09009155.hg.1 ENST00000347136.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09009155.hg.1 ENST00000372406.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09009155.hg.1 ENST00000372410.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09009155.hg.1 ENST00000493417.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09009155.hg.1 NM_001136557.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09009155.hg.1 NM_001136558.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09009155.hg.1 NM_020960.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-544xs:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0