This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01047491_m1
See other GTF3A GE Assays ›
Gene Symbol:
GTF3A
Gene Name:
general transcription factor IIIA
Gene Aliases:
AP2, TFIIIA
Chromosome Location:
Chr.13: 27424544 - 27435709 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01047491_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GTF3A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_002097.2 NP_002088.2 4 - 5 687 71
GenBank mRNA U20272.1 - 4 - 5 487 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1300006683.hg.1
This assay targets the same location as HTA transcript cluster(s) TC13000077.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GTF3A

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
transcription from RNA polymerase III promoter
rRNA transcription
DNA binding
RNA binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR13000791.hg.1 ENST00000381140.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13000791.hg.1 ENST00000470606.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13000791.hg.1 NM_002097.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13000791.hg.1 TC13000077.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000381140.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000381140.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000381140.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000381140.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000381140.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000381140.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000381140.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000381140.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000381140.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000381140.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000381140.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000381140.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000381140.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000381140.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000419181.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000419181.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000419181.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000419181.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000419181.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000419181.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000419181.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000419181.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000419181.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000419181.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000419181.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000419181.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000435781.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000435781.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000435781.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000435781.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000435781.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000435781.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000435781.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000435781.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000438571.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000438571.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000438571.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000438571.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000438571.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000438571.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000438571.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000438571.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000439403.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000439403.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000439403.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000439403.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000439403.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000439403.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000439403.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000439403.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000439403.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000439403.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000439403.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000439403.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000439403.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000439403.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000466776.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000466776.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000466776.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000466776.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000466776.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000470606.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000470606.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000470606.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000470606.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000470606.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000470606.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000470606.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000470606.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000470606.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000470606.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000482655.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000482655.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000482655.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000482655.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000482655.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000482655.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000493903.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000493903.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000493903.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000493903.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000493903.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 ENST00000493903.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 NM_002097.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 NM_002097.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 NM_002097.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 NM_002097.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 NM_002097.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 NM_002097.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 NM_002097.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 NM_002097.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 NM_002097.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 NM_002097.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 NM_002097.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 NM_002097.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 NM_002097.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300006683.hg.1 NM_002097.2 Refseq
Affymetrix Human Clariom D Assay PSR1300145600.hg.1 ENST00000381140.8 Ensembl
Affymetrix Human Clariom D Assay PSR1300145600.hg.1 ENST00000435781.5 Ensembl
Affymetrix Human Clariom D Assay PSR1300145600.hg.1 ENST00000439403.5 Ensembl
Affymetrix Human Clariom D Assay PSR1300145600.hg.1 ENST00000466776.5 Ensembl
Affymetrix Human Clariom D Assay PSR1300145600.hg.1 ENST00000470606.5 Ensembl
Affymetrix Human Clariom D Assay PSR1300145600.hg.1 ENST00000493903.1 Ensembl
Affymetrix Human Clariom D Assay PSR1300145600.hg.1 NM_002097.2 Refseq
Affymetrix Human Clariom D Assay PSR1300145600.hg.1 ENST00000381140.8 Ensembl
Affymetrix Human Clariom D Assay PSR1300145600.hg.1 ENST00000435781.5 Ensembl
Affymetrix Human Clariom D Assay PSR1300145600.hg.1 ENST00000439403.5 Ensembl
Affymetrix Human Clariom D Assay PSR1300145600.hg.1 ENST00000466776.5 Ensembl
Affymetrix Human Clariom D Assay PSR1300145600.hg.1 ENST00000470606.5 Ensembl
Affymetrix Human Clariom D Assay PSR1300145600.hg.1 ENST00000493903.1 Ensembl
Affymetrix Human Clariom D Assay PSR1300145600.hg.1 NM_002097.2 Refseq
Affymetrix Human Clariom D Assay PSR1300145600.hg.1 TC1300006683.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000381140.8 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000381140.8 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000381140.8 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000381140.8 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000381140.8 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000381140.8 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000381140.8 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000381140.8 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000381140.8 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000381140.8 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000381140.8 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000381140.8 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000381140.8 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000381140.8 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000419181.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000419181.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000419181.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000419181.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000419181.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000419181.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000419181.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000419181.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000419181.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000419181.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000419181.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000419181.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000435781.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000435781.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000435781.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000435781.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000435781.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000435781.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000435781.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000435781.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000438571.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000438571.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000438571.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000438571.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000438571.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000438571.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000438571.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000438571.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000439403.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000439403.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000439403.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000439403.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000439403.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000439403.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000439403.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000439403.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000439403.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000439403.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000439403.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000439403.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000439403.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000439403.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000466776.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000466776.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000466776.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000466776.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000466776.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000470606.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000470606.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000470606.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000470606.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000470606.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000470606.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000470606.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000470606.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000470606.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000470606.5 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000482655.2 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000482655.2 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000482655.2 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000482655.2 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000482655.2 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000482655.2 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000493903.1 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000493903.1 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000493903.1 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000493903.1 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000493903.1 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 ENST00000493903.1 Ensembl
Affymetrix Human Clariom D Assay TC1300006683.hg.1 NM_002097.2 Refseq
Affymetrix Human Clariom D Assay TC1300006683.hg.1 NM_002097.2 Refseq
Affymetrix Human Clariom D Assay TC1300006683.hg.1 NM_002097.2 Refseq
Affymetrix Human Clariom D Assay TC1300006683.hg.1 NM_002097.2 Refseq
Affymetrix Human Clariom D Assay TC1300006683.hg.1 NM_002097.2 Refseq
Affymetrix Human Clariom D Assay TC1300006683.hg.1 NM_002097.2 Refseq
Affymetrix Human Clariom D Assay TC1300006683.hg.1 NM_002097.2 Refseq
Affymetrix Human Clariom D Assay TC1300006683.hg.1 NM_002097.2 Refseq
Affymetrix Human Clariom D Assay TC1300006683.hg.1 NM_002097.2 Refseq
Affymetrix Human Clariom D Assay TC1300006683.hg.1 NM_002097.2 Refseq
Affymetrix Human Clariom D Assay TC1300006683.hg.1 NM_002097.2 Refseq
Affymetrix Human Clariom D Assay TC1300006683.hg.1 NM_002097.2 Refseq
Affymetrix Human Clariom D Assay TC1300006683.hg.1 NM_002097.2 Refseq
Affymetrix Human Clariom D Assay TC1300006683.hg.1 NM_002097.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline