This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01047785_m1
See other DHRS9 GE Assays ›
Gene Symbol:
DHRS9
Gene Name:
dehydrogenase/reductase 9
Gene Aliases:
3-alpha-HSD, 3ALPHA-HSD, RDH-E2, RDH-TBE, RDH15, RDHL, RDHTBE, RETSDR8, SDR9C4
Chromosome Location:
Chr.2: 169067035 - 169096167 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01047785_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DHRS9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_199204.1 NP_954674.1 2 - 3 498 125
GenBank mRNA AF240698.1 - 1 - 2 65 125
AF343729.1 - 1 - 2 94 125
AK313220.1 - 2 - 3 323 125
AY017349.1 - 2 - 3 498 125
BC058883.1 - 1 - 2 51 5168747 125

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200009905.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02001000.hg.1

Additional Information:

This assay is in the 5'UTR region of the AF240698.1,AF343729.1,AK313220.1,AY017349.1,BC058883.1,NM_199204.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene DHRS9

Panther Classification:

Molecular Function -

dehydrogenase oxidoreductase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

retinoic acid biosynthetic process
androgen metabolic process
epithelial cell differentiation
progesterone metabolic process
retinol metabolic process
9-cis-retinoic acid biosynthetic process
oxidation-reduction process
alcohol dehydrogenase (NAD) activity
retinol dehydrogenase activity
racemase and epimerase activity
testosterone dehydrogenase (NAD+) activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000357546.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000357546.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000357546.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000357546.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000357546.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000357546.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000357546.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000357546.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000357546.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000357546.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000357546.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000357546.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000357546.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000412271.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000412271.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000412271.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000412271.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000412271.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000412271.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000412271.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000412271.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000412271.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000421653.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000421653.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000421653.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000421653.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000421653.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000421653.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000421653.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000428522.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000428522.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000428522.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000428522.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000428522.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000428522.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000428522.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000428522.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000428522.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000428522.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000428522.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000428522.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000432060.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000432060.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000432060.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000432060.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000432060.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000432060.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000432060.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000432060.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000432060.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000432060.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000436483.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000436483.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000436483.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000436483.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000436483.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000436483.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000436483.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000436483.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000436483.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000436483.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000436483.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000450153.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000450153.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000450153.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000450153.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000450153.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000450153.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000602501.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000602501.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000602501.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000602501.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000602501.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000602501.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000602501.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000602501.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000602501.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000602501.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000602501.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000602501.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142270.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142270.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142270.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142270.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142270.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142270.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142270.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142270.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142270.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142270.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001289763.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001289763.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001289763.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001289763.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001289763.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001289763.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001289763.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001289763.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001289763.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001289763.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_199204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_199204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_199204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_199204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_199204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_199204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_199204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_199204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_199204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_199204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_199204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_199204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_199204.1 Refseq
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 116895 - -
ABI Human Genome Survey Microarray Version 2.0 202451 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline