This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01047787_m1
See other DHRS9 GE Assays ›
Gene Symbol:
DHRS9
Gene Name:
dehydrogenase/reductase 9
Gene Aliases:
3-alpha-HSD, 3ALPHA-HSD, RDH-E2, RDH-TBE, RDH15, RDHL, RDHTBE, RETSDR8, SDR9C4
Chromosome Location:
Chr.2: 169067035 - 169096167 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01047787_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DHRS9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001142270.1 NP_001135742.1 4 - 5 955 82
NM_001142271.1 NP_001135743.1 3 - 4 678 82
NM_001289763.1 NP_001276692.1 3 - 4 865 82
NM_199204.1 NP_954674.1 4 - 5 1130 82
XM_011510475.1 XP_011508777.1 5 - 6 1288 82
GenBank mRNA AF067174.1 - 3 - 4 592 82
AF240698.1 - 3 - 4 697 82
AF295380.1 - 2 - 3 573 82
AF343729.1 - 3 - 4 726 82
AF529286.1 - 6 - 7 2076 82
AF529287.1 - 4 - 5 1030 82
AF529288.1 - 2 - 3 638 82
AK296625.1 - 3 - 4 823 82
AK313220.1 - 4 - 5 955 82
AY017349.1 - 4 - 5 1130 82
AY359046.1 - 3 - 4 683 82
BC058883.1 - 3 - 4 683 5168747 82

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200009905.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02001000.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene DHRS9

Panther Classification:

Molecular Function -

dehydrogenase oxidoreductase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

retinoic acid biosynthetic process
androgen metabolic process
epithelial cell differentiation
progesterone metabolic process
retinol metabolic process
9-cis-retinoic acid biosynthetic process
oxidation-reduction process
alcohol dehydrogenase (NAD) activity
retinol dehydrogenase activity
racemase and epimerase activity
testosterone dehydrogenase (NAD+) activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000357546.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000357546.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000357546.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000357546.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000357546.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000357546.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000357546.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000357546.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000357546.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000357546.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000357546.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000357546.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000357546.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000412271.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000412271.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000412271.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000412271.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000412271.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000412271.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000412271.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000412271.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000412271.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000421653.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000421653.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000421653.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000421653.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000421653.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000421653.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000421653.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000428522.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000428522.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000428522.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000428522.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000428522.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000428522.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000428522.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000428522.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000428522.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000428522.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000428522.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000428522.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000432060.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000432060.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000432060.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000432060.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000432060.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000432060.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000432060.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000432060.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000432060.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000432060.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000436483.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000436483.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000436483.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000436483.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000436483.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000436483.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000436483.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000436483.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000436483.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000436483.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000436483.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000450153.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000450153.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000450153.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000450153.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000450153.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000450153.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000602501.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000602501.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000602501.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000602501.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000602501.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000602501.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000602501.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000602501.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000602501.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000602501.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000602501.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000602501.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142270.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142270.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142270.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142270.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142270.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142270.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142270.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142270.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142270.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142270.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001289763.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001289763.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001289763.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001289763.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001289763.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001289763.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001289763.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001289763.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001289763.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001289763.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_199204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_199204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_199204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_199204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_199204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_199204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_199204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_199204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_199204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_199204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_199204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_199204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_199204.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hwq2n:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0