This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01047954_m1
See other UNC50 GE Assays ›
Gene Symbol:
UNC50
Gene Name:
unc-50 inner nuclear membrane RNA binding protein
Gene Aliases:
GMH1, HSD23, UNCL, URP
Chromosome Location:
Chr.2: 98608569 - 98618515 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01047954_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UNC50
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA BC017019.1 - 1 - 2 171 3888349 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200008573.hg.1

Additional Information:

This assay is in the 5'UTR region of the BC017019.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens 1700 Probe spans exons

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000357765.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000357765.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000357765.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000357765.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000357765.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000357765.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000357765.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000393493.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000393493.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000393493.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000393493.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000409347.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000409347.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000409347.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000409347.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000409347.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000409347.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000409347.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000409347.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000409975.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000409975.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000409975.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000409975.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000409975.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000409975.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000409975.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000409975.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000409975.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000409975.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000409975.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000423713.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000423713.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000423713.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000466492.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000466492.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000466492.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000466492.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 NM_014044.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 NM_014044.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 NM_014044.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 NM_014044.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 NM_014044.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 NM_014044.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 NM_014044.6 Refseq
Affymetrix Human Clariom D Assay PSR0200159855.hg.1 ENST00000409347.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200159855.hg.1 ENST00000409975.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200159855.hg.1 ENST00000409347.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200159855.hg.1 ENST00000409975.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200159855.hg.1 TC0200008573.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0200159853.hg.1 ENST00000409347.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200159853.hg.1 ENST00000409975.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200159853.hg.1 ENST00000409347.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200159853.hg.1 ENST00000409975.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200159853.hg.1 TC0200008573.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000357765.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000357765.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000357765.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000357765.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000357765.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000357765.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000357765.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000393493.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000393493.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000393493.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000393493.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000409347.5 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000409347.5 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000409347.5 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000409347.5 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000409347.5 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000409347.5 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000409347.5 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000409347.5 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000409975.5 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000409975.5 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000409975.5 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000409975.5 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000409975.5 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000409975.5 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000409975.5 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000409975.5 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000409975.5 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000409975.5 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000409975.5 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000423713.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000423713.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000423713.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000466492.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000466492.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000466492.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000466492.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 NM_014044.6 Refseq
Affymetrix Human Clariom D Assay TC0200008573.hg.1 NM_014044.6 Refseq
Affymetrix Human Clariom D Assay TC0200008573.hg.1 NM_014044.6 Refseq
Affymetrix Human Clariom D Assay TC0200008573.hg.1 NM_014044.6 Refseq
Affymetrix Human Clariom D Assay TC0200008573.hg.1 NM_014044.6 Refseq
Affymetrix Human Clariom D Assay TC0200008573.hg.1 NM_014044.6 Refseq
Affymetrix Human Clariom D Assay TC0200008573.hg.1 NM_014044.6 Refseq
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 191105 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0