This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01047956_m1
See other UNC50 GE Assays ›
Gene Symbol:
UNC50
Gene Name:
unc-50 inner nuclear membrane RNA binding protein
Gene Aliases:
GMH1, HSD23, UNCL, URP
Chromosome Location:
Chr.2: 98608569 - 98618515 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01047956_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UNC50
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_014044.5 NP_054763.2 3 - 4 550 80
XM_005263913.4 XP_005263970.1 3 - 4 646 80
XM_006712403.3 XP_006712466.1 3 - 4 646 80
XM_006712404.2 XP_006712467.1 3 - 4 560 80
GenBank mRNA AF077038.1 - 3 - 4 502 80
AK222599.1 - 3 - 4 516 80
AL080115.1 - 3 - 4 554 80
AY017215.1 - 3 - 4 518 80
AY251534.1 - 2 - 3 411 80
BC017019.1 - 3 - 4 683 3888349 80
BC025992.2 - 2 - 3 1578 4830706 80
BC039078.1 - 2 - 3 1554 5166015 80
BC091481.1 - 3 - 4 540 30337350 80
CR533464.1 - 2 - 3 398 80
KU178586.1 - 2 - 3 398 80

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200008573.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02000600.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene UNC50

Gene Ontology Categories:

Function(s) Process(es)

transport
cell surface receptor signaling pathway
protein transport
RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000357765.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000357765.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000357765.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000357765.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000357765.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000357765.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000357765.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000393493.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000393493.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000393493.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000393493.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000409347.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000409347.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000409347.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000409347.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000409347.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000409347.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000409347.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000409347.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000409975.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000409975.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000409975.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000409975.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000409975.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000409975.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000409975.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000409975.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000409975.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000409975.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000409975.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000423713.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000423713.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000423713.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000466492.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000466492.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000466492.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 ENST00000466492.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 NM_014044.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 NM_014044.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 NM_014044.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 NM_014044.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 NM_014044.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 NM_014044.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008573.hg.1 NM_014044.6 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02009440.hg.1 TC02000600.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR02009440.hg.1 ENST00000357765.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02009440.hg.1 ENST00000409347.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02009440.hg.1 ENST00000409975.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02009440.hg.1 NM_014044.6 Refseq
Affymetrix Human Clariom D Assay PSR0200159860.hg.1 ENST00000357765.6 Ensembl
Affymetrix Human Clariom D Assay PSR0200159860.hg.1 ENST00000393493.6 Ensembl
Affymetrix Human Clariom D Assay PSR0200159860.hg.1 ENST00000409347.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200159860.hg.1 ENST00000409975.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200159860.hg.1 ENST00000423713.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200159860.hg.1 NM_014044.6 Refseq
Affymetrix Human Clariom D Assay PSR0200159860.hg.1 ENST00000357765.6 Ensembl
Affymetrix Human Clariom D Assay PSR0200159860.hg.1 ENST00000393493.6 Ensembl
Affymetrix Human Clariom D Assay PSR0200159860.hg.1 ENST00000409347.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200159860.hg.1 ENST00000409975.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200159860.hg.1 ENST00000423713.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200159860.hg.1 NM_014044.6 Refseq
Affymetrix Human Clariom D Assay PSR0200159860.hg.1 TC0200008573.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000357765.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000357765.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000357765.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000357765.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000357765.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000357765.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000357765.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000393493.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000393493.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000393493.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000393493.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000409347.5 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000409347.5 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000409347.5 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000409347.5 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000409347.5 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000409347.5 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000409347.5 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000409347.5 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000409975.5 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000409975.5 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000409975.5 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000409975.5 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000409975.5 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000409975.5 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000409975.5 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000409975.5 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000409975.5 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000409975.5 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000409975.5 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000423713.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000423713.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000423713.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000466492.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000466492.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000466492.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 ENST00000466492.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008573.hg.1 NM_014044.6 Refseq
Affymetrix Human Clariom D Assay TC0200008573.hg.1 NM_014044.6 Refseq
Affymetrix Human Clariom D Assay TC0200008573.hg.1 NM_014044.6 Refseq
Affymetrix Human Clariom D Assay TC0200008573.hg.1 NM_014044.6 Refseq
Affymetrix Human Clariom D Assay TC0200008573.hg.1 NM_014044.6 Refseq
Affymetrix Human Clariom D Assay TC0200008573.hg.1 NM_014044.6 Refseq
Affymetrix Human Clariom D Assay TC0200008573.hg.1 NM_014044.6 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-r7bw6:80/100.66.78.215:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0