This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01048152_m1
See other BACE2 GE Assays ›
Gene Symbol:
BACE2
Gene Name:
beta-site APP-cleaving enzyme 2
Gene Aliases:
AEPLC, ALP56, ASP1, ASP21, BAE2, CDA13, CEAP1, DRAP
Chromosome Location:
Chr.21: 41167801 - 41282534 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01048152_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BACE2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_012105.4 NP_036237.2 2 - 3 868 71
NM_138991.2 NP_620476.1 2 - 3 868 71
NM_138992.2 NP_620477.1 2 - 3 868 71
XM_017028314.1 XP_016883803.1 2 - 3 540 71
GenBank mRNA AF050171.1 - 2 - 3 513 71
AF117892.1 - 2 - 3 505 71
AF178532.1 - 2 - 3 868 71
AF188276.1 - 2 - 3 868 71
AF188277.1 - 2 - 3 868 71
AF200192.1 - 2 - 3 482 71
AF200342.1 - 2 - 3 405 71
AF204944.1 - 2 - 3 495 71
AF212252.1 - 2 - 3 226 71
AK075539.1 - 3 - 4 362 71
AK292056.1 - 2 - 3 541 71
AY358927.1 - 2 - 3 498 71
BC014453.1 - 2 - 3 496 4868925 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2100007198.hg.1
This assay targets the same location as HTA transcript cluster(s) TC21000185.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene BACE2

Panther Classification:

Molecular Function -

protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
membrane protein ectodomain proteolysis
peptide hormone processing
protein catabolic process
negative regulation of amyloid precursor protein biosynthetic process
beta-amyloid metabolic process
beta-amyloid binding
aspartic-type endopeptidase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000463674.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000463674.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000463674.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000463674.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000463674.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000463674.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000463674.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000463674.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000463674.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000463674.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000465326.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000465326.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000465326.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000465326.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000465326.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000465326.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000465326.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000465326.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000466122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000466122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000466122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000466122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000466122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000466122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000466122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000466122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000466122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000466122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000466122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000466122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000466122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000470864.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000470864.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000470864.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000470864.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000475618.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000475618.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000487994.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000487994.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000487994.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000487994.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000487994.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000487994.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000487994.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000487994.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000487994.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000491838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000491838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000491838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000491838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000491838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000491838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000491838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline