This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01048155_m1
See other BACE2 GE Assays ›
Gene Symbol:
BACE2
Gene Name:
beta-site APP-cleaving enzyme 2
Gene Aliases:
AEPLC, ALP56, ASP1, ASP21, BAE2, CDA13, CEAP1, DRAP
Chromosome Location:
Chr.21: 41167801 - 41282534 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01048155_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BACE2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_012105.4 NP_036237.2 5 - 6 1348 70
NM_138991.2 NP_620476.1 5 - 6 1348 70
NM_138992.2 NP_620477.1 5 - 6 1348 70
XM_017028314.1 XP_016883803.1 5 - 6 1020 70
GenBank mRNA AF050171.1 - 5 - 6 993 70
AF117892.1 - 5 - 6 985 70
AF178532.1 - 5 - 6 1348 70
AF188276.1 - 5 - 6 1348 70
AF188277.1 - 5 - 6 1348 70
AF200192.1 - 5 - 6 962 70
AF200342.1 - 5 - 6 885 70
AF204944.1 - 5 - 6 975 70
AF212252.1 - 5 - 6 706 70
AK075539.1 - 6 - 7 842 70
AK292056.1 - 5 - 6 1021 70
AY358927.1 - 5 - 6 978 70
BC014453.1 - 5 - 6 976 4868925 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2100007198.hg.1
This assay targets the same location as HTA transcript cluster(s) TC21000185.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene BACE2

Panther Classification:

Molecular Function -

protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
membrane protein ectodomain proteolysis
peptide hormone processing
protein catabolic process
negative regulation of amyloid precursor protein biosynthetic process
beta-amyloid metabolic process
beta-amyloid binding
aspartic-type endopeptidase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000463674.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000463674.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000463674.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000463674.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000463674.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000463674.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000463674.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000463674.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000463674.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000463674.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000465326.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000465326.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000465326.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000465326.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000465326.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000465326.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000465326.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000465326.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000466122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000466122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000466122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000466122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000466122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000466122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000466122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000466122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000466122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000466122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000466122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000466122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000466122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000470864.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000470864.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000470864.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000470864.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000475618.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000475618.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000487994.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000487994.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000487994.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000487994.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000487994.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000487994.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000487994.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000487994.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000487994.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000491838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000491838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000491838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000491838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000491838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000491838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000491838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline