This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01048158_m1
See other BACE2 GE Assays ›
Gene Symbol:
BACE2
Gene Name:
beta-site APP-cleaving enzyme 2
Gene Aliases:
AEPLC, ALP56, ASP1, ASP21, BAE2, CDA13, CEAP1, DRAP
Chromosome Location:
Chr.21: 41167801 - 41282534 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01048158_m1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BACE2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_138991.2 NP_620476.1 6 - 7 1444 99
GenBank mRNA AF188276.1 - 6 - 7 1444 99

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Protease Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene BACE2

Panther Classification:

Molecular Function -

protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
membrane protein ectodomain proteolysis
peptide hormone processing
protein catabolic process
negative regulation of amyloid precursor protein biosynthetic process
beta-amyloid metabolic process
beta-amyloid binding
aspartic-type endopeptidase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000463674.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000463674.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000463674.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000463674.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000463674.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000463674.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000463674.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000463674.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000463674.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000463674.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000465326.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000465326.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000465326.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000465326.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000465326.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000465326.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000465326.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000465326.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000466122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000466122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000466122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000466122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000466122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000466122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000466122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000466122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000466122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000466122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000466122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000466122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000466122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000470864.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000470864.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000470864.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000470864.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000475618.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000475618.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000487994.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000487994.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000487994.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000487994.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000487994.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000487994.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000487994.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000487994.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000487994.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000491838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000491838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000491838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000491838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000491838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000491838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000491838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline