This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01049569_g1
See other GFPT2 GE Assays ›
Gene Symbol:
GFPT2
Gene Name:
glutamine-fructose-6-phosphate transaminase 2
Gene Aliases:
GFAT, GFAT 2, GFAT2
Chromosome Location:
Chr.5: 180300690 - 180353387 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01049569_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GFPT2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_005110.3 NP_005101.1 8 - 9 848 85
XM_006714942.2 XP_006715005.1 8 - 9 797 85
XM_017010100.1 XP_016865589.1 7 - 8 733 85
GenBank mRNA AB016789.1 - 8 - 9 777 85
AK022507.1 - 8 - 9 788 85
AK297750.1 - 6 - 7 484 85
AK314551.1 - 8 - 9 788 85
BC000012.2 - 8 - 9 786 3506723 85
BT009818.1 - 7 - 8 678 85
KU178494.1 - 7 - 8 678 85
KU178495.1 - 7 - 8 678 85

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0500013089.hg.1
This assay targets the same location as HTA transcript cluster(s) TC05002146.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GFPT2

Panther Classification:

Molecular Function -

transaminase

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
fructose 6-phosphate metabolic process
UDP-N-acetylglucosamine metabolic process
UDP-N-acetylglucosamine biosynthetic process
energy reserve metabolic process
glutamine metabolic process
glutamine-fructose-6-phosphate transaminase (isomerizing) activity
protein binding
carbohydrate binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000253778.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000253778.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000253778.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000253778.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000253778.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000253778.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000253778.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000253778.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000253778.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000253778.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000253778.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000253778.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000253778.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000253778.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000253778.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000253778.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000253778.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000253778.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000253778.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000253778.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000253778.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000253778.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000253778.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000253778.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000503228.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000503228.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000503546.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000503546.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000503546.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000503546.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000503546.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000503546.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000510122.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000510122.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000510122.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000510122.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000510122.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000518158.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000518158.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000518158.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000518158.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000518158.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000518158.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000518158.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000518158.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000518185.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000518185.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000518185.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000518185.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000518185.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000518185.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000518906.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000518906.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000518906.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000518906.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000518906.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000518906.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000518906.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000518906.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000520165.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000520165.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000520165.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000520165.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000520165.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000520165.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000520165.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 NM_005110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 NM_005110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 NM_005110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 NM_005110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 NM_005110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 NM_005110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 NM_005110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 NM_005110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 NM_005110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 NM_005110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 NM_005110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 NM_005110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 NM_005110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 NM_005110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 NM_005110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 NM_005110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 NM_005110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 NM_005110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 NM_005110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 NM_005110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 NM_005110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 NM_005110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 NM_005110.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline