This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01051130_g1
See other COX11 GE Assays ›
Gene Symbol:
COX11
Gene Name:
COX11, cytochrome c oxidase copper chaperone
Gene Aliases:
COX11P
Chromosome Location:
Chr.17: 54951898 - 54971024 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01051130_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: COX11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001162861.2 NP_001156333.1 1 - 2 503 62
NM_001162862.2 NP_001156334.1 1 - 2 503 62
NM_001321518.1 NP_001308447.1 1 - 2 503 62
NM_004375.4 NP_004366.1 1 - 2 503 62
NR_027941.2 1 - 2 503 62
NR_027942.2 1 - 2 503 62
NR_135677.1 1 - 2 503 62
XM_011524342.2 XP_011522644.1 1 - 2 2742 62
XM_017024192.1 XP_016879681.1 1 - 2 2742 62
XM_017024193.1 XP_016879682.1 1 - 2 2742 62
XM_017024194.1 XP_016879683.1 1 - 2 498 62
XM_017024195.1 XP_016879684.1 1 - 2 498 62
XM_017024196.1 XP_016879685.1 1 - 2 498 62
GenBank mRNA AF044321.1 - 1 - 2 379 62
AK293851.1 - 1 - 2 336 62
AK295381.1 - 1 - 2 421 62
BC005895.1 - 1 - 2 374 4282744 62
BF725369.1 - 1 - 2 503 62
BI598312.1 - 1 - 2 419 62
CR541837.1 - 1 - 2 364 62

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene COX11

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

respiratory gaseous exchange
respiratory chain complex IV assembly
aerobic respiration
negative regulation of glucokinase activity
hydrogen ion transmembrane transport
cytochrome-c oxidase activity
copper ion binding
electron carrier activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0