This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01051134_s1
See other COX11 GE Assays ›
Gene Symbol:
COX11
Gene Name:
COX11, cytochrome c oxidase copper chaperone
Gene Aliases:
COX11P
Chromosome Location:
Chr.17: 54951898 - 54971024 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01051134_s1
Size
Availability Made To Order
Catalog # 4426961
Price 434.00
Your Price
Online offer: 407.65
407.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: COX11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK124809.1 - 1 2728 124
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DB306740.1 - 1 Chr. 17 - 54951906 - 54952445 [+] on Build GRCh38 124
GenBankmRNA AW104476.1 - 1 Chr. 17 - 54951902 - 54952281 [+] on Build GRCh38 124
GenBankmRNA AA194498.1 - 1 Chr. 17 - 54951935 - 54952402 [+] on Build GRCh38 124
GenBankmRNA DB299831.1 - 1 Chr. 17 - 54951906 - 54952481 [+] on Build GRCh38 124
GenBankmRNA BX481798.1 - 1 Chr. 17 - 54951901 - 54952463 [+] on Build GRCh38 124
GenBankmRNA AA928058.1 - 1 Chr. 17 - 54951902 - 54952281 [+] on Build GRCh38 124

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream kinesin family member 2B KIF2B -1.1mb Sense Chr. 17 - 53822878 - 53825212 on NCBI Build GRCh38
Downstream syntaxin binding protein 4 STXBP4 Synip 16.3kb Sense Chr. 17 - 54968728 - 55164285 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0