This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01051144_s1
See other COX11 GE Assays ›
Gene Symbol:
COX11
Gene Name:
COX11, cytochrome c oxidase copper chaperone
Gene Aliases:
COX11P
Chromosome Location:
Chr.17: 54951898 - 54971024 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01051144_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: COX11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017024194.1 XP_016879683.1 5 6445 127
XM_017024195.1 XP_016879684.1 5 6449 127
GenBank mRNA AK124809.1 - 1 886 127
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AW665723.1 - 1 Chr. 17 - 54953731 - 54954366 [+] on Build GRCh38 127
GenBankmRNA AW003220.1 - 1 Chr. 17 - 54953730 - 54954384 [+] on Build GRCh38 128

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream kinesin family member 2B KIF2B -1.1mb Anti-sense Chr. 17 - 53822878 - 53825212 on NCBI Build GRCh38
Downstream syntaxin binding protein 4 STXBP4 Synip 9.3kb Anti-sense Chr. 17 - 54968728 - 55164285 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

respiratory gaseous exchange
respiratory chain complex IV assembly
aerobic respiration
negative regulation of glucokinase activity
hydrogen ion transmembrane transport
cytochrome-c oxidase activity
copper ion binding
electron carrier activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000299335.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000299335.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000299335.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000299335.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000299335.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000299335.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000299335.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000299335.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000299335.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000299335.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000571584.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000571584.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000571584.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000571584.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000571584.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000572088.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000572088.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000572088.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000572088.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000572088.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000572088.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000572558.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000572558.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000572558.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000572558.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000572558.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000572558.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000572558.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000572558.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000573912.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000573912.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000573912.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000573912.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000573912.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000574821.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000574821.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000574821.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000574821.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000574821.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000574989.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000576084.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000576084.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000576084.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000576084.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000576370.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000576370.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000576370.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000576370.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000576370.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000576370.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000576370.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000576370.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000576370.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000576370.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 ENST00000576370.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 NM_001162861.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 NM_001162861.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 NM_001162861.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 NM_001162861.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 NM_001162861.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 NM_001162861.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 NM_001162862.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 NM_001162862.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 NM_001162862.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 NM_001162862.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 NM_001162862.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 NM_001162862.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 NM_004375.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 NM_004375.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 NM_004375.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 NM_004375.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 NM_004375.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 NM_004375.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 NM_004375.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 NM_004375.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 NM_004375.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 NR_027941.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 NR_027941.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 NR_027941.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 NR_027941.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 NR_027941.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 NR_027941.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 NR_027941.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 NR_027941.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 NR_027941.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 NR_027942.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 NR_027942.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 NR_027942.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 NR_027942.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 NR_027942.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 NR_027942.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 NR_027942.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 NR_027942.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011171.hg.1 NR_027942.2 Refseq
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000299335.7 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000299335.7 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000299335.7 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000299335.7 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000299335.7 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000299335.7 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000299335.7 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000299335.7 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000299335.7 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000299335.7 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000571584.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000571584.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000571584.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000571584.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000571584.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000572088.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000572088.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000572088.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000572088.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000572088.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000572088.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000572558.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000572558.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000572558.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000572558.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000572558.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000572558.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000572558.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000572558.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000573912.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000573912.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000573912.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000573912.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000573912.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000574821.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000574821.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000574821.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000574821.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000574821.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000574989.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000576084.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000576084.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000576084.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000576084.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000576370.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000576370.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000576370.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000576370.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000576370.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000576370.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000576370.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000576370.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000576370.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000576370.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 ENST00000576370.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011171.hg.1 NM_001162861.2 Refseq
Affymetrix Human Clariom D Assay TC1700011171.hg.1 NM_001162861.2 Refseq
Affymetrix Human Clariom D Assay TC1700011171.hg.1 NM_001162861.2 Refseq
Affymetrix Human Clariom D Assay TC1700011171.hg.1 NM_001162861.2 Refseq
Affymetrix Human Clariom D Assay TC1700011171.hg.1 NM_001162861.2 Refseq
Affymetrix Human Clariom D Assay TC1700011171.hg.1 NM_001162861.2 Refseq
Affymetrix Human Clariom D Assay TC1700011171.hg.1 NM_001162862.2 Refseq
Affymetrix Human Clariom D Assay TC1700011171.hg.1 NM_001162862.2 Refseq
Affymetrix Human Clariom D Assay TC1700011171.hg.1 NM_001162862.2 Refseq
Affymetrix Human Clariom D Assay TC1700011171.hg.1 NM_001162862.2 Refseq
Affymetrix Human Clariom D Assay TC1700011171.hg.1 NM_001162862.2 Refseq
Affymetrix Human Clariom D Assay TC1700011171.hg.1 NM_001162862.2 Refseq
Affymetrix Human Clariom D Assay TC1700011171.hg.1 NM_004375.4 Refseq
Affymetrix Human Clariom D Assay TC1700011171.hg.1 NM_004375.4 Refseq
Affymetrix Human Clariom D Assay TC1700011171.hg.1 NM_004375.4 Refseq
Affymetrix Human Clariom D Assay TC1700011171.hg.1 NM_004375.4 Refseq
Affymetrix Human Clariom D Assay TC1700011171.hg.1 NM_004375.4 Refseq
Affymetrix Human Clariom D Assay TC1700011171.hg.1 NM_004375.4 Refseq
Affymetrix Human Clariom D Assay TC1700011171.hg.1 NM_004375.4 Refseq
Affymetrix Human Clariom D Assay TC1700011171.hg.1 NM_004375.4 Refseq
Affymetrix Human Clariom D Assay TC1700011171.hg.1 NM_004375.4 Refseq
Affymetrix Human Clariom D Assay TC1700011171.hg.1 NR_027941.2 Refseq
Affymetrix Human Clariom D Assay TC1700011171.hg.1 NR_027941.2 Refseq
Affymetrix Human Clariom D Assay TC1700011171.hg.1 NR_027941.2 Refseq
Affymetrix Human Clariom D Assay TC1700011171.hg.1 NR_027941.2 Refseq
Affymetrix Human Clariom D Assay TC1700011171.hg.1 NR_027941.2 Refseq
Affymetrix Human Clariom D Assay TC1700011171.hg.1 NR_027941.2 Refseq
Affymetrix Human Clariom D Assay TC1700011171.hg.1 NR_027941.2 Refseq
Affymetrix Human Clariom D Assay TC1700011171.hg.1 NR_027941.2 Refseq
Affymetrix Human Clariom D Assay TC1700011171.hg.1 NR_027941.2 Refseq
Affymetrix Human Clariom D Assay TC1700011171.hg.1 NR_027942.2 Refseq
Affymetrix Human Clariom D Assay TC1700011171.hg.1 NR_027942.2 Refseq
Affymetrix Human Clariom D Assay TC1700011171.hg.1 NR_027942.2 Refseq
Affymetrix Human Clariom D Assay TC1700011171.hg.1 NR_027942.2 Refseq
Affymetrix Human Clariom D Assay TC1700011171.hg.1 NR_027942.2 Refseq
Affymetrix Human Clariom D Assay TC1700011171.hg.1 NR_027942.2 Refseq
Affymetrix Human Clariom D Assay TC1700011171.hg.1 NR_027942.2 Refseq
Affymetrix Human Clariom D Assay TC1700011171.hg.1 NR_027942.2 Refseq
Affymetrix Human Clariom D Assay TC1700011171.hg.1 NR_027942.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0