This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01053592_g1
See other NDST2 GE Assays ›
Gene Symbol:
NDST2
Gene Name:
N-deacetylase and N-sulfotransferase 2
Gene Aliases:
HSST2, NST2
Chromosome Location:
Chr.10: 73801911 - 73811831 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01053592_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NDST2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005270255.3 XP_005270312.1 13 - 14 3186 64

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

transferase

Gene Ontology Categories:

Function(s) Process(es)

glycosaminoglycan biosynthetic process
heparan sulfate proteoglycan biosynthetic process
heparin biosynthetic process
[heparan sulfate]-glucosamine N-sulfotransferase activity
N-acetylglucosamine deacetylase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7gdsv:80/100.66.79.88:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0