This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01053742_m1
See other SMARCA5 GE Assays ›
Gene Symbol:
SMARCA5
Gene Name:
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 5
Gene Aliases:
ISWI, SNF2H, WCRF135, hISWI, hSNF2H
Chromosome Location:
Chr.4: 143513463 - 143557489 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01053742_m1
Size
Availability Made To Order
Catalog # 4351372
Price 417.00
Your Price
Online offer: 387.65
387.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SMARCA5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_003601.3 NP_003592.3 2 - 3 716 114
GenBank mRNA AB010882.1 - 2 - 3 456 114
AK302841.1 - 3 - 4 351 114
AK308424.1 - 2 - 3 446 114
AK316444.1 - 3 - 4 518 114
BC023144.1 - 2 - 3 467 4340921 114

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0400008882.hg.1
This assay targets the same location as HTA transcript cluster(s) TC04000701.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SMARCA5

Panther Classification:

Molecular Function -

DNA helicase

Gene Ontology Categories:

Function(s) Process(es)

chromatin silencing at rDNA
double-strand break repair
nucleosome assembly
chromatin remodeling
DNA-templated transcription, initiation
regulation of transcription from RNA polymerase II promoter
covalent chromatin modification
nucleosome positioning
CENP-A containing nucleosome assembly
ATP-dependent chromatin remodeling
positive regulation of gene expression, epigenetic
positive regulation of transcription, DNA-templated
DNA binding
helicase activity
protein binding
ATP binding
ATPase activity
nucleosome binding
histone binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR04009959.hg.1 TC04000701.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR04009959.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04009959.hg.1 ENST00000535006.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04009959.hg.1 ENST00000536484.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04009959.hg.1 NM_003601.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 ENST00000508573.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 ENST00000508573.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 ENST00000508573.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 ENST00000515531.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 ENST00000515531.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 ENST00000515531.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Clariom D Assay PSR0400160969.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Clariom D Assay PSR0400160969.hg.1 NM_003601.3 Refseq
Affymetrix Human Clariom D Assay PSR0400160969.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Clariom D Assay PSR0400160969.hg.1 NM_003601.3 Refseq
Affymetrix Human Clariom D Assay PSR0400160969.hg.1 TC0400008882.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Clariom D Assay TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Clariom D Assay TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Clariom D Assay TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Clariom D Assay TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Clariom D Assay TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Clariom D Assay TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Clariom D Assay TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Clariom D Assay TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Clariom D Assay TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Clariom D Assay TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Clariom D Assay TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Clariom D Assay TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Clariom D Assay TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Clariom D Assay TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Clariom D Assay TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Clariom D Assay TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Clariom D Assay TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Clariom D Assay TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Clariom D Assay TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Clariom D Assay TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Clariom D Assay TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Clariom D Assay TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Clariom D Assay TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Clariom D Assay TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Clariom D Assay TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Clariom D Assay TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Clariom D Assay TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Clariom D Assay TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Clariom D Assay TC0400008882.hg.1 ENST00000283131.3 Ensembl
Affymetrix Human Clariom D Assay TC0400008882.hg.1 ENST00000508573.1 Ensembl
Affymetrix Human Clariom D Assay TC0400008882.hg.1 ENST00000508573.1 Ensembl
Affymetrix Human Clariom D Assay TC0400008882.hg.1 ENST00000508573.1 Ensembl
Affymetrix Human Clariom D Assay TC0400008882.hg.1 ENST00000515531.1 Ensembl
Affymetrix Human Clariom D Assay TC0400008882.hg.1 ENST00000515531.1 Ensembl
Affymetrix Human Clariom D Assay TC0400008882.hg.1 ENST00000515531.1 Ensembl
Affymetrix Human Clariom D Assay TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Clariom D Assay TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Clariom D Assay TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Clariom D Assay TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Clariom D Assay TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Clariom D Assay TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Clariom D Assay TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Clariom D Assay TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Clariom D Assay TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Clariom D Assay TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Clariom D Assay TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Clariom D Assay TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Clariom D Assay TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Clariom D Assay TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Clariom D Assay TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Clariom D Assay TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Clariom D Assay TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Clariom D Assay TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Clariom D Assay TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Clariom D Assay TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Clariom D Assay TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Clariom D Assay TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Clariom D Assay TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Clariom D Assay TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Clariom D Assay TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Clariom D Assay TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Clariom D Assay TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Clariom D Assay TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Clariom D Assay TC0400008882.hg.1 NM_003601.3 Refseq
Affymetrix Human Clariom D Assay TC0400008882.hg.1 NM_003601.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-m9fgr:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline