This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01053788_m1
See other ABCG2 GE Assays ›
Gene Symbol:
ABCG2
Gene Name:
ATP binding cassette subfamily G member 2 (Junior blood group)
Gene Aliases:
ABC15, ABCP, BCRP, BCRP1, BMDP, CD338, CDw338, EST157481, GOUT1, MRX, MXR, MXR-1, MXR1, UAQTL1
Chromosome Location:
Chr.4: 88090264 - 88231417 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01053788_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ABCG2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005263354.3 XP_005263411.1 1 - 2 311 115
XM_017008852.1 XP_016864341.1 1 - 2 311 115
GenBank mRNA DQ459565.1 - 1 - 2 89 114
DQ459566.1 - 1 - 2 116
DQ459568.1 - 1 - 2 89 115
DQ459576.1 - 1 - 2 95 115
DQ459577.1 - 1 - 2 93 115
DQ459578.1 - 1 - 2 89 115
DQ459579.1 - 1 - 2 89 115
DQ459580.1 - 1 - 2 46 115
DQ459581.1 - 1 - 2 41 115
DQ459595.1 - 1 - 2 90 115
HQ387007.1 - 1 - 2 89 115

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the HQ387007.1, XM_005263354.3, XM_017008852.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

ATP-binding cassette (ABC) transporter primary active transporter transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
drug transmembrane transport
cellular iron ion homeostasis
response to iron ion
heme transport
response to drug
xenobiotic transport
urate metabolic process
drug export
response to folic acid
embryonic process involved in female pregnancy
cellular response to dexamethasone stimulus
transporter activity
protein binding
ATP binding
xenobiotic-transporting ATPase activity
heme transporter activity
ATPase activity, coupled to transmembrane movement of substances
protein homodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000237612.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000237612.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000237612.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000237612.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000237612.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000237612.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000237612.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000237612.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000237612.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000237612.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000237612.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000237612.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000237612.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000237612.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000237612.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000237612.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000237612.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000237612.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000237612.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000237612.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000237612.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000237612.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000237612.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000237612.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000237612.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000237612.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000503830.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000503830.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000503830.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000503830.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000503830.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000503830.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000505480.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000505480.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000505480.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000505480.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000505480.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000505480.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000515655.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000515655.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000515655.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000515655.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000515655.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000515655.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000515655.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000515655.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000515655.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000515655.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000515655.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000515655.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000515655.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000515655.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000515655.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000515655.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000515655.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000515655.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000515655.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000515655.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000515655.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 ENST00000515655.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 NM_001257386.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 NM_001257386.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 NM_001257386.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 NM_001257386.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 NM_001257386.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 NM_001257386.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 NM_001257386.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 NM_001257386.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 NM_001257386.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 NM_001257386.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 NM_001257386.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 NM_001257386.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 NM_001257386.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 NM_001257386.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 NM_001257386.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 NM_001257386.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 NM_001257386.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 NM_001257386.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 NM_001257386.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 NM_001257386.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 NM_001257386.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 NM_001257386.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 NM_004827.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 NM_004827.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 NM_004827.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 NM_004827.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 NM_004827.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 NM_004827.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 NM_004827.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 NM_004827.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 NM_004827.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 NM_004827.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 NM_004827.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 NM_004827.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 NM_004827.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 NM_004827.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 NM_004827.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 NM_004827.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 NM_004827.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 NM_004827.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 NM_004827.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 NM_004827.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 NM_004827.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 NM_004827.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 NM_004827.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 NM_004827.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011279.hg.1 NM_004827.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007686.hg.1 ENST00000611559.1 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000237612.7 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000237612.7 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000237612.7 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000237612.7 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000237612.7 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000237612.7 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000237612.7 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000237612.7 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000237612.7 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000237612.7 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000237612.7 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000237612.7 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000237612.7 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000237612.7 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000237612.7 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000237612.7 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000237612.7 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000237612.7 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000237612.7 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000237612.7 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000237612.7 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000237612.7 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000237612.7 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000237612.7 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000237612.7 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000237612.7 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000503830.1 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000503830.1 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000503830.1 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000503830.1 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000503830.1 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000503830.1 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000505480.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000505480.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000505480.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000505480.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000505480.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000505480.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000515655.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000515655.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000515655.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000515655.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000515655.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000515655.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000515655.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000515655.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000515655.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000515655.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000515655.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000515655.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000515655.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000515655.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000515655.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000515655.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000515655.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000515655.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000515655.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000515655.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000515655.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 ENST00000515655.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011279.hg.1 NM_001257386.2 Refseq
Affymetrix Human Clariom D Assay TC0400011279.hg.1 NM_001257386.2 Refseq
Affymetrix Human Clariom D Assay TC0400011279.hg.1 NM_001257386.2 Refseq
Affymetrix Human Clariom D Assay TC0400011279.hg.1 NM_001257386.2 Refseq
Affymetrix Human Clariom D Assay TC0400011279.hg.1 NM_001257386.2 Refseq
Affymetrix Human Clariom D Assay TC0400011279.hg.1 NM_001257386.2 Refseq
Affymetrix Human Clariom D Assay TC0400011279.hg.1 NM_001257386.2 Refseq
Affymetrix Human Clariom D Assay TC0400011279.hg.1 NM_001257386.2 Refseq
Affymetrix Human Clariom D Assay TC0400011279.hg.1 NM_001257386.2 Refseq
Affymetrix Human Clariom D Assay TC0400011279.hg.1 NM_001257386.2 Refseq
Affymetrix Human Clariom D Assay TC0400011279.hg.1 NM_001257386.2 Refseq
Affymetrix Human Clariom D Assay TC0400011279.hg.1 NM_001257386.2 Refseq
Affymetrix Human Clariom D Assay TC0400011279.hg.1 NM_001257386.2 Refseq
Affymetrix Human Clariom D Assay TC0400011279.hg.1 NM_001257386.2 Refseq
Affymetrix Human Clariom D Assay TC0400011279.hg.1 NM_001257386.2 Refseq
Affymetrix Human Clariom D Assay TC0400011279.hg.1 NM_001257386.2 Refseq
Affymetrix Human Clariom D Assay TC0400011279.hg.1 NM_001257386.2 Refseq
Affymetrix Human Clariom D Assay TC0400011279.hg.1 NM_001257386.2 Refseq
Affymetrix Human Clariom D Assay TC0400011279.hg.1 NM_001257386.2 Refseq
Affymetrix Human Clariom D Assay TC0400011279.hg.1 NM_001257386.2 Refseq
Affymetrix Human Clariom D Assay TC0400011279.hg.1 NM_001257386.2 Refseq
Affymetrix Human Clariom D Assay TC0400011279.hg.1 NM_001257386.2 Refseq
Affymetrix Human Clariom D Assay TC0400011279.hg.1 NM_004827.2 Refseq
Affymetrix Human Clariom D Assay TC0400011279.hg.1 NM_004827.2 Refseq
Affymetrix Human Clariom D Assay TC0400011279.hg.1 NM_004827.2 Refseq
Affymetrix Human Clariom D Assay TC0400011279.hg.1 NM_004827.2 Refseq
Affymetrix Human Clariom D Assay TC0400011279.hg.1 NM_004827.2 Refseq
Affymetrix Human Clariom D Assay TC0400011279.hg.1 NM_004827.2 Refseq
Affymetrix Human Clariom D Assay TC0400011279.hg.1 NM_004827.2 Refseq
Affymetrix Human Clariom D Assay TC0400011279.hg.1 NM_004827.2 Refseq
Affymetrix Human Clariom D Assay TC0400011279.hg.1 NM_004827.2 Refseq
Affymetrix Human Clariom D Assay TC0400011279.hg.1 NM_004827.2 Refseq
Affymetrix Human Clariom D Assay TC0400011279.hg.1 NM_004827.2 Refseq
Affymetrix Human Clariom D Assay TC0400011279.hg.1 NM_004827.2 Refseq
Affymetrix Human Clariom D Assay TC0400011279.hg.1 NM_004827.2 Refseq
Affymetrix Human Clariom D Assay TC0400011279.hg.1 NM_004827.2 Refseq
Affymetrix Human Clariom D Assay TC0400011279.hg.1 NM_004827.2 Refseq
Affymetrix Human Clariom D Assay TC0400011279.hg.1 NM_004827.2 Refseq
Affymetrix Human Clariom D Assay TC0400011279.hg.1 NM_004827.2 Refseq
Affymetrix Human Clariom D Assay TC0400011279.hg.1 NM_004827.2 Refseq
Affymetrix Human Clariom D Assay TC0400011279.hg.1 NM_004827.2 Refseq
Affymetrix Human Clariom D Assay TC0400011279.hg.1 NM_004827.2 Refseq
Affymetrix Human Clariom D Assay TC0400011279.hg.1 NM_004827.2 Refseq
Affymetrix Human Clariom D Assay TC0400011279.hg.1 NM_004827.2 Refseq
Affymetrix Human Clariom D Assay TC0400011279.hg.1 NM_004827.2 Refseq
Affymetrix Human Clariom D Assay TC0400011279.hg.1 NM_004827.2 Refseq
Affymetrix Human Clariom D Assay TC0400011279.hg.1 NM_004827.2 Refseq
Affymetrix Human Clariom D Assay TC1500007686.hg.1 ENST00000611559.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0