This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01053817_m1
See other SLCO6A1 GE Assays ›
Gene Symbol:
SLCO6A1
Gene Name:
solute carrier organic anion transporter family member 6A1
Gene Aliases:
CT48, GST, OATP-I, OATP6A1, OATPY
Chromosome Location:
Chr.5: 102371782 - 102502213 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01053817_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLCO6A1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001289002.1 NP_001275931.1 3 - 4 973 79
NM_173488.4 NP_775759.3 3 - 4 973 79
XM_005271874.3 XP_005271931.1 3 - 4 4170 79
XM_011543147.2 XP_011541449.1 3 - 4 4065 79
XM_011543148.2 XP_011541450.1 3 - 4 3933 79
XM_011543150.1 XP_011541452.1 2 - 3 183 79
XM_011543154.2 XP_011541456.1 3 - 4 4170 79
GenBank mRNA AF505657.1 - 3 - 4 944 79
AY273897.1 - 3 - 4 936 79

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC05001642.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SLCO6A1

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

sodium-independent organic anion transport
sodium-independent organic anion transmembrane transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR05022327.hg.1 ENST00000379807.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05022327.hg.1 ENST00000379810.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05022327.hg.1 ENST00000389019.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05022327.hg.1 ENST00000506729.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05022327.hg.1 ENST00000513675.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05022327.hg.1 ENST00000514551.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05022327.hg.1 NM_173488.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05022327.hg.1 TC05001642.hg.1 HTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline