This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01054475_g1
See other RALGAPA1 GE Assays ›
Gene Symbol:
RALGAPA1 RALGAPA1P1
Gene Name:
Ral GTPase activating protein catalytic alpha subunit 1
Ral GTPase activating protein catalytic alpha subunit 1 pseudogene 1
Gene Aliases:
GARNL1, GRIPE, RalGAPalpha1, TULIP1, p240
GARNL2, GARNL2P, RALGAPA1P, bA235C23.1
Chromosome Location:
Chr.14: 35538352 - 35809304 on Build GRCh38
Chromosome Location:
Chr.9: 105519824 - 105527319 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
-
Assay ID Hs01054475_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RALGAPA1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001283043.1 NP_001269972.1 32 - 33 4730 95
NM_001283044.1 NP_001269973.1 32 - 33 4832 95
NM_014990.1 NP_055805.1 31 - 32 4691 95
NM_194301.2 NP_919277.2 31 - 32 4691 95
XM_005267491.3 XP_005267548.1 32 - 33 4910 95
XM_005267492.3 XP_005267549.1 32 - 33 4910 95
XM_005267493.3 XP_005267550.1 31 - 32 4769 95
XM_005267498.4 XP_005267555.1 17 - 18 2520 95
XM_006720098.2 XP_006720161.1 32 - 33 6287 95
XM_006720099.2 XP_006720162.1 32 - 33 6287 95
XM_006720100.3 XP_006720163.1 31 - 32 6146 95
XM_006720101.2 XP_006720164.1 32 - 33 6287 95
XM_006720102.3 XP_006720165.1 32 - 33 5993 95
XM_006720103.2 XP_006720166.1 32 - 33 4910 95
XM_011536615.2 XP_011534917.1 32 - 33 6287 95
XM_017021141.1 XP_016876630.1 31 - 32 6146 95
XM_017021142.1 XP_016876631.1 31 - 32 6146 95
XM_017021143.1 XP_016876632.1 32 - 33 6287 95
XM_017021144.1 XP_016876633.1 32 - 33 4910 95
XM_017021145.1 XP_016876634.1 31 - 32 4769 95
XM_017021146.1 XP_016876635.1 17 - 18 3956 95
XM_017021147.1 XP_016876636.1 17 - 18 2842 95
GenBank mRNA AB511280.1 - 32 - 33 4441 95
AK126975.1 - 2 - 3 220 95
AL834362.1 - 8 - 9 802 95
AY596970.1 - 31 - 32 4691 95
AY596971.1 - 31 - 32 4691 95
BC042045.1 - 8 - 9 862 95
BC150596.1 - 32 - 33 4438 9056966 95
Gene Symbol: RALGAPA1P1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_104269.1 1 4730 95

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1400008945.hg.1
This assay targets the same location as HTA transcript cluster(s) TC14001038.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene RALGAPA1
Click here to find siRNAs for Gene RALGAPA1P1

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
regulation of small GTPase mediated signal transduction
activation of GTPase activity
GTPase activator activity
protein heterodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR14011872.hg.1 ENST00000258840.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14011872.hg.1 ENST00000307138.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14011872.hg.1 ENST00000335518.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14011872.hg.1 ENST00000382366.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14011872.hg.1 ENST00000389698.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14011872.hg.1 NM_014990.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR14011872.hg.1 NM_194301.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR14011872.hg.1 TC14001038.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1400165848.hg.1 ENST00000307138.10 Ensembl
Affymetrix Human Clariom D Assay PSR1400165848.hg.1 ENST00000382366.7 Ensembl
Affymetrix Human Clariom D Assay PSR1400165848.hg.1 ENST00000389698.7 Ensembl
Affymetrix Human Clariom D Assay PSR1400165848.hg.1 ENST00000553892.2 Ensembl
Affymetrix Human Clariom D Assay PSR1400165848.hg.1 ENST00000554259.5 Ensembl
Affymetrix Human Clariom D Assay PSR1400165848.hg.1 NM_001283043.2 Refseq
Affymetrix Human Clariom D Assay PSR1400165848.hg.1 NM_001283044.2 Refseq
Affymetrix Human Clariom D Assay PSR1400165848.hg.1 NM_014990.2 Refseq
Affymetrix Human Clariom D Assay PSR1400165848.hg.1 NM_194301.3 Refseq
Affymetrix Human Clariom D Assay PSR1400165848.hg.1 ENST00000307138.10 Ensembl
Affymetrix Human Clariom D Assay PSR1400165848.hg.1 ENST00000382366.7 Ensembl
Affymetrix Human Clariom D Assay PSR1400165848.hg.1 ENST00000389698.7 Ensembl
Affymetrix Human Clariom D Assay PSR1400165848.hg.1 ENST00000553892.2 Ensembl
Affymetrix Human Clariom D Assay PSR1400165848.hg.1 ENST00000554259.5 Ensembl
Affymetrix Human Clariom D Assay PSR1400165848.hg.1 NM_001283043.2 Refseq
Affymetrix Human Clariom D Assay PSR1400165848.hg.1 NM_001283044.2 Refseq
Affymetrix Human Clariom D Assay PSR1400165848.hg.1 NM_014990.2 Refseq
Affymetrix Human Clariom D Assay PSR1400165848.hg.1 NM_194301.3 Refseq
Affymetrix Human Clariom D Assay PSR1400165848.hg.1 TC1400008945.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-x4xlz:80/100.66.78.26:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0