This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01055245_g1
See other DNA2 GE Assays ›
Gene Symbol:
DNA2
Gene Name:
DNA replication helicase/nuclease 2
Gene Aliases:
DNA2L, hDNA2
Chromosome Location:
Chr.10: 68414064 - 68472521 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01055245_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DNA2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001080449.2 NP_001073918.2 18 - 19 2898 99
NR_102264.1 19 - 20 2763 99
XM_006717680.3 XP_006717743.1 19 - 20 3278 99
XM_011539417.1 XP_011537719.1 12 - 13 1822 99
XM_017015799.1 XP_016871288.1 15 - 16 2287 99
GenBank mRNA AK307939.1 - 7 - 8 848 99
BC028188.1 - 5 - 6 764 99
BC041115.2 - 6 - 7 916 99
BC053574.1 - 8 - 9 1092 99
BC063664.1 - 14 - 15 2097 99
BC073945.1 - 4 - 5 505 99
BC111740.1 - 19 - 20 2763 40038149 99
D42046.1 - 18 - 19 2840 99

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000010847.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10001347.hg.1

Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

DNA replication checkpoint
telomere maintenance via recombination
telomere maintenance
DNA double-strand break processing
DNA synthesis involved in DNA repair
strand displacement
DNA replication
mitochondrial DNA replication
base-excision repair
DNA duplex unwinding
DNA replication, Okazaki fragment processing
DNA replication, removal of RNA primer
mitochondrial DNA repair
positive regulation of DNA replication
nucleic acid phosphodiester bond hydrolysis
regulation of signal transduction by p53 class mediator
mitotic telomere maintenance via semi-conservative replication
DNA binding
DNA helicase activity
helicase activity
nuclease activity
protein binding
ATP binding
ATPase activity
site-specific endodeoxyribonuclease activity, specific for altered base
5'-flap endonuclease activity
5'-3' DNA helicase activity
single-stranded DNA-dependent ATPase activity
metal ion binding
4 iron, 4 sulfur cluster binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000358410.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000358410.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000358410.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000358410.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000358410.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000358410.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000358410.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000358410.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000358410.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000358410.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000358410.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000358410.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000358410.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000358410.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000358410.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000358410.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000358410.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000358410.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000358410.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000358410.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000358410.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000358410.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000358410.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000358410.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000358410.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000358410.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000358410.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000358410.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000399179.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000399179.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000399179.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000399179.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000399179.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000399179.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000399179.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000399179.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000399179.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000399179.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000399179.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000399179.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000399179.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000399179.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000399179.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000399179.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000399179.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000399179.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000399179.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000399179.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000399179.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000399179.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000399179.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000399179.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000399179.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000399180.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000399180.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000399180.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000399180.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000399180.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000399180.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000399180.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000399180.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000399180.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000399180.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000399180.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000399180.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000399180.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000399180.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000399180.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000399180.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000399180.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000399180.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000399180.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000399180.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000399180.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000399180.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000399180.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000399180.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000399180.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000440722.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000440722.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000440722.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000440722.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000440722.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000440722.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000440722.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000440722.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000440722.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000478029.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000478029.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000478029.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000550357.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000550357.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000550357.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000550357.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000550545.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000550545.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000550545.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000551118.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000551118.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000551118.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000551118.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000551118.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000551118.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000551118.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000551118.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000551118.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000551118.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000551118.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000551118.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000551118.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000551118.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000551118.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000551118.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000551118.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000551118.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000551118.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000551118.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 ENST00000551118.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NM_001080449.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NM_001080449.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NM_001080449.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NM_001080449.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NM_001080449.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NM_001080449.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NM_001080449.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NM_001080449.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NM_001080449.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NM_001080449.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NM_001080449.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NM_001080449.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NM_001080449.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NM_001080449.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NM_001080449.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NM_001080449.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NM_001080449.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NM_001080449.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NM_001080449.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NM_001080449.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NM_001080449.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NM_001080449.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NM_001080449.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NM_001080449.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NM_001080449.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NM_001080449.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NM_001080449.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NM_001080449.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NM_001080449.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NR_102264.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NR_102264.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NR_102264.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NR_102264.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NR_102264.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NR_102264.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NR_102264.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NR_102264.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NR_102264.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NR_102264.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NR_102264.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NR_102264.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NR_102264.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NR_102264.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NR_102264.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NR_102264.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NR_102264.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NR_102264.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NR_102264.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NR_102264.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NR_102264.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NR_102264.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NR_102264.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NR_102264.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NR_102264.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010847.hg.1 NR_102264.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0