This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01055329_m1
See other GDNF GE Assays ›
Gene Symbol:
GDNF
Gene Name:
glial cell derived neurotrophic factor
Gene Aliases:
ATF1, ATF2, HFB1-GDNF, HSCR3
Chromosome Location:
Chr.5: 37812677 - 37840044 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01055329_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GDNF
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001190469.1 NP_001177398.1 2 - 3 293 142
NM_199231.2 NP_954701.1 2 - 3 196 142
XM_017009337.1 XP_016864826.1 2 - 3 637 142
GenBank mRNA AJ001899.1 - 2 - 3 196 142
AJ001900.1 - 1 - 2 185 142
AY052832.1 - 1 - 2 73 142
KU178094.1 - 1 - 2 73 142

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0500010523.hg.1
This assay targets the same location as HTA transcript cluster(s) TC05001280.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GDNF

Panther Classification:

Molecular Function -

neurotrophic factor

Gene Ontology Categories:

Function(s) Process(es)

MAPK cascade
metanephros development
branching involved in ureteric bud morphogenesis
neural crest cell migration
organ induction
postsynaptic membrane organization
mesenchymal to epithelial transition involved in metanephros morphogenesis
signal transduction
nervous system development
positive regulation of cell proliferation
adult locomotory behavior
regulation of gene expression
postganglionic parasympathetic fiber development
peristalsis
neuron projection development
positive regulation of monooxygenase activity
positive regulation of dopamine secretion
negative regulation of apoptotic process
negative regulation of neuron apoptotic process
positive regulation of GTPase activity
positive regulation of cell differentiation
positive regulation of transcription from RNA polymerase II promoter
mRNA stabilization
enteric nervous system development
sympathetic nervous system development
regulation of dopamine uptake involved in synaptic transmission
ureteric bud formation
regulation of morphogenesis of a branching structure
positive regulation of ureteric bud formation
positive regulation of mesenchymal to epithelial transition involved in metanephros morphogenesis
positive regulation of branching involved in ureteric bud morphogenesis
regulation of stem cell differentiation
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
Ras guanyl-nucleotide exchange factor activity
receptor binding
protein binding
growth factor activity
protein homodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR05017995.hg.1 TC05001280.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR05017995.hg.1 ENST00000344622.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05017995.hg.1 NM_199231.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 ENST00000326524.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 ENST00000326524.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 ENST00000326524.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 ENST00000326524.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 ENST00000326524.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 ENST00000326524.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 ENST00000326524.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 ENST00000326524.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 ENST00000344622.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 ENST00000344622.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 ENST00000344622.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 ENST00000344622.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 ENST00000344622.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 ENST00000344622.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 ENST00000344622.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 ENST00000381826.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 ENST00000381826.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 ENST00000381826.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 ENST00000381826.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 ENST00000381826.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 ENST00000381826.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 ENST00000427982.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 ENST00000427982.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 ENST00000427982.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 ENST00000427982.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 ENST00000427982.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 ENST00000427982.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 ENST00000427982.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 ENST00000502572.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 ENST00000502572.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 ENST00000502572.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 ENST00000502572.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 ENST00000510177.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 ENST00000510177.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 ENST00000510177.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 ENST00000510177.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 ENST00000515058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 ENST00000515058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 ENST00000515058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 ENST00000515058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 ENST00000515058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 ENST00000620847.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 ENST00000620847.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 ENST00000620847.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 ENST00000620847.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 ENST00000620847.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 ENST00000620847.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 ENST00000620847.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 NM_000514.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 NM_000514.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 NM_000514.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 NM_000514.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 NM_000514.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 NM_000514.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 NM_000514.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 NM_000514.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 NM_001190468.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 NM_001190468.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 NM_001190468.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 NM_001190468.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 NM_001190468.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 NM_001190468.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 NM_001190468.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 NM_001190468.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 NM_001190468.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 NM_001190469.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 NM_001190469.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 NM_001190469.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 NM_001190469.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 NM_001190469.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 NM_001190469.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 NM_001190469.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 NM_001190469.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 NM_001278098.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 NM_001278098.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 NM_001278098.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 NM_001278098.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 NM_001278098.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 NM_001278098.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 NM_001278098.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 NM_199231.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 NM_199231.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 NM_199231.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 NM_199231.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 NM_199231.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 NM_199231.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010523.hg.1 NM_199231.2 Refseq
Affymetrix Human Clariom D Assay PSR0500173364.hg.1 ENST00000344622.8 Ensembl
Affymetrix Human Clariom D Assay PSR0500173364.hg.1 ENST00000502572.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500173364.hg.1 NM_199231.2 Refseq
Affymetrix Human Clariom D Assay PSR0500173364.hg.1 ENST00000344622.8 Ensembl
Affymetrix Human Clariom D Assay PSR0500173364.hg.1 ENST00000502572.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500173364.hg.1 NM_199231.2 Refseq
Affymetrix Human Clariom D Assay PSR0500173364.hg.1 TC0500010523.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0500010523.hg.1 ENST00000326524.6 Ensembl
Affymetrix Human Clariom D Assay TC0500010523.hg.1 ENST00000326524.6 Ensembl
Affymetrix Human Clariom D Assay TC0500010523.hg.1 ENST00000326524.6 Ensembl
Affymetrix Human Clariom D Assay TC0500010523.hg.1 ENST00000326524.6 Ensembl
Affymetrix Human Clariom D Assay TC0500010523.hg.1 ENST00000326524.6 Ensembl
Affymetrix Human Clariom D Assay TC0500010523.hg.1 ENST00000326524.6 Ensembl
Affymetrix Human Clariom D Assay TC0500010523.hg.1 ENST00000326524.6 Ensembl
Affymetrix Human Clariom D Assay TC0500010523.hg.1 ENST00000326524.6 Ensembl
Affymetrix Human Clariom D Assay TC0500010523.hg.1 ENST00000344622.8 Ensembl
Affymetrix Human Clariom D Assay TC0500010523.hg.1 ENST00000344622.8 Ensembl
Affymetrix Human Clariom D Assay TC0500010523.hg.1 ENST00000344622.8 Ensembl
Affymetrix Human Clariom D Assay TC0500010523.hg.1 ENST00000344622.8 Ensembl
Affymetrix Human Clariom D Assay TC0500010523.hg.1 ENST00000344622.8 Ensembl
Affymetrix Human Clariom D Assay TC0500010523.hg.1 ENST00000344622.8 Ensembl
Affymetrix Human Clariom D Assay TC0500010523.hg.1 ENST00000344622.8 Ensembl
Affymetrix Human Clariom D Assay TC0500010523.hg.1 ENST00000381826.8 Ensembl
Affymetrix Human Clariom D Assay TC0500010523.hg.1 ENST00000381826.8 Ensembl
Affymetrix Human Clariom D Assay TC0500010523.hg.1 ENST00000381826.8 Ensembl
Affymetrix Human Clariom D Assay TC0500010523.hg.1 ENST00000381826.8 Ensembl
Affymetrix Human Clariom D Assay TC0500010523.hg.1 ENST00000381826.8 Ensembl
Affymetrix Human Clariom D Assay TC0500010523.hg.1 ENST00000381826.8 Ensembl
Affymetrix Human Clariom D Assay TC0500010523.hg.1 ENST00000427982.5 Ensembl
Affymetrix Human Clariom D Assay TC0500010523.hg.1 ENST00000427982.5 Ensembl
Affymetrix Human Clariom D Assay TC0500010523.hg.1 ENST00000427982.5 Ensembl
Affymetrix Human Clariom D Assay TC0500010523.hg.1 ENST00000427982.5 Ensembl
Affymetrix Human Clariom D Assay TC0500010523.hg.1 ENST00000427982.5 Ensembl
Affymetrix Human Clariom D Assay TC0500010523.hg.1 ENST00000427982.5 Ensembl
Affymetrix Human Clariom D Assay TC0500010523.hg.1 ENST00000427982.5 Ensembl
Affymetrix Human Clariom D Assay TC0500010523.hg.1 ENST00000502572.1 Ensembl
Affymetrix Human Clariom D Assay TC0500010523.hg.1 ENST00000502572.1 Ensembl
Affymetrix Human Clariom D Assay TC0500010523.hg.1 ENST00000502572.1 Ensembl
Affymetrix Human Clariom D Assay TC0500010523.hg.1 ENST00000502572.1 Ensembl
Affymetrix Human Clariom D Assay TC0500010523.hg.1 ENST00000510177.5 Ensembl
Affymetrix Human Clariom D Assay TC0500010523.hg.1 ENST00000510177.5 Ensembl
Affymetrix Human Clariom D Assay TC0500010523.hg.1 ENST00000510177.5 Ensembl
Affymetrix Human Clariom D Assay TC0500010523.hg.1 ENST00000510177.5 Ensembl
Affymetrix Human Clariom D Assay TC0500010523.hg.1 ENST00000515058.5 Ensembl
Affymetrix Human Clariom D Assay TC0500010523.hg.1 ENST00000515058.5 Ensembl
Affymetrix Human Clariom D Assay TC0500010523.hg.1 ENST00000515058.5 Ensembl
Affymetrix Human Clariom D Assay TC0500010523.hg.1 ENST00000515058.5 Ensembl
Affymetrix Human Clariom D Assay TC0500010523.hg.1 ENST00000515058.5 Ensembl
Affymetrix Human Clariom D Assay TC0500010523.hg.1 ENST00000620847.1 Ensembl
Affymetrix Human Clariom D Assay TC0500010523.hg.1 ENST00000620847.1 Ensembl
Affymetrix Human Clariom D Assay TC0500010523.hg.1 ENST00000620847.1 Ensembl
Affymetrix Human Clariom D Assay TC0500010523.hg.1 ENST00000620847.1 Ensembl
Affymetrix Human Clariom D Assay TC0500010523.hg.1 ENST00000620847.1 Ensembl
Affymetrix Human Clariom D Assay TC0500010523.hg.1 ENST00000620847.1 Ensembl
Affymetrix Human Clariom D Assay TC0500010523.hg.1 ENST00000620847.1 Ensembl
Affymetrix Human Clariom D Assay TC0500010523.hg.1 NM_000514.3 Refseq
Affymetrix Human Clariom D Assay TC0500010523.hg.1 NM_000514.3 Refseq
Affymetrix Human Clariom D Assay TC0500010523.hg.1 NM_000514.3 Refseq
Affymetrix Human Clariom D Assay TC0500010523.hg.1 NM_000514.3 Refseq
Affymetrix Human Clariom D Assay TC0500010523.hg.1 NM_000514.3 Refseq
Affymetrix Human Clariom D Assay TC0500010523.hg.1 NM_000514.3 Refseq
Affymetrix Human Clariom D Assay TC0500010523.hg.1 NM_000514.3 Refseq
Affymetrix Human Clariom D Assay TC0500010523.hg.1 NM_000514.3 Refseq
Affymetrix Human Clariom D Assay TC0500010523.hg.1 NM_001190468.1 Refseq
Affymetrix Human Clariom D Assay TC0500010523.hg.1 NM_001190468.1 Refseq
Affymetrix Human Clariom D Assay TC0500010523.hg.1 NM_001190468.1 Refseq
Affymetrix Human Clariom D Assay TC0500010523.hg.1 NM_001190468.1 Refseq
Affymetrix Human Clariom D Assay TC0500010523.hg.1 NM_001190468.1 Refseq
Affymetrix Human Clariom D Assay TC0500010523.hg.1 NM_001190468.1 Refseq
Affymetrix Human Clariom D Assay TC0500010523.hg.1 NM_001190468.1 Refseq
Affymetrix Human Clariom D Assay TC0500010523.hg.1 NM_001190468.1 Refseq
Affymetrix Human Clariom D Assay TC0500010523.hg.1 NM_001190468.1 Refseq
Affymetrix Human Clariom D Assay TC0500010523.hg.1 NM_001190469.1 Refseq
Affymetrix Human Clariom D Assay TC0500010523.hg.1 NM_001190469.1 Refseq
Affymetrix Human Clariom D Assay TC0500010523.hg.1 NM_001190469.1 Refseq
Affymetrix Human Clariom D Assay TC0500010523.hg.1 NM_001190469.1 Refseq
Affymetrix Human Clariom D Assay TC0500010523.hg.1 NM_001190469.1 Refseq
Affymetrix Human Clariom D Assay TC0500010523.hg.1 NM_001190469.1 Refseq
Affymetrix Human Clariom D Assay TC0500010523.hg.1 NM_001190469.1 Refseq
Affymetrix Human Clariom D Assay TC0500010523.hg.1 NM_001190469.1 Refseq
Affymetrix Human Clariom D Assay TC0500010523.hg.1 NM_001278098.1 Refseq
Affymetrix Human Clariom D Assay TC0500010523.hg.1 NM_001278098.1 Refseq
Affymetrix Human Clariom D Assay TC0500010523.hg.1 NM_001278098.1 Refseq
Affymetrix Human Clariom D Assay TC0500010523.hg.1 NM_001278098.1 Refseq
Affymetrix Human Clariom D Assay TC0500010523.hg.1 NM_001278098.1 Refseq
Affymetrix Human Clariom D Assay TC0500010523.hg.1 NM_001278098.1 Refseq
Affymetrix Human Clariom D Assay TC0500010523.hg.1 NM_001278098.1 Refseq
Affymetrix Human Clariom D Assay TC0500010523.hg.1 NM_199231.2 Refseq
Affymetrix Human Clariom D Assay TC0500010523.hg.1 NM_199231.2 Refseq
Affymetrix Human Clariom D Assay TC0500010523.hg.1 NM_199231.2 Refseq
Affymetrix Human Clariom D Assay TC0500010523.hg.1 NM_199231.2 Refseq
Affymetrix Human Clariom D Assay TC0500010523.hg.1 NM_199231.2 Refseq
Affymetrix Human Clariom D Assay TC0500010523.hg.1 NM_199231.2 Refseq
Affymetrix Human Clariom D Assay TC0500010523.hg.1 NM_199231.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7kcm7:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0