This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01055783_g1
See other PSMG1 GE Assays ›
Gene Symbol:
PSMG1
Gene Name:
proteasome assembly chaperone 1
Gene Aliases:
C21LRP, DSCR2, LRPC21, PAC-1, PAC1
Chromosome Location:
Chr.21: 39175446 - 39183514 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01055783_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PSMG1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001320795.1 NP_001307724.1 3 - 4 711 150
NM_003720.3 NP_003711.1 3 - 4 523 150
NR_049728.1 2 - 3 416 150
GenBank mRNA AF417108.1 - 2 - 3 287 150
AJ006291.1 - 3 - 4 457 150
AK308614.1 - 2 - 3 394 150
AK315406.1 - 3 - 4 425 150
BC003619.2 - 3 - 4 440 2961575 150
BC010424.1 - 3 - 4 432 3343732 150
BC011755.2 - 3 - 4 440 3531528 150
BC012809.2 - 3 - 4 440 2961575 150
BM013494.1 - 3 - 4 391 150
BM460414.1 - 3 - 4 416 150
CR541821.1 - 3 - 4 394 150
CR541852.1 - 3 - 4 394 150

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2100008164.hg.1
This assay targets the same location as HTA transcript cluster(s) TC21000458.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PSMG1

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

cerebellar granule cell precursor proliferation
proteasome assembly
proteasome core complex assembly
protein binding
proteasome binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 ENST00000331573.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 ENST00000331573.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 ENST00000331573.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 ENST00000331573.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 ENST00000331573.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 ENST00000331573.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 ENST00000331573.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 ENST00000331573.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 ENST00000331573.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 ENST00000331573.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 ENST00000331573.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 ENST00000331573.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 ENST00000331573.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 ENST00000331573.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 ENST00000380900.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 ENST00000380900.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 ENST00000380900.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 ENST00000380900.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 ENST00000380900.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 ENST00000380900.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 ENST00000380900.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 ENST00000380900.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 ENST00000380900.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 ENST00000411828.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 ENST00000411828.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 ENST00000411828.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 ENST00000411828.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 ENST00000411828.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 ENST00000411828.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 ENST00000411828.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 ENST00000411828.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 ENST00000431628.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 ENST00000431628.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 ENST00000431628.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 ENST00000431628.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 ENST00000431628.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 ENST00000431628.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 ENST00000431628.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 ENST00000481921.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 ENST00000481921.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 ENST00000481921.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 ENST00000481921.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 ENST00000481921.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 NM_001261824.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 NM_001261824.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 NM_001261824.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 NM_001261824.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 NM_001261824.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 NM_001261824.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 NM_001261824.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 NM_001261824.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 NM_001261824.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 NM_001261824.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 NM_001261824.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 NM_001261824.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 NM_003720.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 NM_003720.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 NM_003720.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 NM_003720.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 NM_003720.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 NM_003720.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 NM_003720.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 NM_003720.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 NM_003720.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 NM_003720.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 NM_003720.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 NM_003720.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 NM_203433.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 NM_203433.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 NM_203433.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 NM_203433.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 NM_203433.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 NM_203433.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 NM_203433.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 NM_203433.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 NM_203433.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 NM_203433.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 NM_203433.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 NR_049728.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 NR_049728.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 NR_049728.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 NR_049728.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 NR_049728.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 NR_049728.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 NR_049728.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 NR_049728.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 NR_049728.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 NR_049728.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008164.hg.1 NR_049728.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR21005337.hg.1 ENST00000331573.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR21005337.hg.1 NM_003720.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR21005337.hg.1 TC21000458.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC2100008164.hg.1 ENST00000331573.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008164.hg.1 ENST00000331573.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008164.hg.1 ENST00000331573.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008164.hg.1 ENST00000331573.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008164.hg.1 ENST00000331573.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008164.hg.1 ENST00000331573.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008164.hg.1 ENST00000331573.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008164.hg.1 ENST00000331573.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008164.hg.1 ENST00000331573.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008164.hg.1 ENST00000331573.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008164.hg.1 ENST00000331573.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008164.hg.1 ENST00000331573.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008164.hg.1 ENST00000331573.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008164.hg.1 ENST00000331573.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008164.hg.1 ENST00000380900.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008164.hg.1 ENST00000380900.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008164.hg.1 ENST00000380900.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008164.hg.1 ENST00000380900.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008164.hg.1 ENST00000380900.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008164.hg.1 ENST00000380900.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008164.hg.1 ENST00000380900.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008164.hg.1 ENST00000380900.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008164.hg.1 ENST00000380900.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008164.hg.1 ENST00000411828.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008164.hg.1 ENST00000411828.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008164.hg.1 ENST00000411828.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008164.hg.1 ENST00000411828.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008164.hg.1 ENST00000411828.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008164.hg.1 ENST00000411828.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008164.hg.1 ENST00000411828.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008164.hg.1 ENST00000411828.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008164.hg.1 ENST00000431628.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008164.hg.1 ENST00000431628.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008164.hg.1 ENST00000431628.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008164.hg.1 ENST00000431628.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008164.hg.1 ENST00000431628.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008164.hg.1 ENST00000431628.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008164.hg.1 ENST00000431628.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008164.hg.1 ENST00000481921.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008164.hg.1 ENST00000481921.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008164.hg.1 ENST00000481921.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008164.hg.1 ENST00000481921.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008164.hg.1 ENST00000481921.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008164.hg.1 NM_001261824.1 Refseq
Affymetrix Human Clariom D Assay TC2100008164.hg.1 NM_001261824.1 Refseq
Affymetrix Human Clariom D Assay TC2100008164.hg.1 NM_001261824.1 Refseq
Affymetrix Human Clariom D Assay TC2100008164.hg.1 NM_001261824.1 Refseq
Affymetrix Human Clariom D Assay TC2100008164.hg.1 NM_001261824.1 Refseq
Affymetrix Human Clariom D Assay TC2100008164.hg.1 NM_001261824.1 Refseq
Affymetrix Human Clariom D Assay TC2100008164.hg.1 NM_001261824.1 Refseq
Affymetrix Human Clariom D Assay TC2100008164.hg.1 NM_001261824.1 Refseq
Affymetrix Human Clariom D Assay TC2100008164.hg.1 NM_001261824.1 Refseq
Affymetrix Human Clariom D Assay TC2100008164.hg.1 NM_001261824.1 Refseq
Affymetrix Human Clariom D Assay TC2100008164.hg.1 NM_001261824.1 Refseq
Affymetrix Human Clariom D Assay TC2100008164.hg.1 NM_001261824.1 Refseq
Affymetrix Human Clariom D Assay TC2100008164.hg.1 NM_003720.3 Refseq
Affymetrix Human Clariom D Assay TC2100008164.hg.1 NM_003720.3 Refseq
Affymetrix Human Clariom D Assay TC2100008164.hg.1 NM_003720.3 Refseq
Affymetrix Human Clariom D Assay TC2100008164.hg.1 NM_003720.3 Refseq
Affymetrix Human Clariom D Assay TC2100008164.hg.1 NM_003720.3 Refseq
Affymetrix Human Clariom D Assay TC2100008164.hg.1 NM_003720.3 Refseq
Affymetrix Human Clariom D Assay TC2100008164.hg.1 NM_003720.3 Refseq
Affymetrix Human Clariom D Assay TC2100008164.hg.1 NM_003720.3 Refseq
Affymetrix Human Clariom D Assay TC2100008164.hg.1 NM_003720.3 Refseq
Affymetrix Human Clariom D Assay TC2100008164.hg.1 NM_003720.3 Refseq
Affymetrix Human Clariom D Assay TC2100008164.hg.1 NM_003720.3 Refseq
Affymetrix Human Clariom D Assay TC2100008164.hg.1 NM_003720.3 Refseq
Affymetrix Human Clariom D Assay TC2100008164.hg.1 NM_203433.2 Refseq
Affymetrix Human Clariom D Assay TC2100008164.hg.1 NM_203433.2 Refseq
Affymetrix Human Clariom D Assay TC2100008164.hg.1 NM_203433.2 Refseq
Affymetrix Human Clariom D Assay TC2100008164.hg.1 NM_203433.2 Refseq
Affymetrix Human Clariom D Assay TC2100008164.hg.1 NM_203433.2 Refseq
Affymetrix Human Clariom D Assay TC2100008164.hg.1 NM_203433.2 Refseq
Affymetrix Human Clariom D Assay TC2100008164.hg.1 NM_203433.2 Refseq
Affymetrix Human Clariom D Assay TC2100008164.hg.1 NM_203433.2 Refseq
Affymetrix Human Clariom D Assay TC2100008164.hg.1 NM_203433.2 Refseq
Affymetrix Human Clariom D Assay TC2100008164.hg.1 NM_203433.2 Refseq
Affymetrix Human Clariom D Assay TC2100008164.hg.1 NM_203433.2 Refseq
Affymetrix Human Clariom D Assay TC2100008164.hg.1 NR_049728.1 Refseq
Affymetrix Human Clariom D Assay TC2100008164.hg.1 NR_049728.1 Refseq
Affymetrix Human Clariom D Assay TC2100008164.hg.1 NR_049728.1 Refseq
Affymetrix Human Clariom D Assay TC2100008164.hg.1 NR_049728.1 Refseq
Affymetrix Human Clariom D Assay TC2100008164.hg.1 NR_049728.1 Refseq
Affymetrix Human Clariom D Assay TC2100008164.hg.1 NR_049728.1 Refseq
Affymetrix Human Clariom D Assay TC2100008164.hg.1 NR_049728.1 Refseq
Affymetrix Human Clariom D Assay TC2100008164.hg.1 NR_049728.1 Refseq
Affymetrix Human Clariom D Assay TC2100008164.hg.1 NR_049728.1 Refseq
Affymetrix Human Clariom D Assay TC2100008164.hg.1 NR_049728.1 Refseq
Affymetrix Human Clariom D Assay TC2100008164.hg.1 NR_049728.1 Refseq
Affymetrix Human Clariom D Assay PSR2100155036.hg.1 ENST00000331573.7 Ensembl
Affymetrix Human Clariom D Assay PSR2100155036.hg.1 ENST00000411828.5 Ensembl
Affymetrix Human Clariom D Assay PSR2100155036.hg.1 ENST00000431628.1 Ensembl
Affymetrix Human Clariom D Assay PSR2100155036.hg.1 NM_001261824.1 Refseq
Affymetrix Human Clariom D Assay PSR2100155036.hg.1 NM_003720.3 Refseq
Affymetrix Human Clariom D Assay PSR2100155036.hg.1 NR_049728.1 Refseq
Affymetrix Human Clariom D Assay PSR2100155036.hg.1 TC2100008164.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2100155036.hg.1 ENST00000331573.7 Ensembl
Affymetrix Human Clariom D Assay PSR2100155036.hg.1 ENST00000411828.5 Ensembl
Affymetrix Human Clariom D Assay PSR2100155036.hg.1 ENST00000431628.1 Ensembl
Affymetrix Human Clariom D Assay PSR2100155036.hg.1 NM_001261824.1 Refseq
Affymetrix Human Clariom D Assay PSR2100155036.hg.1 NM_003720.3 Refseq
Affymetrix Human Clariom D Assay PSR2100155036.hg.1 NR_049728.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0