This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01056225_m1
See other TP73 GE Assays ›
Gene Symbol:
TP73
Gene Name:
tumor protein p73
Gene Aliases:
P73
Chromosome Location:
Chr.1: 3652565 - 3736201 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01056225_m1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TP73
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001126240.2 NP_001119712.1 9 - 10 1434 59
NM_001126241.2 NP_001119713.1 9 - 10 1434 59
NM_001204184.1 NP_001191113.1 11 - 12 1457 59
NM_001204192.1 NP_001191121.1 9 - 10 1267 59
NM_005427.3 NP_005418.1 11 - 12 1457 59
GenBank mRNA AB055065.1 - 9 - 10 1434 59
AB055066.1 - 9 - 10 1434 59
AK302118.1 - 9 - 10 1255 59
AK304784.1 - 9 - 10 1384 59
AY040827.1 - 9 - 10 1434 59
AY040828.1 - 9 - 10 1434 59
BC117251.1 - 11 - 12 1457 40125802 59
BC117253.1 - 11 - 12 1457 40125804 59
Y11416.1 - 11 - 12 1457 59

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100006620.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01000071.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TP73

Panther Classification:

Molecular Function -

P53-like transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
activation of MAPK activity
kidney development
mismatch repair
transcription from RNA polymerase II promoter
cellular response to DNA damage stimulus
DNA damage response, signal transduction by p53 class mediator resulting in transcription of p21 class mediator
cell cycle arrest
regulation of mitotic cell cycle
intrinsic apoptotic signaling pathway in response to DNA damage
response to X-ray
response to organonitrogen compound
response to gamma radiation
regulation of gene expression
viral process
mitotic G1 DNA damage checkpoint
cellular response to UV
response to drug
intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
regulation of apoptotic process
negative regulation of JUN kinase activity
negative regulation of neuron apoptotic process
negative regulation of neuron differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of oligodendrocyte differentiation
protein tetramerization
negative regulation of cardiac muscle cell proliferation
positive regulation of cell cycle arrest
positive regulation of protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
regulation of signal transduction by p53 class mediator
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
p53 binding
chromatin binding
damaged DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
transcription factor binding
protein kinase binding
identical protein binding
transcription regulatory region DNA binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR01001142.hg.1 TC01000071.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01001142.hg.1 ENST00000354437.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01001142.hg.1 ENST00000357733.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01001142.hg.1 ENST00000378280.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01001142.hg.1 ENST00000378285.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01001142.hg.1 ENST00000378288.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01001142.hg.1 ENST00000378290.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01001142.hg.1 ENST00000378295.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01001142.hg.1 NM_001126240.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01001142.hg.1 NM_001126241.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01001142.hg.1 NM_001126242.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01001142.hg.1 NM_001204184.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01001142.hg.1 NM_001204185.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01001142.hg.1 NM_001204187.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01001142.hg.1 NM_001204190.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01001142.hg.1 NM_001204192.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01001142.hg.1 NM_005427.3 Refseq
Affymetrix Human Clariom D Assay PSR0100145801.hg.1 ENST00000354437.8 Ensembl
Affymetrix Human Clariom D Assay PSR0100145801.hg.1 ENST00000357733.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100145801.hg.1 ENST00000378280.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100145801.hg.1 ENST00000378285.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100145801.hg.1 ENST00000378288.8 Ensembl
Affymetrix Human Clariom D Assay PSR0100145801.hg.1 ENST00000378290.4 Ensembl
Affymetrix Human Clariom D Assay PSR0100145801.hg.1 ENST00000378295.8 Ensembl
Affymetrix Human Clariom D Assay PSR0100145801.hg.1 ENST00000603362.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100145801.hg.1 ENST00000604566.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100145801.hg.1 NM_001126240.2 Refseq
Affymetrix Human Clariom D Assay PSR0100145801.hg.1 NM_001126241.2 Refseq
Affymetrix Human Clariom D Assay PSR0100145801.hg.1 NM_001126242.2 Refseq
Affymetrix Human Clariom D Assay PSR0100145801.hg.1 NM_001204184.1 Refseq
Affymetrix Human Clariom D Assay PSR0100145801.hg.1 NM_001204185.1 Refseq
Affymetrix Human Clariom D Assay PSR0100145801.hg.1 NM_001204187.1 Refseq
Affymetrix Human Clariom D Assay PSR0100145801.hg.1 NM_001204190.1 Refseq
Affymetrix Human Clariom D Assay PSR0100145801.hg.1 NM_001204192.1 Refseq
Affymetrix Human Clariom D Assay PSR0100145801.hg.1 NM_005427.3 Refseq
Affymetrix Human Clariom D Assay PSR0100145801.hg.1 ENST00000354437.8 Ensembl
Affymetrix Human Clariom D Assay PSR0100145801.hg.1 ENST00000357733.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100145801.hg.1 ENST00000378280.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100145801.hg.1 ENST00000378285.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100145801.hg.1 ENST00000378288.8 Ensembl
Affymetrix Human Clariom D Assay PSR0100145801.hg.1 ENST00000378290.4 Ensembl
Affymetrix Human Clariom D Assay PSR0100145801.hg.1 ENST00000378295.8 Ensembl
Affymetrix Human Clariom D Assay PSR0100145801.hg.1 ENST00000603362.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100145801.hg.1 ENST00000604566.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100145801.hg.1 NM_001126240.2 Refseq
Affymetrix Human Clariom D Assay PSR0100145801.hg.1 NM_001126241.2 Refseq
Affymetrix Human Clariom D Assay PSR0100145801.hg.1 NM_001126242.2 Refseq
Affymetrix Human Clariom D Assay PSR0100145801.hg.1 NM_001204184.1 Refseq
Affymetrix Human Clariom D Assay PSR0100145801.hg.1 NM_001204185.1 Refseq
Affymetrix Human Clariom D Assay PSR0100145801.hg.1 NM_001204187.1 Refseq
Affymetrix Human Clariom D Assay PSR0100145801.hg.1 NM_001204190.1 Refseq
Affymetrix Human Clariom D Assay PSR0100145801.hg.1 NM_001204192.1 Refseq
Affymetrix Human Clariom D Assay PSR0100145801.hg.1 NM_005427.3 Refseq
Affymetrix Human Clariom D Assay PSR0100145801.hg.1 TC0100006620.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nbrrd:80/100.66.77.69:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0