This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01056722_m1
See other IGSF6 GE Assays ›
Gene Symbol:
IGSF6
Gene Name:
immunoglobulin superfamily member 6
Gene Aliases:
DORA
Chromosome Location:
Chr.16: 21641284 - 21652666 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01056722_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IGSF6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_005849.3 NP_005840.2 2 - 3 491 75
GenBank mRNA AJ223183.1 - 2 - 3 467 75
AK223286.1 - 2 - 3 433 75
AK290910.1 - 2 - 3 433 75
AK313860.1 - 2 - 3 433 75
BC017844.1 - 2 - 3 433 4616809 75
CD369238.1 - 4 - 5 75

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1600009683.hg.1
This assay targets the same location as HTA transcript cluster(s) TC16000939.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene IGSF6

Panther Classification:

Molecular Function -

immunoglobulin

Gene Ontology Categories:

Function(s) Process(es)

immune response
cell surface receptor signaling pathway
transmembrane signaling receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR16013655.hg.1 ENST00000268389.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16013655.hg.1 NM_005849.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16013655.hg.1 TC16000939.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1600009683.hg.1 ENST00000268389.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009683.hg.1 ENST00000268389.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009683.hg.1 ENST00000268389.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009683.hg.1 ENST00000268389.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009683.hg.1 ENST00000268389.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009683.hg.1 ENST00000268389.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009683.hg.1 ENST00000268389.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009683.hg.1 ENST00000268389.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009683.hg.1 ENST00000268389.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009683.hg.1 ENST00000268389.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009683.hg.1 ENST00000565499.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009683.hg.1 ENST00000565499.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009683.hg.1 ENST00000565499.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009683.hg.1 ENST00000565499.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009683.hg.1 ENST00000569602.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009683.hg.1 ENST00000569602.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009683.hg.1 ENST00000569602.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009683.hg.1 NM_005849.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009683.hg.1 NM_005849.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009683.hg.1 NM_005849.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009683.hg.1 NM_005849.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009683.hg.1 NM_005849.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009683.hg.1 NM_005849.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009683.hg.1 NM_005849.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009683.hg.1 NM_005849.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009683.hg.1 NM_005849.3 Refseq
Affymetrix Human Clariom D Assay TC1600009683.hg.1 ENST00000268389.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009683.hg.1 ENST00000268389.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009683.hg.1 ENST00000268389.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009683.hg.1 ENST00000268389.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009683.hg.1 ENST00000268389.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009683.hg.1 ENST00000268389.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009683.hg.1 ENST00000268389.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009683.hg.1 ENST00000268389.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009683.hg.1 ENST00000268389.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009683.hg.1 ENST00000268389.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009683.hg.1 ENST00000565499.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009683.hg.1 ENST00000565499.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009683.hg.1 ENST00000565499.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009683.hg.1 ENST00000565499.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009683.hg.1 ENST00000569602.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009683.hg.1 ENST00000569602.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009683.hg.1 ENST00000569602.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009683.hg.1 NM_005849.3 Refseq
Affymetrix Human Clariom D Assay TC1600009683.hg.1 NM_005849.3 Refseq
Affymetrix Human Clariom D Assay TC1600009683.hg.1 NM_005849.3 Refseq
Affymetrix Human Clariom D Assay TC1600009683.hg.1 NM_005849.3 Refseq
Affymetrix Human Clariom D Assay TC1600009683.hg.1 NM_005849.3 Refseq
Affymetrix Human Clariom D Assay TC1600009683.hg.1 NM_005849.3 Refseq
Affymetrix Human Clariom D Assay TC1600009683.hg.1 NM_005849.3 Refseq
Affymetrix Human Clariom D Assay TC1600009683.hg.1 NM_005849.3 Refseq
Affymetrix Human Clariom D Assay TC1600009683.hg.1 NM_005849.3 Refseq
Affymetrix Human Clariom D Assay PSR1600174483.hg.1 ENST00000268389.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600174483.hg.1 ENST00000565499.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600174483.hg.1 NM_005849.3 Refseq
Affymetrix Human Clariom D Assay PSR1600174483.hg.1 ENST00000268389.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600174483.hg.1 ENST00000565499.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600174483.hg.1 NM_005849.3 Refseq
Affymetrix Human Clariom D Assay PSR1600174483.hg.1 TC1600009683.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1600174481.hg.1 ENST00000268389.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600174481.hg.1 ENST00000569602.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600174481.hg.1 NM_005849.3 Refseq
Affymetrix Human Clariom D Assay PSR1600174481.hg.1 ENST00000268389.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600174481.hg.1 ENST00000569602.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600174481.hg.1 NM_005849.3 Refseq
Affymetrix Human Clariom D Assay PSR1600174481.hg.1 TC1600009683.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0