This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01056908_m1
See other PLA1A GE Assays ›
Gene Symbol:
PLA1A
Gene Name:
phospholipase A1 member A
Gene Aliases:
PS-PLA1, PSPLA1
Chromosome Location:
Chr.3: 119597848 - 119629811 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01056908_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PLA1A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001206960.1 NP_001193889.1 2 - 3 340 69
NM_001293225.1 NP_001280154.1 2 - 3 335 69
NM_015900.3 NP_056984.1 2 - 3 340 69
NR_120610.1 2 - 3 340 69
XM_017006572.1 XP_016862061.1 3 - 4 372 69
GenBank mRNA AF035268.1 - 2 - 3 305 69
AF035269.1 - 2 - 3 305 69
AK222705.1 - 2 - 3 303 69
AK313519.1 - 2 - 3 303 69
BC035347.1 - 2 - 3 292 69
BC047703.2 - 2 - 3 292 6061929 69
BX647516.1 - 2 - 3 335 69
DB189568.1 - 2 - 3 341 69
DB255030.1 - 2 - 3 314 69
U37591.1 - 2 - 3 300 69

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300008484.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03000611.hg.1

Additional Information:

This assay is in the 5'UTR region of the XM_017006572.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PLA1A

Panther Classification:

Molecular Function -

lipase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
phosphatidylserine metabolic process
lipid catabolic process
phosphatidylcholine 1-acylhydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000472126.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000472126.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000472126.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000475963.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000475963.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000475963.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000475963.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000475963.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000475963.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000475963.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000475963.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline