This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01056914_m1
See other PLA1A GE Assays ›
Gene Symbol:
PLA1A
Gene Name:
phospholipase A1 member A
Gene Aliases:
PS-PLA1, PSPLA1
Chromosome Location:
Chr.3: 119597848 - 119629811 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01056914_m1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PLA1A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001206960.1 NP_001193889.1 8 - 9 1037 103
NM_001206961.1 NP_001193890.1 7 - 8 883 103
NM_001293225.1 NP_001280154.1 8 - 9 1080 103
NM_015900.3 NP_056984.1 8 - 9 1085 103
NR_120610.1 8 - 9 1085 103
XM_017006572.1 XP_016862061.1 9 - 10 1117 103
GenBank mRNA AF035268.1 - 8 - 9 1050 103
AF035269.1 - 8 - 9 1050 103
AK222705.1 - 8 - 9 1048 103
AK301880.1 - 7 - 8 856 103
AK313519.1 - 8 - 9 1048 103
BC047703.2 - 8 - 9 989 6061929 103
BX647516.1 - 8 - 9 1080 103
U37591.1 - 8 - 9 1045 103

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300008484.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03000611.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PLA1A

Panther Classification:

Molecular Function -

lipase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
phosphatidylserine metabolic process
lipid catabolic process
phosphatidylcholine 1-acylhydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000472126.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000472126.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000472126.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000475963.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000475963.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000475963.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000475963.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000475963.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000475963.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000475963.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000475963.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline