This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01056947_m1
See other NQO2 GE Assays ›
Gene Symbol:
NQO2
Gene Name:
NAD(P)H quinone dehydrogenase 2
Gene Aliases:
DHQV, DIA6, NMOR2, QR2
Chromosome Location:
Chr.6: 2999816 - 3019876 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01056947_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NQO2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000904.4 NP_000895.2 2 - 3 362 84
NM_001290221.1 NP_001277150.1 5 - 6 877 84
NM_001290222.1 NP_001277151.1 2 - 3 362 84
NM_001318940.1 NP_001305869.1 2 - 3 302 84
GenBank mRNA AK056981.1 - 5 - 6 719 84
AK311746.1 - 2 - 3 321 84
BC006096.2 - 1 - 2 93 3507945 84
BI333292.1 - 2 - 3 301 84
BI670476.1 - 2 - 3 350 84
CD014013.1 - 2 - 3 314 84
J02888.1 - 2 - 3 182 84

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0600014055.hg.1
This assay targets the same location as HTA transcript cluster(s) TC06004062.hg.1

Additional Information:

This assay is in the 5'UTR region of the AK056981.1,AK311746.1,BC006096.2,J02888.1,NM_000904.4,NM_001290221.1,NM_001290222.1,NM_001318940.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NQO2

Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s) Process(es)

memory
oxidation-reduction process
protein binding
NADPH dehydrogenase (quinone) activity
electron carrier activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 ENST00000338130.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 ENST00000338130.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 ENST00000338130.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 ENST00000338130.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 ENST00000338130.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 ENST00000338130.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 ENST00000338130.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 ENST00000338130.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 ENST00000338130.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 ENST00000338130.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 ENST00000338130.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 ENST00000338130.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 ENST00000380430.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 ENST00000380430.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 ENST00000380430.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 ENST00000380430.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 ENST00000380430.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 ENST00000380430.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 ENST00000380430.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 ENST00000380430.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 ENST00000380430.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 ENST00000380441.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 ENST00000380441.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 ENST00000380441.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 ENST00000380441.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 ENST00000380441.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 ENST00000380441.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 ENST00000380441.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 ENST00000380441.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 ENST00000380441.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 ENST00000380454.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 ENST00000380454.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 ENST00000380454.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 ENST00000380454.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 ENST00000380454.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 ENST00000380454.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 ENST00000380454.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 ENST00000380454.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 ENST00000397717.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 ENST00000397717.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 ENST00000397717.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 ENST00000397717.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 ENST00000397717.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 ENST00000397717.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 ENST00000397717.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 ENST00000397717.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 ENST00000606474.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 ENST00000606474.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 ENST00000606474.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 ENST00000606474.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 NM_000904.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 NM_000904.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 NM_000904.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 NM_000904.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 NM_000904.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 NM_000904.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 NM_000904.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 NM_000904.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 NM_000904.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 NM_000904.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 NM_000904.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 NM_000904.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 NM_001290221.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 NM_001290221.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 NM_001290221.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 NM_001290221.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 NM_001290221.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 NM_001290221.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 NM_001290221.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 NM_001290221.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 NM_001290221.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 NM_001290221.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 NM_001290221.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 NM_001290221.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 NM_001290221.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 NM_001290221.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 NM_001290221.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 NM_001290222.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 NM_001290222.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 NM_001290222.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 NM_001290222.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 NM_001290222.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 NM_001290222.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 NM_001290222.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 NM_001290222.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 NM_001290222.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 NM_001290222.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014055.hg.1 NM_001290222.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600006647.hg.1 ENST00000612789.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06000146.hg.1 TC06004062.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR06000146.hg.1 ENST00000338130.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06000146.hg.1 ENST00000380430.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06000146.hg.1 ENST00000380441.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06000146.hg.1 ENST00000380454.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06000146.hg.1 ENST00000380455.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06000146.hg.1 ENST00000538898.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06000146.hg.1 NM_000904.4 Refseq
Affymetrix Human Clariom D Assay PSR0600197167.hg.1 ENST00000338130.6 Ensembl
Affymetrix Human Clariom D Assay PSR0600197167.hg.1 ENST00000380430.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600197167.hg.1 ENST00000380441.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600197167.hg.1 ENST00000380454.8 Ensembl
Affymetrix Human Clariom D Assay PSR0600197167.hg.1 ENST00000397717.6 Ensembl
Affymetrix Human Clariom D Assay PSR0600197167.hg.1 ENST00000606474.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600197167.hg.1 NM_000904.4 Refseq
Affymetrix Human Clariom D Assay PSR0600197167.hg.1 NM_001290221.1 Refseq
Affymetrix Human Clariom D Assay PSR0600197167.hg.1 NM_001290222.1 Refseq
Affymetrix Human Clariom D Assay PSR0600197167.hg.1 ENST00000338130.6 Ensembl
Affymetrix Human Clariom D Assay PSR0600197167.hg.1 ENST00000380430.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600197167.hg.1 ENST00000380441.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600197167.hg.1 ENST00000380454.8 Ensembl
Affymetrix Human Clariom D Assay PSR0600197167.hg.1 ENST00000397717.6 Ensembl
Affymetrix Human Clariom D Assay PSR0600197167.hg.1 ENST00000606474.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600197167.hg.1 NM_000904.4 Refseq
Affymetrix Human Clariom D Assay PSR0600197167.hg.1 NM_001290221.1 Refseq
Affymetrix Human Clariom D Assay PSR0600197167.hg.1 NM_001290222.1 Refseq
Affymetrix Human Clariom D Assay PSR0600197167.hg.1 TC0600014055.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0600006647.hg.1 ENST00000612789.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014055.hg.1 ENST00000338130.6 Ensembl
Affymetrix Human Clariom D Assay TC0600014055.hg.1 ENST00000338130.6 Ensembl
Affymetrix Human Clariom D Assay TC0600014055.hg.1 ENST00000338130.6 Ensembl
Affymetrix Human Clariom D Assay TC0600014055.hg.1 ENST00000338130.6 Ensembl
Affymetrix Human Clariom D Assay TC0600014055.hg.1 ENST00000338130.6 Ensembl
Affymetrix Human Clariom D Assay TC0600014055.hg.1 ENST00000338130.6 Ensembl
Affymetrix Human Clariom D Assay TC0600014055.hg.1 ENST00000338130.6 Ensembl
Affymetrix Human Clariom D Assay TC0600014055.hg.1 ENST00000338130.6 Ensembl
Affymetrix Human Clariom D Assay TC0600014055.hg.1 ENST00000338130.6 Ensembl
Affymetrix Human Clariom D Assay TC0600014055.hg.1 ENST00000338130.6 Ensembl
Affymetrix Human Clariom D Assay TC0600014055.hg.1 ENST00000338130.6 Ensembl
Affymetrix Human Clariom D Assay TC0600014055.hg.1 ENST00000338130.6 Ensembl
Affymetrix Human Clariom D Assay TC0600014055.hg.1 ENST00000380430.5 Ensembl
Affymetrix Human Clariom D Assay TC0600014055.hg.1 ENST00000380430.5 Ensembl
Affymetrix Human Clariom D Assay TC0600014055.hg.1 ENST00000380430.5 Ensembl
Affymetrix Human Clariom D Assay TC0600014055.hg.1 ENST00000380430.5 Ensembl
Affymetrix Human Clariom D Assay TC0600014055.hg.1 ENST00000380430.5 Ensembl
Affymetrix Human Clariom D Assay TC0600014055.hg.1 ENST00000380430.5 Ensembl
Affymetrix Human Clariom D Assay TC0600014055.hg.1 ENST00000380430.5 Ensembl
Affymetrix Human Clariom D Assay TC0600014055.hg.1 ENST00000380430.5 Ensembl
Affymetrix Human Clariom D Assay TC0600014055.hg.1 ENST00000380430.5 Ensembl
Affymetrix Human Clariom D Assay TC0600014055.hg.1 ENST00000380441.5 Ensembl
Affymetrix Human Clariom D Assay TC0600014055.hg.1 ENST00000380441.5 Ensembl
Affymetrix Human Clariom D Assay TC0600014055.hg.1 ENST00000380441.5 Ensembl
Affymetrix Human Clariom D Assay TC0600014055.hg.1 ENST00000380441.5 Ensembl
Affymetrix Human Clariom D Assay TC0600014055.hg.1 ENST00000380441.5 Ensembl
Affymetrix Human Clariom D Assay TC0600014055.hg.1 ENST00000380441.5 Ensembl
Affymetrix Human Clariom D Assay TC0600014055.hg.1 ENST00000380441.5 Ensembl
Affymetrix Human Clariom D Assay TC0600014055.hg.1 ENST00000380441.5 Ensembl
Affymetrix Human Clariom D Assay TC0600014055.hg.1 ENST00000380441.5 Ensembl
Affymetrix Human Clariom D Assay TC0600014055.hg.1 ENST00000380454.8 Ensembl
Affymetrix Human Clariom D Assay TC0600014055.hg.1 ENST00000380454.8 Ensembl
Affymetrix Human Clariom D Assay TC0600014055.hg.1 ENST00000380454.8 Ensembl
Affymetrix Human Clariom D Assay TC0600014055.hg.1 ENST00000380454.8 Ensembl
Affymetrix Human Clariom D Assay TC0600014055.hg.1 ENST00000380454.8 Ensembl
Affymetrix Human Clariom D Assay TC0600014055.hg.1 ENST00000380454.8 Ensembl
Affymetrix Human Clariom D Assay TC0600014055.hg.1 ENST00000380454.8 Ensembl
Affymetrix Human Clariom D Assay TC0600014055.hg.1 ENST00000380454.8 Ensembl
Affymetrix Human Clariom D Assay TC0600014055.hg.1 ENST00000397717.6 Ensembl
Affymetrix Human Clariom D Assay TC0600014055.hg.1 ENST00000397717.6 Ensembl
Affymetrix Human Clariom D Assay TC0600014055.hg.1 ENST00000397717.6 Ensembl
Affymetrix Human Clariom D Assay TC0600014055.hg.1 ENST00000397717.6 Ensembl
Affymetrix Human Clariom D Assay TC0600014055.hg.1 ENST00000397717.6 Ensembl
Affymetrix Human Clariom D Assay TC0600014055.hg.1 ENST00000397717.6 Ensembl
Affymetrix Human Clariom D Assay TC0600014055.hg.1 ENST00000397717.6 Ensembl
Affymetrix Human Clariom D Assay TC0600014055.hg.1 ENST00000397717.6 Ensembl
Affymetrix Human Clariom D Assay TC0600014055.hg.1 ENST00000606474.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014055.hg.1 ENST00000606474.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014055.hg.1 ENST00000606474.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014055.hg.1 ENST00000606474.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014055.hg.1 NM_000904.4 Refseq
Affymetrix Human Clariom D Assay TC0600014055.hg.1 NM_000904.4 Refseq
Affymetrix Human Clariom D Assay TC0600014055.hg.1 NM_000904.4 Refseq
Affymetrix Human Clariom D Assay TC0600014055.hg.1 NM_000904.4 Refseq
Affymetrix Human Clariom D Assay TC0600014055.hg.1 NM_000904.4 Refseq
Affymetrix Human Clariom D Assay TC0600014055.hg.1 NM_000904.4 Refseq
Affymetrix Human Clariom D Assay TC0600014055.hg.1 NM_000904.4 Refseq
Affymetrix Human Clariom D Assay TC0600014055.hg.1 NM_000904.4 Refseq
Affymetrix Human Clariom D Assay TC0600014055.hg.1 NM_000904.4 Refseq
Affymetrix Human Clariom D Assay TC0600014055.hg.1 NM_000904.4 Refseq
Affymetrix Human Clariom D Assay TC0600014055.hg.1 NM_000904.4 Refseq
Affymetrix Human Clariom D Assay TC0600014055.hg.1 NM_000904.4 Refseq
Affymetrix Human Clariom D Assay TC0600014055.hg.1 NM_001290221.1 Refseq
Affymetrix Human Clariom D Assay TC0600014055.hg.1 NM_001290221.1 Refseq
Affymetrix Human Clariom D Assay TC0600014055.hg.1 NM_001290221.1 Refseq
Affymetrix Human Clariom D Assay TC0600014055.hg.1 NM_001290221.1 Refseq
Affymetrix Human Clariom D Assay TC0600014055.hg.1 NM_001290221.1 Refseq
Affymetrix Human Clariom D Assay TC0600014055.hg.1 NM_001290221.1 Refseq
Affymetrix Human Clariom D Assay TC0600014055.hg.1 NM_001290221.1 Refseq
Affymetrix Human Clariom D Assay TC0600014055.hg.1 NM_001290221.1 Refseq
Affymetrix Human Clariom D Assay TC0600014055.hg.1 NM_001290221.1 Refseq
Affymetrix Human Clariom D Assay TC0600014055.hg.1 NM_001290221.1 Refseq
Affymetrix Human Clariom D Assay TC0600014055.hg.1 NM_001290221.1 Refseq
Affymetrix Human Clariom D Assay TC0600014055.hg.1 NM_001290221.1 Refseq
Affymetrix Human Clariom D Assay TC0600014055.hg.1 NM_001290221.1 Refseq
Affymetrix Human Clariom D Assay TC0600014055.hg.1 NM_001290221.1 Refseq
Affymetrix Human Clariom D Assay TC0600014055.hg.1 NM_001290221.1 Refseq
Affymetrix Human Clariom D Assay TC0600014055.hg.1 NM_001290222.1 Refseq
Affymetrix Human Clariom D Assay TC0600014055.hg.1 NM_001290222.1 Refseq
Affymetrix Human Clariom D Assay TC0600014055.hg.1 NM_001290222.1 Refseq
Affymetrix Human Clariom D Assay TC0600014055.hg.1 NM_001290222.1 Refseq
Affymetrix Human Clariom D Assay TC0600014055.hg.1 NM_001290222.1 Refseq
Affymetrix Human Clariom D Assay TC0600014055.hg.1 NM_001290222.1 Refseq
Affymetrix Human Clariom D Assay TC0600014055.hg.1 NM_001290222.1 Refseq
Affymetrix Human Clariom D Assay TC0600014055.hg.1 NM_001290222.1 Refseq
Affymetrix Human Clariom D Assay TC0600014055.hg.1 NM_001290222.1 Refseq
Affymetrix Human Clariom D Assay TC0600014055.hg.1 NM_001290222.1 Refseq
Affymetrix Human Clariom D Assay TC0600014055.hg.1 NM_001290222.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-6dxlp:80/100.66.78.215:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0