This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01057073_m1
See other EPB41 GE Assays ›
Gene Symbol:
EPB41
Gene Name:
erythrocyte membrane protein band 4.1
Gene Aliases:
4.1R, EL1, HE
Chromosome Location:
Chr.1: 28887091 - 29120046 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01057073_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EPB41
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_004437.3 NP_004428.1 13 - 14 2030 127
XM_005245764.1 XP_005245821.1 12 - 13 2042 127
XM_011540960.1 XP_011539262.1 12 - 13 2042 127
XM_017000594.1 XP_016856083.1 12 - 13 1938 127
GenBank mRNA BC096105.1 - 11 - 12 1574 40001729 127
BC114450.1 - 11 - 12 1574 127
M14993.1 - 13 - 14 2011 127

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100007574.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01000394.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene EPB41

Gene Ontology Categories:

Function(s) Process(es)

blood circulation
actin cytoskeleton organization
cortical actin cytoskeleton organization
positive regulation of protein binding
actin binding
structural constituent of cytoskeleton
protein binding
calmodulin binding
1-phosphatidylinositol binding
spectrin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR01006077.hg.1 TC01000394.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01006077.hg.1 ENST00000343067.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01006077.hg.1 ENST00000349460.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01006077.hg.1 ENST00000356093.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01006077.hg.1 ENST00000373797.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01006077.hg.1 ENST00000373798.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01006077.hg.1 ENST00000373800.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01006077.hg.1 ENST00000398865.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01006077.hg.1 NM_001166005.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01006077.hg.1 NM_001166006.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01006077.hg.1 NM_004437.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01006077.hg.1 NM_203342.2 Refseq
Affymetrix Human Clariom D Assay PSR0100153180.hg.1 ENST00000343067.8 Ensembl
Affymetrix Human Clariom D Assay PSR0100153180.hg.1 ENST00000349460.8 Ensembl
Affymetrix Human Clariom D Assay PSR0100153180.hg.1 ENST00000356093.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100153180.hg.1 ENST00000373797.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100153180.hg.1 ENST00000373798.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100153180.hg.1 ENST00000373800.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100153180.hg.1 NM_001166005.1 Refseq
Affymetrix Human Clariom D Assay PSR0100153180.hg.1 NM_001166006.1 Refseq
Affymetrix Human Clariom D Assay PSR0100153180.hg.1 NM_004437.3 Refseq
Affymetrix Human Clariom D Assay PSR0100153180.hg.1 NM_203342.2 Refseq
Affymetrix Human Clariom D Assay PSR0100153180.hg.1 ENST00000343067.8 Ensembl
Affymetrix Human Clariom D Assay PSR0100153180.hg.1 ENST00000349460.8 Ensembl
Affymetrix Human Clariom D Assay PSR0100153180.hg.1 ENST00000356093.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100153180.hg.1 ENST00000373797.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100153180.hg.1 ENST00000373798.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100153180.hg.1 ENST00000373800.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100153180.hg.1 NM_001166005.1 Refseq
Affymetrix Human Clariom D Assay PSR0100153180.hg.1 NM_001166006.1 Refseq
Affymetrix Human Clariom D Assay PSR0100153180.hg.1 NM_004437.3 Refseq
Affymetrix Human Clariom D Assay PSR0100153180.hg.1 NM_203342.2 Refseq
Affymetrix Human Clariom D Assay PSR0100153180.hg.1 TC0100007574.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9hc65:80/100.66.78.230:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0