This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01057075_m1
See other EPB41 GE Assays ›
Gene Symbol:
EPB41
Gene Name:
erythrocyte membrane protein band 4.1
Gene Aliases:
4.1R, EL1, HE
Chromosome Location:
Chr.1: 28887091 - 29120046 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01057075_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EPB41
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001166005.1 NP_001159477.1 16 - 17 2311 101
NM_001166007.1 NP_001159479.1 13 - 14 2132 101
NM_004437.3 NP_004428.1 15 - 16 2275 101
NM_203342.2 NP_976217.1 16 - 17 2349 101
NM_203343.2 NP_976218.1 12 - 13 2044 101
XM_005245761.1 XP_005245818.1 16 - 17 2386 101
XM_005245763.1 XP_005245820.1 15 - 16 2344 101
XM_005245764.1 XP_005245821.1 14 - 15 2287 101
XM_005245765.1 XP_005245822.1 14 - 15 2281 101
XM_005245769.1 XP_005245826.1 13 - 14 2224 101
XM_005245770.1 XP_005245827.1 13 - 14 2221 101
XM_017000589.1 XP_016856078.1 12 - 13 2119 101
XM_017000590.1 XP_016856079.1 12 - 13 2116 101
GenBank mRNA AB209649.1 - 13 - 14 2132 101
AF156225.1 - 15 - 16 2184 101
AK299093.1 - 1 - 2 191 101
BC096104.1 - 10 - 11 1438 101
BC096105.1 - 13 - 14 1819 40001729 101
BC114450.1 - 13 - 14 1819 101
J03796.1 - 12 - 13 1963 101
M14993.1 - 15 - 16 2256 101
M61733.1 - 16 - 17 2330 101

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100007574.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01000394.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene EPB41

Gene Ontology Categories:

Function(s) Process(es)

blood circulation
actin cytoskeleton organization
cortical actin cytoskeleton organization
positive regulation of protein binding
actin binding
structural constituent of cytoskeleton
protein binding
calmodulin binding
1-phosphatidylinositol binding
spectrin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR01006084.hg.1 TC01000394.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01006084.hg.1 ENST00000343067.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01006084.hg.1 ENST00000347529.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01006084.hg.1 ENST00000349460.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01006084.hg.1 ENST00000356093.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01006084.hg.1 ENST00000358989.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01006084.hg.1 ENST00000373798.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01006084.hg.1 ENST00000373800.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01006084.hg.1 ENST00000398861.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01006084.hg.1 ENST00000398865.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01006084.hg.1 ENST00000460378.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01006084.hg.1 NM_001166005.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01006084.hg.1 NM_001166007.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01006084.hg.1 NM_004437.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01006084.hg.1 NM_203342.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01006084.hg.1 NM_203343.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01006085.hg.1 TC01000394.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01006085.hg.1 ENST00000343067.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01006085.hg.1 ENST00000347529.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01006085.hg.1 ENST00000349460.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01006085.hg.1 ENST00000356093.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01006085.hg.1 ENST00000358989.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01006085.hg.1 ENST00000373798.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01006085.hg.1 ENST00000373800.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01006085.hg.1 ENST00000398861.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01006085.hg.1 ENST00000398865.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01006085.hg.1 ENST00000460378.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01006085.hg.1 NM_001166005.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01006085.hg.1 NM_001166007.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01006085.hg.1 NM_004437.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01006085.hg.1 NM_203342.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01006085.hg.1 NM_203343.2 Refseq
Affymetrix Human Clariom D Assay PSR0100153187.hg.1 ENST00000343067.8 Ensembl
Affymetrix Human Clariom D Assay PSR0100153187.hg.1 ENST00000347529.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100153187.hg.1 ENST00000349460.8 Ensembl
Affymetrix Human Clariom D Assay PSR0100153187.hg.1 ENST00000356093.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100153187.hg.1 ENST00000373798.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100153187.hg.1 ENST00000373800.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100153187.hg.1 ENST00000460378.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100153187.hg.1 ENST00000462032.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100153187.hg.1 ENST00000488277.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100153187.hg.1 ENST00000490308.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100153187.hg.1 NM_001166005.1 Refseq
Affymetrix Human Clariom D Assay PSR0100153187.hg.1 NM_001166007.1 Refseq
Affymetrix Human Clariom D Assay PSR0100153187.hg.1 NM_004437.3 Refseq
Affymetrix Human Clariom D Assay PSR0100153187.hg.1 NM_203342.2 Refseq
Affymetrix Human Clariom D Assay PSR0100153187.hg.1 NM_203343.2 Refseq
Affymetrix Human Clariom D Assay PSR0100153187.hg.1 ENST00000343067.8 Ensembl
Affymetrix Human Clariom D Assay PSR0100153187.hg.1 ENST00000347529.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100153187.hg.1 ENST00000349460.8 Ensembl
Affymetrix Human Clariom D Assay PSR0100153187.hg.1 ENST00000356093.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100153187.hg.1 ENST00000373798.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100153187.hg.1 ENST00000373800.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100153187.hg.1 ENST00000460378.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100153187.hg.1 ENST00000462032.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100153187.hg.1 ENST00000488277.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100153187.hg.1 ENST00000490308.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100153187.hg.1 NM_001166005.1 Refseq
Affymetrix Human Clariom D Assay PSR0100153187.hg.1 NM_001166007.1 Refseq
Affymetrix Human Clariom D Assay PSR0100153187.hg.1 NM_004437.3 Refseq
Affymetrix Human Clariom D Assay PSR0100153187.hg.1 NM_203342.2 Refseq
Affymetrix Human Clariom D Assay PSR0100153187.hg.1 NM_203343.2 Refseq
Affymetrix Human Clariom D Assay PSR0100153187.hg.1 TC0100007574.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0100153188.hg.1 ENST00000343067.8 Ensembl
Affymetrix Human Clariom D Assay PSR0100153188.hg.1 ENST00000347529.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100153188.hg.1 ENST00000349460.8 Ensembl
Affymetrix Human Clariom D Assay PSR0100153188.hg.1 ENST00000356093.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100153188.hg.1 ENST00000373798.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100153188.hg.1 ENST00000373800.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100153188.hg.1 ENST00000460378.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100153188.hg.1 ENST00000462032.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100153188.hg.1 ENST00000490308.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100153188.hg.1 NM_001166005.1 Refseq
Affymetrix Human Clariom D Assay PSR0100153188.hg.1 NM_001166007.1 Refseq
Affymetrix Human Clariom D Assay PSR0100153188.hg.1 NM_004437.3 Refseq
Affymetrix Human Clariom D Assay PSR0100153188.hg.1 NM_203342.2 Refseq
Affymetrix Human Clariom D Assay PSR0100153188.hg.1 NM_203343.2 Refseq
Affymetrix Human Clariom D Assay PSR0100153188.hg.1 ENST00000343067.8 Ensembl
Affymetrix Human Clariom D Assay PSR0100153188.hg.1 ENST00000347529.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100153188.hg.1 ENST00000349460.8 Ensembl
Affymetrix Human Clariom D Assay PSR0100153188.hg.1 ENST00000356093.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100153188.hg.1 ENST00000373798.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100153188.hg.1 ENST00000373800.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100153188.hg.1 ENST00000460378.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100153188.hg.1 ENST00000462032.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100153188.hg.1 ENST00000490308.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100153188.hg.1 NM_001166005.1 Refseq
Affymetrix Human Clariom D Assay PSR0100153188.hg.1 NM_001166007.1 Refseq
Affymetrix Human Clariom D Assay PSR0100153188.hg.1 NM_004437.3 Refseq
Affymetrix Human Clariom D Assay PSR0100153188.hg.1 NM_203342.2 Refseq
Affymetrix Human Clariom D Assay PSR0100153188.hg.1 NM_203343.2 Refseq
Affymetrix Human Clariom D Assay PSR0100153188.hg.1 TC0100007574.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4krp5:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0