This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01057668_m1
See other PNLIP GE Assays ›
Gene Symbol:
PNLIP
Gene Name:
pancreatic lipase
Gene Aliases:
PL, PNLIPD, PTL
Chromosome Location:
Chr.10: 116545916 - 116567855 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01057668_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PNLIP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA A24059.1 - 2 - 3 301 139
AK313941.1 - 4 - 5 353 139
BC014309.2 - 3 - 4 325 4042766 139
J05125.1 - 3 - 4 337 139
M93285.1 - 3 - 4 325 139

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000009004.hg.1

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

lipase

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 ENST00000470562.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 ENST00000470562.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 ENST00000470562.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 ENST00000470562.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 ENST00000470562.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Clariom D Assay PSR1000161629.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Clariom D Assay PSR1000161629.hg.1 ENST00000470562.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000161629.hg.1 NM_000936.3 Refseq
Affymetrix Human Clariom D Assay PSR1000161629.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Clariom D Assay PSR1000161629.hg.1 ENST00000470562.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000161629.hg.1 NM_000936.3 Refseq
Affymetrix Human Clariom D Assay PSR1000161629.hg.1 TC1000009004.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1000161630.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Clariom D Assay PSR1000161630.hg.1 ENST00000470562.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000161630.hg.1 NM_000936.3 Refseq
Affymetrix Human Clariom D Assay PSR1000161630.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Clariom D Assay PSR1000161630.hg.1 ENST00000470562.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000161630.hg.1 NM_000936.3 Refseq
Affymetrix Human Clariom D Assay PSR1000161630.hg.1 TC1000009004.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009004.hg.1 ENST00000470562.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009004.hg.1 ENST00000470562.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009004.hg.1 ENST00000470562.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009004.hg.1 ENST00000470562.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009004.hg.1 ENST00000470562.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Clariom D Assay TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Clariom D Assay TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Clariom D Assay TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Clariom D Assay TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Clariom D Assay TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Clariom D Assay TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Clariom D Assay TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Clariom D Assay TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Clariom D Assay TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Clariom D Assay TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Clariom D Assay TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Clariom D Assay TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Clariom D Assay TC1000009004.hg.1 NM_000936.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0