This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01057672_m1
See other PNLIP GE Assays ›
Gene Symbol:
PNLIP
Gene Name:
pancreatic lipase
Gene Aliases:
PL, PNLIPD, PTL
Chromosome Location:
Chr.10: 116545916 - 116567855 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01057672_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PNLIP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA A24059.1 - 8 - 9 1032 97
AK313941.1 - 10 - 11 1084 97
BC014309.2 - 9 - 10 1056 4042766 97
J05125.1 - 9 - 10 1068 97
M93285.1 - 9 - 10 1056 97

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000009004.hg.1

Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

lipase

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 ENST00000470562.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 ENST00000470562.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 ENST00000470562.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 ENST00000470562.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 ENST00000470562.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Clariom D Assay TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009004.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009004.hg.1 ENST00000470562.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009004.hg.1 ENST00000470562.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009004.hg.1 ENST00000470562.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009004.hg.1 ENST00000470562.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009004.hg.1 ENST00000470562.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Clariom D Assay TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Clariom D Assay TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Clariom D Assay TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Clariom D Assay TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Clariom D Assay TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Clariom D Assay TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Clariom D Assay TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Clariom D Assay TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Clariom D Assay TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Clariom D Assay TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Clariom D Assay TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Clariom D Assay TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Clariom D Assay TC1000009004.hg.1 NM_000936.3 Refseq
Affymetrix Human Clariom D Assay PSR1000161637.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Clariom D Assay PSR1000161637.hg.1 NM_000936.3 Refseq
Affymetrix Human Clariom D Assay PSR1000161637.hg.1 ENST00000369221.2 Ensembl
Affymetrix Human Clariom D Assay PSR1000161637.hg.1 NM_000936.3 Refseq
Affymetrix Human Clariom D Assay PSR1000161637.hg.1 TC1000009004.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0