This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01058303_m1
See other ACMSD GE Assays ›
Gene Symbol:
ACMSD
Gene Name:
aminocarboxymuconate semialdehyde decarboxylase
Gene Aliases:
-
Chromosome Location:
Chr.2: 134838319 - 134902906 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01058303_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ACMSD
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_138326.2 NP_612199.2 1 - 2 130 132
GenBank mRNA AK125008.1 - 1 - 2 130 132

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200009424.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02000877.hg.1

Additional Information:

This assay is in the 5'UTR region of the AK125008.1,NM_138326.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ACMSD

Panther Classification:

Molecular Function -

decarboxylase

Gene Ontology Categories:

Function(s) Process(es)

tryptophan catabolic process
quinolinate metabolic process
protein oligomerization
regulation of quinolinate biosynthetic process
picolinic acid biosynthetic process
regulation of 'de novo' NAD biosynthetic process from tryptophan
aminocarboxymuconate-semialdehyde decarboxylase activity
protein binding
zinc ion binding
hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR02012935.hg.1 TC02000877.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR02012935.hg.1 ENST00000283054.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02012935.hg.1 ENST00000356140.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02012935.hg.1 ENST00000392928.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02012935.hg.1 NM_138326.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000485893.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000485893.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000485893.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000485893.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000485893.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000485893.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000485893.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000485893.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000485893.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000485893.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000498093.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000498093.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000498093.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000498093.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_001307983.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_001307983.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_001307983.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_001307983.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_001307983.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_001307983.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_001307983.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_001307983.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_001307983.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_001307983.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_001307983.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_001307983.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_001307983.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_138326.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_138326.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_138326.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_138326.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_138326.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_138326.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_138326.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_138326.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_138326.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_138326.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_138326.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_138326.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02012938.hg.1 TC02000877.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR02012938.hg.1 ENST00000283054.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02012938.hg.1 ENST00000356140.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02012938.hg.1 ENST00000392928.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02012938.hg.1 NM_138326.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02012934.hg.1 TC02000877.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR02012934.hg.1 ENST00000283054.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02012934.hg.1 ENST00000356140.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02012934.hg.1 ENST00000392928.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02012934.hg.1 NM_138326.2 Refseq
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000485893.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000485893.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000485893.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000485893.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000485893.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000485893.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000485893.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000485893.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000485893.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000485893.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000498093.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000498093.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000498093.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000498093.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_001307983.1 Refseq
Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_001307983.1 Refseq
Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_001307983.1 Refseq
Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_001307983.1 Refseq
Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_001307983.1 Refseq
Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_001307983.1 Refseq
Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_001307983.1 Refseq
Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_001307983.1 Refseq
Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_001307983.1 Refseq
Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_001307983.1 Refseq
Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_001307983.1 Refseq
Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_001307983.1 Refseq
Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_001307983.1 Refseq
Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_138326.2 Refseq
Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_138326.2 Refseq
Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_138326.2 Refseq
Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_138326.2 Refseq
Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_138326.2 Refseq
Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_138326.2 Refseq
Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_138326.2 Refseq
Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_138326.2 Refseq
Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_138326.2 Refseq
Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_138326.2 Refseq
Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_138326.2 Refseq
Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_138326.2 Refseq
Affymetrix Human Clariom D Assay PSR0200165884.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Clariom D Assay PSR0200165884.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200165884.hg.1 ENST00000485893.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200165884.hg.1 ENST00000498093.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200165884.hg.1 NM_001307983.1 Refseq
Affymetrix Human Clariom D Assay PSR0200165884.hg.1 NM_138326.2 Refseq
Affymetrix Human Clariom D Assay PSR0200165884.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Clariom D Assay PSR0200165884.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200165884.hg.1 ENST00000485893.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200165884.hg.1 ENST00000498093.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200165884.hg.1 NM_001307983.1 Refseq
Affymetrix Human Clariom D Assay PSR0200165884.hg.1 NM_138326.2 Refseq
Affymetrix Human Clariom D Assay PSR0200165884.hg.1 TC0200009424.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0