This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01059049_m1
See other PLCE1 GE Assays ›
Gene Symbol:
PLCE1
Gene Name:
phospholipase C epsilon 1
Gene Aliases:
NPHS3, PLCE, PPLC
Chromosome Location:
Chr.10: 93993989 - 94329447 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01059049_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PLCE1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001165979.2 NP_001159451.1 22 - 23 4316 93
NM_001288989.1 NP_001275918.1 23 - 24 5757 93
NM_016341.3 NP_057425.3 23 - 24 5805 93
XM_006717885.3 XP_006717948.1 24 - 25 5795 93
XM_006717888.3 XP_006717951.1 24 - 25 5792 93
XM_006717889.3 XP_006717952.1 24 - 25 5747 93
XM_006717890.2 XP_006717953.1 23 - 24 4311 93
XM_011539849.2 XP_011538151.1 24 - 25 6952 93
XM_011539850.2 XP_011538152.1 23 - 24 5440 93
XM_017016310.1 XP_016871799.1 25 - 26 5842 93
XM_017016311.1 XP_016871800.1 24 - 25 5795 93
XM_017016312.1 XP_016871801.1 22 - 23 4221 93
GenBank mRNA AB040949.2 - 23 - 24 5805 93
AF117948.1 - 14 - 15 1880 93
AF170071.1 - 22 - 23 4247 93
AF190642.2 - 22 - 23 5406 93
AK074239.1 - 2 - 3 192 93
BC140705.1 - 22 - 23 5231 9021573 93
BC144286.1 - 22 - 23 5183 9052806 93
BC151854.1 - 23 - 24 5805 8860521 93

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000008495.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10000670.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PLCE1

Panther Classification:

Molecular Function -

phospholipase

Gene Ontology Categories:

Function(s) Process(es)

activation of MAPK activity
regulation of cell growth
phospholipid metabolic process
diacylglycerol biosynthetic process
regulation of smooth muscle contraction
cytoskeleton organization
epidermal growth factor receptor signaling pathway
phospholipase C-activating G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
protein kinase C-activating G-protein coupled receptor signaling pathway
Ras protein signal transduction
heart development
regulation of G-protein coupled receptor protein signaling pathway
cell proliferation
lipid catabolic process
calcium-mediated signaling
glomerulus development
positive regulation of GTPase activity
inositol phosphate metabolic process
regulation of protein kinase activity
regulation of Ras protein signal transduction
inositol phosphate-mediated signaling
phosphatidylinositol phospholipase C activity
phospholipase C activity
receptor signaling protein activity
guanyl-nucleotide exchange factor activity
protein binding
Ras GTPase binding
enzyme binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR10007596.hg.1 ENST00000260766.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10007596.hg.1 ENST00000371375.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10007596.hg.1 ENST00000371380.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10007596.hg.1 ENST00000371385.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10007596.hg.1 NM_001165979.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10007596.hg.1 NM_016341.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10007596.hg.1 TC10000670.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1000157840.hg.1 ENST00000260766.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000157840.hg.1 ENST00000371375.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000157840.hg.1 ENST00000371380.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000157840.hg.1 ENST00000371385.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000157840.hg.1 NM_001165979.2 Refseq
Affymetrix Human Clariom D Assay PSR1000157840.hg.1 NM_001288989.1 Refseq
Affymetrix Human Clariom D Assay PSR1000157840.hg.1 NM_016341.3 Refseq
Affymetrix Human Clariom D Assay PSR1000157840.hg.1 ENST00000260766.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000157840.hg.1 ENST00000371375.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000157840.hg.1 ENST00000371380.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000157840.hg.1 ENST00000371385.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000157840.hg.1 NM_001165979.2 Refseq
Affymetrix Human Clariom D Assay PSR1000157840.hg.1 NM_001288989.1 Refseq
Affymetrix Human Clariom D Assay PSR1000157840.hg.1 NM_016341.3 Refseq
Affymetrix Human Clariom D Assay PSR1000157840.hg.1 TC1000008495.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0