This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01059459_m1
See other SV2A GE Assays ›
Gene Symbol:
SV2A
Gene Name:
synaptic vesicle glycoprotein 2A
Gene Aliases:
SV2
Chromosome Location:
Chr.1: 149903318 - 149917882 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01059459_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SV2A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_014849.4 NP_055664.3 1 - 2 151 68
GenBank mRNA AB018279.1 - 1 - 2 94 68
AK075480.1 - 1 - 2 111 68
AK291718.1 - 1 - 2 100 68
BC045111.1 - 1 - 2 97 5804662 68
BP214143.1 - 1 - 2 140 68
BX537754.1 - 1 - 2 147 68
DA334809.1 - 1 - 2 151 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100015714.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01003195.hg.1

Additional Information:

This assay is in the 5'UTR region of the AB018279.1,AK075480.1,AK291718.1,BC045111.1,BX537754.1,NM_014849.4 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SV2A

Gene Ontology Categories:

Function(s) Process(es)

neurotransmitter transport
cellular calcium ion homeostasis
transmembrane transport
receptor activity
protein kinase binding
transmembrane transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR01048950.hg.1 TC01003195.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01048950.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01048950.hg.1 ENST00000369146.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01048950.hg.1 NM_014849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01048951.hg.1 TC01003195.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01048951.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01048951.hg.1 ENST00000369146.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01048951.hg.1 NM_014849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01048953.hg.1 TC01003195.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01048953.hg.1 ENST00000369146.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01048953.hg.1 NM_014849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 NM_001278719.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 NM_001278719.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 NM_001278719.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 NM_001278719.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 NM_001278719.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 NM_001278719.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Clariom D Assay PSR0100217643.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100217643.hg.1 NM_014849.4 Refseq
Affymetrix Human Clariom D Assay PSR0100217643.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100217643.hg.1 NM_014849.4 Refseq
Affymetrix Human Clariom D Assay PSR0100217643.hg.1 TC0100015714.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0100217640.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100217640.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100217640.hg.1 NM_014849.4 Refseq
Affymetrix Human Clariom D Assay PSR0100217640.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100217640.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100217640.hg.1 NM_014849.4 Refseq
Affymetrix Human Clariom D Assay PSR0100217640.hg.1 TC0100015714.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0100217641.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100217641.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100217641.hg.1 NM_014849.4 Refseq
Affymetrix Human Clariom D Assay PSR0100217641.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100217641.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100217641.hg.1 NM_014849.4 Refseq
Affymetrix Human Clariom D Assay PSR0100217641.hg.1 TC0100015714.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 NM_001278719.1 Refseq
Affymetrix Human Clariom D Assay TC0100015714.hg.1 NM_001278719.1 Refseq
Affymetrix Human Clariom D Assay TC0100015714.hg.1 NM_001278719.1 Refseq
Affymetrix Human Clariom D Assay TC0100015714.hg.1 NM_001278719.1 Refseq
Affymetrix Human Clariom D Assay TC0100015714.hg.1 NM_001278719.1 Refseq
Affymetrix Human Clariom D Assay TC0100015714.hg.1 NM_001278719.1 Refseq
Affymetrix Human Clariom D Assay TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Clariom D Assay TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Clariom D Assay TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Clariom D Assay TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Clariom D Assay TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Clariom D Assay TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Clariom D Assay TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Clariom D Assay TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Clariom D Assay TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Clariom D Assay TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Clariom D Assay TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Clariom D Assay TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Clariom D Assay TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Clariom D Assay TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Clariom D Assay TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Clariom D Assay TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Clariom D Assay TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Clariom D Assay PSR0100217639.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100217639.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100217639.hg.1 NM_014849.4 Refseq
Affymetrix Human Clariom D Assay PSR0100217639.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100217639.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100217639.hg.1 NM_014849.4 Refseq
Affymetrix Human Clariom D Assay PSR0100217639.hg.1 TC0100015714.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0