This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01059539_s1
See other GJB3 GE Assays ›
Gene Symbol:
GJB3
Gene Name:
gap junction protein beta 3
Gene Aliases:
CX31, DFNA2, DFNA2B, EKV
Chromosome Location:
Chr.1: 34781189 - 34786366 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01059539_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GJB3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AF052692.1 - 1 3561 82
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AL832611.1 - 1 Chr. 1 - 34783031 - 34788133 [-] on Build GRCh38 82

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream gap junction protein beta 4 GJB4 CX30.3,
EKV
-19.3kb Anti-sense Chr. 1 - 34759741 - 34763724 on NCBI Build GRCh38
Downstream gap junction protein alpha 4 GJA4 CX37 4.8kb Anti-sense Chr. 1 - 34792958 - 34795747 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

gap junction

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline