This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01059951_m1
See other PPP6R3 GE Assays ›
Gene Symbol:
PPP6R3
Gene Name:
protein phosphatase 6 regulatory subunit 3
Gene Aliases:
C11orf23, PP6R3, SAP190, SAPL, SAPLa, SAPS3
Chromosome Location:
Chr.11: 68460712 - 68615334 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01059951_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPP6R3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001164164.1 NP_001157636.1 10 - 11 1346 148
NM_018312.4 NP_060782.2 10 - 11 1346 148
XM_006718622.3 XP_006718685.1 10 - 11 1357 148
XM_006718623.3 XP_006718686.1 10 - 11 1340 148
XM_006718624.2 XP_006718687.1 10 - 11 1357 148
XM_011545145.2 XP_011543447.1 10 - 11 1357 148
XM_017017976.1 XP_016873465.1 9 - 10 1240 148
GenBank mRNA AF264779.1 - 9 - 10 1251 148
AF264780.1 - 9 - 10 1251 148

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PPP6R3

Panther Classification:

Molecular Function -

phosphatase modulator

Gene Ontology Categories:

Function(s) Process(es)

regulation of phosphoprotein phosphatase activity
COPII vesicle coating
protein binding
protein phosphatase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0