This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01060256_m1
See other CHD1 GE Assays ›
Gene Symbol:
CHD1
Gene Name:
chromodomain helicase DNA binding protein 1
Gene Aliases:
-
Chromosome Location:
Chr.5: 98855204 - 98929014 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01060256_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CHD1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001270.2 NP_001261.2 24 - 25 3588 124
XM_005271866.4 XP_005271923.1 25 - 26 4064 124
XM_005271867.4 XP_005271924.1 25 - 26 4064 124
XM_011543112.2 XP_011541414.1 25 - 26 4064 124
XM_017008991.1 XP_016864480.1 9 - 10 1124 124
XM_017008992.1 XP_016864481.1 9 - 10 969 124
GenBank mRNA AF006513.1 - 24 - 25 3603 124
BC117134.1 - 24 - 25 3578 40125685 124

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0500011554.hg.1
This assay targets the same location as HTA transcript cluster(s) TC05001628.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CHD1

Gene Ontology Categories:

Function(s) Process(es)

chromatin remodeling
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
covalent chromatin modification
DNA duplex unwinding
positive regulation by host of viral transcription
DNA binding
ATP-dependent DNA helicase activity
protein binding
ATP binding
methylated histone binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0